Search
Search

ap2s1| Gene Symbol: ap2s1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001016325.2 | NP_001016325.1 | 4 - 5 | 345 | 115 | |
| GenBank mRNA | BC170868.1 | - | 4 - 5 | 345 | 115 | |
| BC171019.1 | - | 4 - 5 | 357 | 115 | ||
| CR761434.2 | - | 4 - 5 | 345 | 115 | ||
Set Membership: |
|