Search
Search

crnkl1| Gene Symbol: crnkl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001016895.2 | NP_001016895.1 | 11 - 12 | 1575 | 108 | |
| GenBank mRNA | BC155382.1 | - | 10 - 11 | 1748 | 108 | |
| CR848544.2 | - | 11 - 12 | 1575 | 108 | ||
Set Membership: |
|