Search
Search
| Gene Symbol: Gck | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001270849.1 | NP_001257778.1 | 5 - 6 | 691 | 58 | |
| NM_001270850.1 | NP_001257779.1 | 4 - 5 | 469 | 58 | ||
| NM_012565.2 | NP_036697.1 | 4 - 5 | 592 | 58 | ||
| XM_006251179.2 | XP_006251241.1 | 4 - 5 | 1437 | 58 | ||
| GenBank mRNA | J04218.1 | - | 4 - 5 | 551 | 58 | |
| KJ026953.1 | - | 4 - 5 | 484 | 58 | ||
| M25807.1 | - | 4 - 5 | 469 | 58 | ||
| X53588.1 | - | 4 - 5 | 575 | 58 | ||
| X53589.1 | - | 5 - 6 | 691 | 58 | ||
Set Membership: |
|