Search
dataTaker loggers provide support to read vibrating wire strain gauge and electromechanical sensors which are common within geotechnical applications. This logger is suitable to be used in geotechnical applications such as slope stability, subsidence, dam wall monitoring, tunnel and mining excavation, ground water, tunnel wall monitoring, and site assessment. Either as standalone loggers or part of a network with powerful inbuilt communication options, dataTaker loggers are ready to be deployed for geotechnical monitoring.
| DT80G | DT85G Series |
|---|---|
![]() | ![]() |
| Intermediate data logger | Advanced data logger |
| - | Options for integrated modem |
| 5 analog input channels, expandable to 300 | 16 analog input channels, expandable to 960 |
| 8 digital input/counter channels | |
| ~10,000,000 data points | |
| USB port | |
| 2 serial 'smart sensor' ports | |
| Web and FTP client, RS232/485 | |
| SDI12 | |
| Vibrating wire strain gauge support | |
Increasing the channel capacity of the dataTaker DT80 and DT85 range is made very easy by adding dataTaker Channel Expansion Modules (CEM20). Each CEM20 connects 20 universal data logging channels to the dataTaker data logger.
dataTaker enclosures give you the flexibility to install your dataTaker system in a harsh or otherwise unsuitable environment by providing protection against external conditions such as water, dust, and accidental damage. These enclosures are suitable for all dataTaker systems and can be used for, industrial, weatherproof, or portable applications.