Search
Find replacement consumables specific to your system.
Your lab water purification system is designed to be easy to use and maintain. In order to keep your system performing at its best, certain components should be replaced on a regularly scheduled basis.
Select your system below to review available consumables, the recommended replacement schedules, functions and specifications.
| Description | Catalog Number |
Estimated Lifetime |
Impurities Removed |
Effects if Not Replaced |
| Ultrapure Cartridge | 09.2005 | Up to 6-12 months depending on feed water quality |
Ions | *Low resistivity/high conductivity in water |
| 0.2um Final Filter | 09.1003 | Up to 3-12 months |
Bacteria |
*Interference in analytical method *Bacteria and particles in the water *Slow dispense flow rate |
| UV Lamp UV or UV/UF units only |
09.2002 | Up to 2 years |
Organics Bacteria |
*Increased TOC causing problems in analytical methods and biological methods *Bacteria in system |
| 50133980 | Up to 2 years |
Endotoxins |
*Cell culture growth retarded or cell death *Degradation of DNA and RNA |
|
| CMX25 | N/A |
Bacteria | *Bacteria in water system *Slow dispense flow rate |
| Description | Catalog Number | Estimated Llfetime * | Impurities removed | Effects if not replaced |
| Smart2Pure Pro RO membrane | 50157608 | Up to 12 months | General removal of: Ions Organics Bacteria |
*Increased ion, organic and bacteria impurities in water. *Ultrapure cartridge exhausting quickly |
| Ultrapure cartridge | 09.1020 | Up to 12 months | Ions | *Low resistivity/high conductivity in water *Interference in analytical methods, molecular biology and cell culture |
| 0.1 um filter | 50157375 | Up to 3 -12 months | Bacteria Particulates |
*Bacteria and particles in the water *Mycoplasma contamination *Slow dispense flow rate |
| 0.2 um filter for system and hand remote dispenser | 09.1003 | Up to 12 months | Bacteria Particulates |
*Interference in analytical method *Bacteria and particles in the water *Slow dispense flow rate |
| UV Lamp | 09.1002 | When applicable -Up to 2 years | Organics Bacteria |
*Increased TOC causing problems in analytical methods and biological methods *Bacteria in system |
| Ultrafilter | 50133980 | Up to 2 years |
Endotoxins Nucleases Bacteria |
*Cell culture growth retarded or cell death *Degradation of DNA and RNA |
| Storage reservoir CO2 vent filter | 06.5002 | Up to 12 months | Airborne particulates CO2 and vapors |
*Contaimination of stored water cause water system to work harder |
| Description | Catalog Number | Estimated Lifetime * | Impurities removed | Effects if not replaced |
| RO membrane for Smart2Pure 3 LPH Pretreatment Cartridge | 09.2003 | Up to 12 months | General removal of: Ions Organics Bacteria Particles |
*Increased ion, organic and bacteria impurities in water. *Ultrapure cartridge exhausting quickly |
| RO membrane for Smart2Pure 6 LPH Pretreatment Cartridge | 09.2006 | Up to 12 months | General removal of: Ions Organics Bacteria Particles |
*Increased ion, organic and bacteria impurities in water. *Ultrapure cartridge exhausting quickly |
| Ultrapure cartridge | 09.1020 | Up to 12 months | Ions | *Low resistivity/high conductivity in water *Interference in analytical methods, molecular biology and cell culture |
| 0.2 um filter for system and hand remote dispenser | 09.1003 | Up to 3 -12 months | Bacteria Particulates |
*Interference in analytical method *Bacteria and particles in the water *Slow dispense flow rate |
| UV Lamp for system UV or UV/UF units only |
09.1002 | Up to 2 years |
Organics Bacteria |
*Increased TOC causing problems in analytical methods and biological methods *Bacteria in system |
| Ultrafilter (For UF or UV/UF units only) |
50133981 | Up to 2 years |
Endotoxins Nucleases Bacteria |
*Cell culture growth retarded or cell death *Degradation of DNA and RNA |
| Storage reservoir vent filter | 22.0091 | Airborne particulates | *Contaimination of stored water cause water system to work harder |
| Description | Catalog Number | Eestimated Lifetime * | Impurities removed | Effects if not replaced |
|---|---|---|---|---|
| Smart2Pure 12 LPH Pretreatment Cartridge (RO membrane) | 09.2012 | Up to 12 months | General removal of: Ions Organics Bacteria Particles |
*Increased ion, organic and bacteria impurities in water. *Ultrapure cartridge exhausting quickly |
| Ultrapure cartridge | 09.1020 | Up to 12 months | Ions | *Low resistivity/high conductivity in water *Interference in analytical methods, molecular biology and cell culture |
| 0.2 um filter for system and hand remote dispenser | 09.1003 | Up to 3 -12 months | Bacteria Particulates |
*Interference in analytical method *Bacteria and particles in the water *Slow dispense flow rate |
| UV Lamp for Type I system UV or UV/UF units only |
09.1002 | Up to 2 years |
Organics Bacteria |
*Increased TOC causing problems in analytical methods and biological methods *Bacteria in system |
| Ultrafilter For UF or UV/UF units only | 50133981 | Up to 2 years |
Endotoxins Nucleases Bacteria |
*Cell culture growth retarded or cell death *Degradation of DNA and RNA |
| Storage reservoir CO2 vent filter | 06.5002 | Up to 12 months | Airborne particulates CO2 and vapors |
*Contamination of stored water cause water system to work harder |
| Description | Catalog Number |
Estimated Lifetime |
Impurities Removed |
Effects if Not Replaced |
| LabTower EDI 15 RO membrane (Requires 2) |
22.0046 |
Up to 2 years |
General removal of: Ions Organics Bacteria Particles |
*Increased ion, organic and bacteria impurities in water. |
| LabTower EDI 30 RO membrane (Requires 2) |
22.0087 |
Up to 2 years |
General removal of: |
*Increased ion, organic and bacteria impurities in water. *Ultrapure cartridge exhausting quickly |
| Ultrapure Cartridge | 09.2005 | Up to 6-12 months depending on feed water quality |
Ions | *Low resistivity/high conductivity in water *Interference in analytical methods, molecular biology and cell culture |
Sterile 0.2 micron final filter for system and remote dispenser option |
09.1003 | 3 -6 months |
Bacteria Particulates |
*Interference in analytical method *Bacteria and particles in the water *Slow dispense flow rate |
| 09.2002 | when applicable - up to 2 years |
Organics Bacteria |
*Increased TOC causing problems in analytical methods and biological methods *Bacteria in system |
|
| CO2 vent filter | 09.1003 | Up to 12 months | Airborne particulates CO2 and vapors |
*Contamination of stored water cause water system to work harder |
| Description | Catalog Number | Estimated Lifetime* | Impurities Removed | Effects if Not Replaced |
|---|---|---|---|---|
| Ultrapure Cartridge | 09.1006 | Up to 6-12 months depending on feed water quality | Ions | *Low resistivity/high conductivity in water *Interference in analytical methods, molecular biology and cell culture |
| 0.2um Final Filter | 09.1003 | Up to 3 -12 months | Bacteria Particulates |
*Interference in analytical method *Bacteria and particles in the water *Slow dispense flow rate |
| UV Lamp UV or UV/UF units only |
09.1002 | When applicable -Up to two years | Organics Bacteria |
*Increased TOC causing problems in analytical methods and biological methods *Bacteria in system |
| Ultrafilter (For UF or UV/UF units only) |
50133981 | When applicable -Up to two years | Endotoxins Nucleases Bacteria |
*Cell culture growth retarded or cell death *Degradation of DNA and RNA |
| Sterile Vent Filter - MicroPure ST units only | 22.0091 | When applicable - Up to 6-12 months |
Airborne particulates | *Contamination of stored water cause water system to work harder |
| Cleaning Solution | CMX25 | N/A | Bacteria | *Bacteria in water system *Slow dispense flow rate |
| Description | Catalog Number |
Estimated Lifetime |
Impurities Removed |
Effects if Not Replaced |
| Ultrapure Cartridge | 09.1006 |
Up to 6-12 months depending on feed water quality |
Ions | *Low resistivity/high conductivity water |
| 0.2um Final Filter | 09.1003 | Up to 3-12 months |
Bacteria |
*Interference in analytical method *Bacteria and particles in the water *Slow dispense flow rate |
| UV Lamp |
09.1002 |
Up to 2 years |
Organics Bacteria |
*High TOC causing problems in analytical methods and biological methods |
| 50133981 | Up to 2 years |
Particles Bacteria |
*Particles in water going to Ion Chromatography Instrumentation | |
Sterile Vent Filter |
22.0091 | 6-12 months |
Airborne particulates | *Contamination of stored water causing system consumables work harder |
| Cleaning Solution | CMX25 | N/A | Bacteria | *Bacteria in water system *Slow dispense flow rate |
| Pretreatment RO cartridge (Optional for tap water feed) |
09.2012 | 6-12 months | Ions Organics Bacteria Particles |
*Shortened Ultrapure cartridge life |
| Hardness Stabilizer (Optional for tap water feed) | 06.5204 | 6-12 months | Hardness | *Shortened life of Pretreatment RO Cartridge |
| Description | Catalog Number | Estimated Lifetime * | Impurities removed | Effects if not replaced |
| RO membrane for Pacific TII 3, 7, 12(Requires 1) | 22.0046 | Up to 2 years | General removal of: Ions Organics Bacteria Particles |
*Increased ion, organic and bacteria impurities in water. *DI cartridge exhausting quickly |
| RO membrane for Pacific TII 20 (Requires 2) | 22.0046 | Up to 2 years | General removal of: Ions Organics Bacteria Particles |
*Increased ion, organic and bacteria impurities in water. *DI cartridge exhausting quickly |
| RO membrane for Pacific TII 40 (Requires 2) | 22.0087 | Up to 2 years | General removal of: Ions Organics Bacteria Particles |
*Increased ion, organic and bacteria impurities in water. *DI cartridge exhausting quickly |
| DI Cartridge | 09.4011 | Up to 12 months | Ions | *Low resistivity/high conductivity water *Interference in analytical methods |
| Pacific TII System UV Lamp | 50139226 | when applicable - up to 2 years | Bacteria | *Bacteria growth in system |
| 0.2 um filter for hand remote dispenser option | 09.1003 | 3 -6 months | Bacteria Particulates |
*Bacteria and particles in the water *Slow dispense flow rate |
| CO2 vent filter | 06.5002 | Up to 12 months | Airborne particulates CO2 and vapors |
*Contaimination of stored water cause water system to work harder |
| Description | Catalog Number |
Estimated Lifetime |
Impurities Removed |
Effects if Not Replaced |
| LabTower TII 20 RO membrane (Requires 1) |
22.0046 |
Up to 2 years |
General removal of: Ions Organics Bacteria Particles |
*Increased ion, organic and bacteria impurities in water. |
| LabTower TII 40 RO membranes (Requires 2) |
22.0087 |
Up to 2 years |
General removal of: |
*Increased ion, organic and bacteria impurities in water. *DI cartridge exhausting quickly |
| LabTower TII 60 RO membrane (Requires 2 each) |
22.0046 22.0087 |
Up to 2 years |
General removal of: Ions Organics Bacteria Particles |
*Increased ion, organic and bacteria impurities in water. *DI cartridge exhausting quickly |
| DI Canister |
02.2850-LAB |
Up to 12 months |
Ions | *Low resistivity/high conductivity water *Interference in analytical methods |
| 50139226 | when applicable - up to 2 years |
Bacteria |
*Bacteria growth in system | |
| Sterile 0.2 micron final filter for system and remote dispenser option | 09.1003 |
Up to 12 months |
Bacteria Particulates |
*Bacteria and particles in the water *Slow dispense flow rate |
| Sterile Vent Filter |
50135142 | Up to 12 months |
Airborne particulates CO2 and vapors |
*Contaimination of stored water cause water system to work harder |
| 0.1 micron 10" filter (Installed in LabTower TII, back white canister) | 06.5557 |
Up to 6 months |
Bacteria Particulates |
*Bacteria and particles in the water |
| Description | Catalog Number | Estimated Lifetime * | Impurities removed | Interferes with lab results if not replaced: |
| RO membrane for Pacific RO 3, 7, 12 (Requires 1) | 22.0046 | Up to 2 years | General removal of: Ions Organics Bacteria Particles |
*Increased ion, organic and bacteria impurities in water. *DI cartridge exhausting quickly |
| RO membrane for Pacific TII 20 (Requires 2) | 22.0046 | Up to 2 years | General removal of: Ions Organics Bacteria Particles |
*Increased ion, organic and bacteria impurities in water. *DI cartridge exhausting quickly |
| RO membrane for Pacific RO 40 (Requires 2) | 22.0087 | Up to 2 years | General removal of: Ions Organics Bacteria Particles |
*Increased ion, organic and bacteria impurities in water. *DI cartridge exhausting quickly |
| Vent filter | 50135142 | Up to 12 months | Airborne particulates CO2 and vapors |
*Contamination of stored water cause water system to work harder |
| Description | Catalog Number |
Estimated Lifetime |
Impurities Removed |
Effects if Not Replaced |
| LabTower RO 20 RO membranes (Requires 1) |
22.0046 |
Up to 2 years |
General removal of: Ions Organics Bacteria Particles |
*Increased ion, organic and bacteria impurities in water. |
| LabTower RO 40 RO membranes (Requires 2) |
22.0087 | Up to 2 years |
General removal of: |
*Increased ion, organic and bacteria impurities in water. *DI cartridge exhausting quickly |
| LabTower RO 60 RO membrane (Requires 2) |
22.0046 22.0087 |
Up to 2 years |
General removal of: Ions Organics Bacteria Particles |
*Increased ion, organic and bacteria impurities in water. *DI cartridge exhausting quickly |
| 09.1003 | 3 -6 months |
Bacteria Particulates |
*Bacteria and particles in the water *Slow dispense flow rate |
|
Sterile Vent Filter |
50135142 | Up to 12 months |
Airborne particulates CO2 and vapors |
*Contamination of stored water cause water system to work harder |
| Description | Catalog Number |
Estimated Lifetime |
Impurities Removed |
Effects if Not Replaced |
| Tap Water Cartridge kit (4 holder Epure) | D5028 | Up to 1 year |
Ions | *Low resistivity/high conductivity in water |
| Tap Water - Organic Free Cartridge kit (4 holder Epure) | D5023 | Up to 1 year |
Ions Organics |
*Low resistivity/high conductivity in water *Increased organics *Interference in analytical methods |
| Pretreated Feed Cartridge kit (3 holder Epure) | D5029 | Up to 1 year |
Ions | *Low resistivity/high conductivity in water *Interference in analytical methods |
Pretreated Feed - Organic Free Cartridge kit (3 holder Epure) |
D5022 | Up to 1 year |
Ions Organics |
*Low resistivity/high conductivity in water *Increased organics *Interference in analytical methods |
| D50227 | Up to 1 year |
Ions | *Low resistivity/high conductivity in water *Interference in analytical methods |
|
Pretreated Feed - Organic Free Cartridge kit (4 holder Epure) |
D50228 | Up to 1 year |
Ions Organics |
*Low resistivity/high conductivity in water *Increased organics *Interference in analytical methods |
| D3750 | 3 -6 months |
Bacteria Particulates |
*Bacteria and particles in the water *Slow dispense flow rate |
|
Cleaning cartridges (3 pk) |
D50223 | 6-12 months |
Bacteria | *Bacteria in water system |
| Description | Catalog Number | Estimated Lifetime* | Impurities removed | Effects if not replaced |
| 1 micron prefilter | D502113 | 2 x per year | Rust, sediment | *Reduced RO membrane life leading to higher impurities in water |
| Hardness stabilizer | D502114 | 2 x per year | Water hardness | *Hard water deposits on membrane leading to short consumable life, higher impurities in water |
| Carbon cartridge | D502115 | 2 x per year | Chlorine | *Oxidation of RO membrane leading to short consumable life, higher impurities in water |
| Description | Catalog Number | Estimate Lifetime | Impurities Removed | Effects if Not Replaced |
| Hardness stabilizer | 06.5452 | 6 months | Water hardness, rust, sediment | *Hard water deposits on RO membrane leading to short consumable life, higher impurities in water |
| 5 µm filter and activated carbon cartridge | 06.5201 | 3 - 4 months | Chlorine | *Reduced RO membrane life leading to higher impurities in water |
| 1 µm prefilter | 06.5101 | 4 - 6 months | Rust, sediment | *Hard water deposits on membrane leading to short consumable life, higher impurities in water |
| 5 µm prefilter and hardness stabilizer | 06.5204 | 4 - 6 month | Water hardness | *Oxidation of RO membrane leading to short consumable life, higher impurities in water |
| Salt pellets | N/A | When pellets are gone | Water hardness | *Premature failure of EDI |