Barnstead™ E-Pure™ Cartridge Kits
Thermo Scientific™
Barnstead™ E-Pure™ Cartridge Kits
Replace worn, outdated and otherwise ineffective cartridges with Thermo Scientific™ Barnstead™ E-Pure™ Cartridge Kits.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberFor Use With (Equipment)Feedwater Source
D50228E-Pure 4-holder, Organic freePretreated
D5022E-Pure 3-holder, Organic freePretreated feed
D50227E-Pure 4-holderPretreated
D5023E-Pure 4-holder, Organic freeTap
D5028E-Pure 4-holderTap
D5029E-Pure 3-holderPretreated feed
Catalog number D50228
Price (ZAR)
-
For Use With (Equipment):
E-Pure 4-holder, Organic free
Feedwater Source:
Pretreated
Replace worn, outdated and otherwise ineffective cartridges with Thermo Scientific™ Barnstead™ E-Pure™ Cartridge Kits. These cartridge kits are classified based on type of feed water used; choose the appropriate kit according to your feed water and need for organic free water.
Replace worn, outdated and otherwise ineffective cartridges with Thermo Scientific™ Barnstead™ E-Pure™ Cartridge Kits. These cartridge kits are classified based on type of feed water used; choose the appropriate kit according to your feed water and need for organic free water.
Specifications
DescriptionCartridge Kit for E-Pure 4-holder; Pretreated feed, Organic free
For Use With (Equipment)E-Pure 4-holder, Organic free
TypeCartridge Kit
Feedwater SourcePretreated
Unit SizeEach