Eliminate particles and microorganisms from your sample — reliably
Prevent the accumulation of fine particles that can block your chromatography column. Thermo Scientific Titan3, Target2, and Choice SyringeFilters remove interfering materials, fine particles, and microorganisms, providing you with cleaner sample extracts. These high-quality robust sample filtration solutions are available in a variety of syringe filter sizes, membranes, and housings for a wide range of samples and applications.
Titan3, Target2, and Choice syringe filters
Syringe Filter | Application | ||
---|---|---|---|
Premium Brand | Routine Brand | Basic Brand | |
Titan3 Nylon | Target2 Nylon | Choice Nylon |
|
Titan3 PTFE (Hydrophobic) |
Target2 PTFE (Hydrophobic) |
Choice PTFE (Hydrophobic) |
|
Titan3 PTFE (Hydrophilic) |
— | Choice PTFE (Hydrophilic) |
|
Titan3 PVDF (Hydrophobic) |
Target2 PVDF (Hydrophobic) |
Choice PVDF (Hydrophobic) |
|
— | — | Choice PVDF (Hydrophilic) |
|
Titan3 Regenerated Cellulose (RC) | Target2 Nylon Regenerated Cellulose (RC) | Choice Nylon Regenerated Cellulose (RC) |
|
Titan3 PES (Polyethersulfone) |
Target2 PES (Polyethersulfone) |
Choice PES (Polyethersulfone) |
|
Titan3 GMF (Glass Microfiber) |
Target2 (Glass Microfiber) |
— |
|
Titan3 Polypropylene | Target2 Polypropylene | Choice Polypropylene |
|
Titan3 Cellulose Acetate | Target2 Cellulose Acetate | Choice Cellulose Acetate |
|
What is the difference between Titan3, Target2, and Choice syringe filters?
Titan3 Premium Syringe Filters | Target2 Routine Syringe Filters | Choice Basic Syringe Filters | |
---|---|---|---|
Range of filter sizes | 4mm to 30mm | 4mm to 30mm | 13mm to 25mm |
High-quality filtration solutions | |||
Low extractable membranes and housing | |||
Plain polypropylene housing | |||
HPLC performance tested | |||
Enhanced Luer Lock inlet which provides secure attachment to syringes | |||
Low hold-up volume allows for the treatment of small sample volumes with minimal sample loss | |||
Color coded for easy identification and selection of the correct membrane and pore size | |||
30mm products pressure rated to 120psi for enhanced sample safety | |||
Most 30mm devices have a 1mm borosilicate glass pre-filter for increased filtrate volumes |
How to select your syringe filter
Get this syringe filter technical guide to determine:
- Membrane compatibility with your sample type and technique
- Compatibility of membrane with solvent
- Filter size, which is dependent on sample volume
- Porosity, which depends on the amount of particles to be removed from the sample
Select Chromatography Consumables and Columns for your Pharma, Biopharma, Clinical, Food & Beverage or Environmental workflow.
Find interactive tools and calculators, guides to select the right products, equivalent product finders, and get in contact with our consumables technical support team to answer your application questions.
On-demand Consumables for chromatography webinar series: Learn at your own pace on a wide range of topics from column troubleshooting, method transfer, to tips for successful low-flow and more
Your sample is precious and you cannot afford failures. Take a look at our comprehensive range of products that deliver clean and consistent extractions using innovative sample preparation technologies to accomplish your analytical requirements.