Arcturus® AutoScanXT LCM Software
  • Designed to mimic the user’s workflow from top to bottom in five easy steps
  • All tools to perform a basic microdissection are at the top level—no fussing with right-clicking or hidden menus
  • One-click image documentation
  • Quality control—visualize and document exactly what has been collected on the LCM cap
Collect software update - coming soon
Download ArcturusXT Software v3.4 (software and driver) (.zip)
Download ArcturusXT Software v3.4 (software only) (.zip)
Release notes (.pdf)
Easy workflow to simplify microdissection
1 Load sample
Place your sample in the instrument platform.
2 Inspect sample
Visualize your sample on the monitor.
3 Select cells
Trace the outline of the targeted portion from the sample to dissect.
4 Microdissect
The UV or IR laser dissects the selected target.