Trusted microarray analysis for uncompromised results
Microarrays are an ideal platform for copy number variation (CNV) analysis and molecular cytogenetic research for solid and liquid tumors. Applied Biosystems CytoScan microarrays offer a reliable genome-wide approach for high-resolution DNA copy number analysis to detect gains, losses, loss of heterozygosity (LOH)/absence of heterozygosity (AOH), copy-neutral loss of heterozygosity (cnLOH), regions identical-by-descent, and mosaicism.
All CytoScan arrays are hybrid-SNP chromosomal microarrays that contain large numbers of both SNP probes and non-polymorphic probes. Hybrid-SNP arrays, with more than 99% genotype accuracy, boost confidence in breakpoint determination and provide independent confirmation of copy number events throughout the entire genome.
Our robust microarray solutions are designed to provide reproducible results, save you time and money, reduce error, and deliver quality performance consistent with your laboratory requirements.
Workflow
Thermo Fisher Scientific offers powerful microarray solutions for oncology clinical research that are powered with robust workflows that make it easy to obtain consistent, reliable, and high-quality results.
Our complete microarray platform, the CytoScan Cytogenetics Suite, includes hybrid-SNP arrays, automated and manual target preparation options, fully kitted reagents, the GeneChip System 3000 instrument for array processing, and Chromosome Analysis Suite (ChAS) software for data interpretation and reporting. These tools support laboratories to help maximize operational time.
image of the oncology workflow
Product overview
CytoScan HD Accel Array
image of Cytoscan HD array
With enhanced speed, improved coverage, and lesser sample input requirement, the CytoScan HD Accel Array can help maximize lab productivity by 100%, enabling you to reach a new level of efficiency.
  • Two-day assay workflow
  • 100 ng of genomic DNA input—50% less than other commercially available chromosomal microarrays
  • Consistent target generated by the assay that is hybridized to the array for reproducible and reliable results
  • Challenging sample types (e.g., buccal swabs, saliva, products of conception, amniotic fluid, and chorionic villus sampling (CVS)) included in reference model file to help enable generation of higher-quality results
  • Improved coverage in more than 5,000 regions across entries in OMIM® database, RefSeq, ClinGen, DECIPHER/DDD constitutional regions, and the COSMIC Cancer Gene Census (CGC)
CytoScan HD Array
image of Cytoscan HD array
The benchmark in cytogenetics research with highest genome-wide resolution of CNVs for applications in oncology research. Key highlights of CytoScan HD array are:
  • 2.67 million markers for copy number analysis, including 750,000 SNPs and 1.9 million nonpolymorphic probes.
  • Exceptional coverage across entries in OMIM database, RefSeq, ClinGen, DECIPHER/DDD constitutional regions, and the COSMIC Cancer Gene Census (CGC)
  • Advanced, proprietary manufacturing technology that produces highly reproducible arrays between batches, with no risk of probe dropout that occurs with bead array technology
OncoScan Assay
Catch critical copy number changes with OncoScan CNV Assay. If you work with highly degraded DNA, such as that derived from FFPE tumor samples of various ages or product of conception (POC), and with low amounts of DNA starting material, common in cell-free DNA (cfDNA), learn more why this assay is a natural choice for clinical research and beyond.
OncoScan Assay
Catch critical copy number changes with OncoScan CNV Assay. If you work with highly degraded DNA, such as that derived from FFPE tumor samples of various ages or product of conception (POC), and with low amounts of DNA starting material, common in cell-free DNA (cfDNA), learn more why this assay is a natural choice for clinical research and beyond.
Product specifications
CytoScan HD Accel
CytoScan HD
OncoScan Assay
Research applications
Fastest turnaround time in the industry with improved coverage for highest genome-wide resolution of CNVs for applications in prenatal and postnatal The benchmark in cryogenics research with highest genome-wide resolution of CNVs for applications in prenatal and postnatal research High resolution analysis, up to 50 kb in top cancer genes and 300 kb across whole genome in FFPE and fresh frozen tissues
Sample types
Blood; buccal swabs; saliva; bone marrow;
uncultured or cultured cells; chorionic villi; amniocytes; POC
Blood, buccal swabs, saliva, uncultured or cultured cells, chorionic villi, amniocytes, POC. FFPE, fresh and frozen tissue
Size of aberration*
Losses: 25 kb
Gains: 50 kb
LOH/AOH: 3 Mb
Mosaicism: >15% (approximately)
Losses: 25 kb
Gains: 50 kb
LOH/AOH: 3Mb
Mosaicism: >15% (approximately)
Gains: 50 kb
Losses: 50 kb
LOH/AOH: 10 Mb
Mosaicism (% aberrant cells): 15%
Input DNA**
100 ng 10-250 ng** 80 ng**
Probe structure
2.8 million markers for whole genome coverage 2 million nonpolymorphic markers 750,000 SNP probes for LOH/AOH analysis, duo-trio assessment, and sample tracking
2.67 million markers for whole genome coverage
1.95 million nonpolymorphic markers
-743,000 SNP probes for LOH/AOH analysis, duo-trio assessment, and sample tracking
220,000 molecular inversion probes (MIPs), whole genome coverage
5,700 non-polymorphic probes
216,000 SNP probes
Protocol
2 days 3-4 days 2-3 days
*Size of aberration—The size of the segment call depends on the average marker spacing in the region. Best performance can be achieved in regions with higher marker coverage. Mosaicism detection may depend on the size of the altered segment and the type of aberration involved.
**250 ng is optimal but users have reported success using as little as 10 ng starting DNA.
Ordering information
OncoScan Assay
For Research Use Only. Not for use in diagnostic procedures.