Note: This protocol is intended for use with the specific products mentioned within it. Substituting different products is not recommended.
The use of the annexin V apoptosis assay protocol is a common method for detecting apoptotic cells. The below protocols are recommended for use with the specific flow cytometry kits mentioned. Please see the Annexin V Staining page for a discussion about general experimental conditions and avoiding false positives, or to review a selection guide for all of our annexin V products.
Annexin V flow cytometry protocols
General notes
  • Invitrogen Fixable Viability Dye (FVD) eFluor 450 is not recommended for use with Annexin V Apoptosis Detection Kits.
  • Due to the calcium dependence of the Annexin V:PS interaction, it is critical to avoid buffers containing EDTA or other calcium chelators during Annexin V experiments. 
  • Annexin V can only be used as a marker of apoptosis in cells where the plasma membrane is intact. Destroying the integrity of the plasma membrane will allow binding of Annexin V to PS inside the cell.
Annexin V staining protocol
Materials
  • 12 x 75 mm round-bottom tubes 
  • 1X PBS 
  • Annexin V Apoptosis Detection kit (any one of the kits listed). Each kit includes an Annexin V conjugate.
        ° eFluor 450 (Cat. Nos. 88-8006-72, 88-8006-74)
        ° FITC (Cat. Nos. 88-8005-72, 88-8005-74)
        ° PerCP-eFluor 710 (Cat. Nos. 88-8008-72, 88-8008-74)
        ° PE (Cat. Nos. 88-8102-72, 88-8102-74)
        ° PE-Cyanine7 (Cat. Nos. 88-8103-72, 88-8103-74)
        ° APC (Cat. Nos. 88-8007-72, 88-8007-74)
  • 10X Binding buffer
Note:PerCP-eFluor 710 (Cat. Nos. 88-8008-72, 88-8008-74) do not include a viability dye. We recommend using this format in combination with a Invitrogen Fixable Viability Dye (FVD) such as FVD eFluor 660 (Cat. Nos. 65-0864-14, 65-0864-18), FVD eFluor 506 (Cat. Nos. 65-0866-14, 65-0866-18), or FVD eFluor 780 (Cat. Nos. 65-0865-14, 65-8065-18).
Experimental procedure
  1. Prepare 1X binding buffer by mixing 1 part of 10X binding buffer with 9 parts of distilled water. 
  2. Harvest cells. 
  3. Wash cells once in 1X PBS, then once in 1X binding buffer. 
  4. Resuspend cells in 1X Binding Buffer at 1-5 x 106 cells/mL. 
  5. Add 5 μL of fluorochrome-conjugated Annexin V to 100 μL of the cell suspension. 
  6.  Incubate 10-15 minutes at room temperature. Protect from light. 
  7. Add 2 mL 1X binding buffer and centrifuge at 400-600 x g for 5 minutes at room temperature. Discard supernatant. 
  8. Resuspend cells in 200 µL of 1X binding buffer. 
  9. Add 5 μL of Propidium Iodide Staining Solution or 7-AAD Viability Staining Solution and incubate 5-15 minutes on ice or at room temperature.
    • Note: Propidium iodide and 7-AAD must remain in the buffer during acquisition. Do not wash cells after the addition of propidium iodide or 7-AAD.
  10. Analyze by flow cytometry.
    • Note: Cells should be analyzed within 4 hours after the initial incubation period due to adverse effects on the viability of cells left in the presence of propidium iodide or 7-AAD for prolonged periods. Store at 2–8°C and protect from light until ready for analysis. 
Annexin V staining protocol with Fixable Viability Dyes
Materials
  • 12 x 75 mm round-bottom tubes 
  • 1X PBS 
  • Annexin V Apoptosis Detection kit (any one of the kits listed). Each kit includes an Annexin V conjugate.
        ° eFluor 450 (Cat. Nos. 88-8006-72, 88-8006-74)
        ° FITC (Cat. Nos. 88-8005-72, 88-8005-74)
        ° PerCP-eFluor 710 (Cat. Nos. 88-8008-72, 88-8008-74)
        ° PE (Cat. Nos. 88-8102-72, 88-8102-74)
        ° PE-Cyanine7 (Cat. Nos. 88-8103-72, 88-8103-74)
        ° APC (Cat. Nos. 88-8007-72, 88-8007-74)
  • 10X Binding buffer 
  • Flow Cytometry Staining Buffer (Cat. No. 00-4222) 
  • PBS (azide- and serum/protein-free PBS) 
  • FVD eFluor 660 (Cat. Nos. 65-0864-14, 65-0864-18), FVD eFluor 506 (Cat. Nos. 65-0866-14, 65-0866-18) or FVD eFluor 780 (Cat. Nos. 65-0865-14, 65-8065-18)
Note: FVD eFluor 450 is not recommended for use with the Annexin V Apoptosis Detection Kits.
Experimental procedure
  1. Prepare 1X binding buffer by mixing 1 part of 10X binding buffer with 9 parts of distilled water. 
  2. Wash cells twice in azide-free and serum/protein-free PBS. 
  3. Resuspend cells at 1-10 x 106 cells/mL in azide-free and serum/protein-free PBS. 
  4. Add 1 µL of FVD per 1 mL of cells and vortex immediately. 
  5. Incubate for 30 minutes at 2-8°C. Protect from light. 
  6. Wash cells twice with Flow Cytometry Staining Buffer or equivalent. 
  7. Wash cells once with 1X Binding Buffer. 
  8. Resuspend cells in 1X Binding Buffer at 1-5 x 106 cells/mL. 
  9. Add 5 μL of fluorochrome-conjugated Annexin V to 100 μL of the cell suspension. 
  10. Incubate 10-15 minutes at room temperature. Protect from light. 
  11. Add 2 mL of 1X binding buffer and centrifuge at 400-600 x g for 5 minutes at room temperature. Discard supernatant. 
  12. Resuspend cells in 200 μL of 1X Binding Buffer. 
  13. Analyze by flow cytometry.
Annexin V staining protocol with surface and intracellular staining
Materials
Note: FVD eFluor 450 is not recommended for use with the Annexin V Apoptosis Detection Kits.
Experimental procedure
  1. Prepare 1X binding buffer by mixing 1 part of 10X binding buffer with 9 parts of distilled water. 
  2. Stain cell surface antigen(s). Refer to ‘Staining Cell Surface Targets, Protocol A’. 
  3. Wash cells twice in azide-free and serum/protein-free PBS. 
  4. Resuspend cells at 1-10 x 106 cells/mL in azide-free and serum/protein-free PBS. 
  5. Add 1 µL of Fixable Viability Dye per 1 mL of cells and vortex immediately. 
  6. Incubate for 30 minutes at 2-8°C. Protect from light.  
  7. Wash cells twice in Flow Cytometry Staining Buffer or equivalent. 
  8. Wash cells once with 1X binding buffer. 
  9. Resuspend cells in 1X binding buffer at 1-5 x 106 cells/mL. 
  10. Add 5 μL of fluorochrome-conjugated Annexin V to 100 μL of the cell suspension. 
  11. Incubate 10-15 minutes at room temperature. Protect from light. 
  12. Wash cells once with 1X binding buffer. 
  13. Stain intracellular antigen(s). Refer to ‘Staining Intracellular Antigens, Protocol A or B’. 
  14. Analyze by flow cytometry.
Resources
BioProbes Journal of Cell Biology Applications—Stay up-to-date with highlights of the latest breakthroughs and get information about new technologies and products.
Tools and Support
Flow Cytometry Panel Builder—Design your flow cytometry panel with this online tool for a simplified, customizable experience to fit your needs.
Flow Cytometry Learning Center—Access flow cytometry educational resources for better experiment planning and execution.
Flow Cytometry Support Center—Find technical support recommendations for your flow cytometry workflows, including tips for experimental setup and in-depth troubleshooting help.