Rat embryonic sensory neuron. Rhodamine phalloidin.
A fixed and permeabilized rat embryonic sensory neuron was stained with the filamentous actin (F-actin) stain, rhodamine phalloidin (Cat. no. R415). The image was acquired using filters appropriate for TRITC. Image contributed by Heiti Paves, Laboratory of Molecular Genetics, National Institute of Chemical Physics and Biophysics, Estonia.
A fixed and permeabilized rat embryonic sensory neuron was stained with the filamentous actin (F-actin) stain, rhodamine phalloidin (Cat. no. R415). The image was acquired using filters appropriate for TRITC.  Image contributed by Heiti Paves, Laboratory of Molecular Genetics, National Institute of Chemical Physics and Biophysics, Estonia.
Related Products
Related Images