Bovine pulmonary artery endothelial cells (BPAEC). Paclitaxel, biotin-XX goat anti–mouse IgG and ELF® 97 Cytological Labeling Kit
Bovine pulmonary artery endothelial cells (BPAEC) fixed and permeabilized in the presence of paclitaxel (Cat. no. P3456) to stabilize the microtubules. Microtubules were then labeled with mouse monoclonal anti–ß-tubulin IgG and biotin-XX goat anti–mouse IgG (Cat.no. B2763) and visualized with the ELF® 97 Cytological Labeling Kit (Cat. no. E6603).
Bovine pulmonary artery endothelial cells (BPAEC) fixed and permeabilized in the presence of paclitaxel (Cat. no. P3456) to stabilize the microtubules. Microtubules were then labeled with mouse monoclonal anti–ß-tubulin IgG and biotin-XX goat anti–mouse IgG (Cat.no. B2763) and visualized with the ELF® 97 Cytological Labeling Kit (Cat. no. E6603).
Related Products
Related Images