Search
NIH 3T3 cells stained with MitoTracker® CMXRos (Cat. no. M7512), BODIPY® FL phallacidin (Cat. no. B607) and POPO™-1 (Cat. no. P3580). The cells were incubated with MitoTracker® CMXRos, which stains mitochondria red. After the fixed cells were permeabilized with acetone, they were stained with BODIPY® FL phallacidin, which labels F-actin green, and with POPO™-1, which labels nuclei blue. This photomicrograph was obtained with a single exposure through an Omega Optical triple bandpass filter set.
1% Agarose gel containing 16S and 23S ribosomal RNA (rRNA). SYBR® Green II RNA gel stain. Go ›
Bovine pulmonary artery endothelial cells (BPAEC). MitoTracker® Red CMXRos, SYTOX® Green nucleic acid stain, biotin-XX goat anti–mouse IgG antibody and Cascade Blue® NeutrAvidin biotin-binding protein. Go ›
Endothelial cells. Go ›
Mouse Anti-Alpha Tubulin Monoclonal Antibody (Cat. No. A11126) Go ›
Live cells transduced with Organelle Lights™ or Cellular Lights™ reagents. Go ›
Multicolor fluorescence analysis of muntjac fibroblasts. Go ›
Simultaneous detection of expression of five genes in a whole mount Drosophila embryo by fluorescence in situ hybridization (FISH) with five RNA probes Go ›
Nuclear, cytoskeletal, and mitochondrial staining of Muntjac fibroblasts. Go ›