The 5500 Series Genetic Analysis Systems are scalable and accurate next generation sequencing. The 5500 Series Genetic Analysis Systems support a wide range of research applications on a per-lane basis with independently addressable and configurable lanes on either one or two FlowChips. The Exact Call Chemistry (ECC) module boosts the already industry-leading accuracy of this ligation-based sequencing.
Product Information
View specification sheet
What Level of Accuracy Do You Demand?
  • Do you need to detect somatic mutations present at 1% in your sample?
  • Do you want to detect variants present in 5% of your sample with 1/3 the sequence coverage needed with other platforms
Demand the Most!
See for Yourself! - Data and Publications
See SNP detection with less false positives
Uniform sequence coverage
See SNP detection with less false positives
Uniform sequence coverage