Wall Pellitory Allergen Facts, Symptoms, and Treatment
Learn more about common allergic diseases, symptoms, management paradigms, and testing considerations.
Is your patient a candidate for specific IgE testing?
Get detailed information on whole allergens and allergen components.
Ready to test a patient?
Learn more about common allergic diseases, symptoms, management paradigms, and testing considerations.
Is your patient a candidate for specific IgE testing?
Get detailed information on whole allergens and allergen components.
Ready to test a patient?
A bushy, wind-pollinated, perennial weed, wall pellitory is common in the Mediterranean and along the west coast of Europe. It's also been introduced in other parts of Europe and the Middle East as well as in North and South America and Australia. It prefers to root near surfaces such as rocks, banks, and walls, hence its name.1 Wall pellitory has two long flowering periods, so while its pollen appears at the beginning of spring, it persists throughout the spring and summer and into fall.2 The pollen from this severely allergenic plant causes symptoms of asthma, allergic conjunctivitis, and allergic rhinitis (hay fever).1,3 Wall pellitory is also known by names such as pellitory-of-the-wall, parietaire, spreading pellitory, asthma weed, and sticky-weed.1
Wall pellitory is common in the Mediterranean and along the west coast of Europe. It's also been introduced in other parts of Western Europe, Argentina, and Australia, and two related species are found in Brazil and the United States.1
Many patients with wall pellitory allergy can experience symptoms when exposed to other allergens such as tree, weed, or grass pollens, making it difficult to determine which pollen is causing the symptoms, especially when pollen seasons are overlapping. This is called cross-reactivity and occurs when your body's immune system identifies the proteins, or components, in different substances as being structurally similar or biologically related, thus triggering a response.5 Other respiratory allergens that may cause reactions associated with wall pellitory are certain grasses, trees, and weeds (e.g., birch, alder, juniper, ragweed, olive, goosefoot, pigweed).6
Knowing the proteins, or components, within each allergen that are triggering your symptoms can help guide your management plan. With that in mind, and based on your symptom history, your healthcare provider may suggest something called a specific IgE component test, which can help reveal other pollens and foods you may react to. Results from this test can also help your healthcare provider decide if allergen immunotherapy may reduce your symptoms.5
Already have your specific IgE component test results?
Your component test results will include the name of the components (a series of letters and numbers). Your healthcare provider will likely review the results with you, but here you'll find an at-a-glance breakdown you can use as a reference. Simply match the component names to the list below to see what they mean in terms of symptom management.5
rPar j 2
rPhl p 7
rPhl p 12
MUXF3 (CCD)
Test results should be interpreted by your healthcare provider in the context of your clinical history. Final diagnosis and decision on further management is made by your healthcare provider.
*These products may not be approved for clinical use in your country. Please work with your healthcare provider to understand availability.
The management of allergic rhinitis includes avoidance of relevant allergens, symptomatic treatment, and allergen immunotherapy.6-8
Wall pellitory allergy symptoms can be similar to many other pollen allergies and may include:4,6
If you're sensitized to wall pellitory and have asthma, the weed pollen may trigger or worsen asthma symptoms, such as coughing and wheezing.4,6
Together with your symptom history, skin-prick testing or specific IgE blood testing can help determine if you are sensitized to a particular allergen. If you are diagnosed with an allergy, your healthcare provider will work with you to create a management plan.
*These products may not be approved for clinical use in your country. Please work with your healthcare provider to understand availability.
Wall pellitory has two long flowering periods. So while its pollen appears at the beginning of spring, it persists throughout the spring and summer and into the fall.2 However, most patients sensitized to wall pellitory experience clinical symptoms prevalently in the spring.1