    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Food and Beverage
    • Lab Solutions
    • Pharma and Biopharma
    • Real-Time PCR
    • Semiconductor Analysis
  • Services
    • 360° CDMO and CRO Solutions
    • Digital Solutions
    • Product Support
    • Technical Information
    • Training and Education
          • Primary Antibodies ›
          • WRN Antibodies
          Antibodies Inc
          WRN Monoclonal Antibody (N428/12), FL650
          View all (23) WRN antibodies
          Tech Support
          Cite WRN Monoclonal Antibody (N428/12), FL650
          Additional Information:
          {{banner.description}}
          WRN Monoclonal Antibody (N428/12), FL650
          Product Details
          75-423-FL650
          Applications
          Tested Dilution
          Publications
          Immunohistochemistry (Frozen) (IHC (F))
          Assay-dependent
          -
          Immunocytochemistry (ICC/IF)
          Assay-dependent
          -
          ELISA (ELISA)
          Assay-dependent
          -
          Product Specifications
          Species Reactivity
          Human, Rat
          Host/Isotype
          Mouse / IgG1
          Class
          Monoclonal
          Type
          Antibody
          Clone
          N428/12
          Immunogen
          Fusion protein amino acids 38-236 (exonuclease domain) of human WRN (accession number Q14191) produced recombinantly in E. Coli.
          3D Epitope / Immunogen
          Conjugate
          FL650 FL650 FL650
          • Unconjugated
          • FL490
          • FL550
          • FL594
          • Request custom conjugation
          Excitation/Emission Max
          655/676 nm
          Form
          Liquid
          Concentration
          0.5 mg/mL
          Amount
          100 µg
          Purification
          Protein A
          Storage buffer
          PBS
          Contains
          0.09% sodium azide
          Storage conditions
          Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
          Shipping conditions
          Ambient (domestic); Dry ice (international)
          Product Specific Information
          For maximum recovery of product, centrifuge the vial prior to removing the cap.
          Target Information
          This gene encodes a member of the RecQ subfamily and the DEAH (Asp-Glu-Ala-His) subfamily of DNA and RNA helicases. DNA helicases are involved in many aspects of DNA metabolism, including transcription, replication, recombination, and repair. This protein contains a nuclear localization signal in the C-terminus and shows a predominant nucleolar localization. It possesses an intrinsic 3' to 5' DNA helicase activity, and is also a 3' to 5' exonuclease. Based on interactions between this protein and Ku70/80 heterodimer in DNA end processing, this protein may be involved in the repair of double strand DNA breaks. Defects in this gene are the cause of Werner syndrome, an autosomal recessive disorder characterized by premature aging.
          Bioinformatics
          Protein Aliases: Bifunctional 3'-5' exonuclease/ATP-dependent helicase WRN; DKFZp686C2056; DNA helicase, RecQ-like type 3; exonuclease WRN; RecQ protein-like 2; similar to Werner syndrome helicase homolog; Werner syndrome ATP-dependent helicase; Werner syndrome protein; Werner syndrome RecQ like helicase; Werner syndrome, RecQ helicase-like; WRN
          View more View less
          Gene Aliases: RECQ3; RECQL2; RECQL3; WRN
          View more View less
          UniProt ID: (Human) Q14191
          View more View less
          Entrez Gene ID: (Rat) 290805, (Human) 7486
          View more View less
          Function(s)
          magnesium ion binding Y-form DNA binding bubble DNA binding nucleic acid binding DNA binding DNA helicase activity ATP-dependent DNA helicase activity helicase activity exonuclease activity ATP binding 3'-5' exonuclease activity four-way junction helicase activity ATPase activity manganese ion binding protein complex binding protein homodimerization activity 3'-5' DNA helicase activity ATP-dependent 3'-5' DNA helicase activity G-quadruplex DNA binding nucleotide binding four-way junction DNA binding catalytic activity nuclease activity protein binding hydrolase activity isomerase activity MutLalpha complex binding macromolecular complex binding metal ion binding forked DNA-dependent helicase activity telomeric D-loop binding telomeric G-quadruplex DNA binding 3'-flap-structured DNA binding catalytic activity, acting on DNA 8-hydroxy-2'-deoxyguanosine DNA binding DNA helicase
          Process(es)
          telomere maintenance DNA synthesis involved in DNA repair replicative cell aging DNA metabolic process DNA replication DNA repair base-excision repair double-strand break repair DNA recombination cellular response to DNA damage stimulus response to oxidative stress cell aging cellular response to starvation response to UV-C multicellular organism aging replication fork processing nucleolus to nucleoplasm transport DNA duplex unwinding regulation of growth rate regulation of apoptotic process positive regulation of hydrolase activity cellular response to gamma radiation nucleic acid phosphodiester bond hydrolysis double-strand break repair via homologous recombination nucleobase-containing compound metabolic process mismatch repair determination of adult lifespan telomere maintenance via semi-conservative replication DNA geometric change telomeric D-loop disassembly cellular senescence replicative senescence t-circle formation positive regulation of strand invasion protein localization to nucleolus
          Taiwan flag icon
          Taiwan
          Your items have has been added!
          Host server : magellan-search-blue-67655d8755-vzwvz:80/100.66.79.173:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline