  • Applications
    • Real-Time PCR
    • Cloning
    • Protein Biology
  • Online Order
    • Custom Primers & TaqMan Probes
    • miRNA Mimics & Inhibitors
    • Stealth RNAi / siRNA
    • Silencer Select siRNAs
    • Custom DNA Oligos
    • GeneArt Services
    • GeneArt Strings DNA Fragments
    • TrueGuide CRISPR gRNA
    • TaqMan Real-Time PCR Assays
    • See all online orderable products
  • Services
    • Custom Services
    • Instrument Qualification Services
    • Technical Services
    • Pipette Services
    • Sample Request
    • Instrument Maintenance Services
    • Repairs and Relocation Services
    • OEM and Licensing Services
    • Events, Seminars, Training
    • Unity Lab Services
    • See all services
  • Support
    • Catalogs
    • Application Notes
    • Safety Data Sheets
    • Legal and Regulatory
    • Certificates of Analysis Search
    • Nunc/Nalgene Product Certificates
    • e-learning
    • FAQs
            • Connect Your Lab
            • Custom Products & Projects
            • Instrument Management
          • Primary Antibodies ›
          • Cytochrome C Antibodies
          Proteintech
          Cytochrome c Recombinant Rabbit Monoclonal Antibody (230503E10), CoraLite® Plus 488
          View all (59) Cytochrome C antibodies
          Tech Support
          Cite Cytochrome c Recombinant Rabbit Monoclonal Antibody (230503E10), CoraLite® Plus 488
          Additional Information:
          {{banner.description}}
          • Antibody Testing Data (1)
          Cytochrome c Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          Cytochrome c Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          FIGURE: 1 / 1
          Cytochrome c Antibody (CL488-83276) in ICC/IF
          Immunofluorescent analysis of (-20°C Methanol) fixed A431 cells using CoraLite Plus 488 Cytochrome c antibody (CL488-83276, Clone: 230503E10 ) at dilution of 1:200. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product
          {{$ctrl.videoDesc}}
          {{$ctrl.videoLongDesc}}
          Cytochrome c Antibody in Immunocytochemistry (ICC/IF)
          Cytochrome c Recombinant Rabbit Monoclonal Antibody (230503E10), CoraLite® Plus 488
          Product Details
          CL488-83276
          Applications
          Tested Dilution
          Publications
          Immunocytochemistry (ICC/IF)
          1:50-1:500
          -
          Product Specifications
          Species Reactivity
          Human, Mouse
          Host/Isotype
          Rabbit / IgG
          Expression System
          HEK293F
          Class
          Recombinant Monoclonal
          Type
          Antibody
          Clone
          230503E10
          Immunogen
          Cytochrome c fusion protein Ag1455
          3D Epitope / Immunogen
          Conjugate
          CoraLite® Plus 488 CoraLite® Plus 488 CoraLite® Plus 488
          • Unconjugated
          • CoraLite 594
          • CoraLite Plus 750
          • Request custom conjugation
          Excitation/Emission Max
          493/522 nm
          Form
          Liquid
          Concentration
          1000 µg/mL
          Amount
          100 µg
          Purification
          Affinity chromatography
          Storage buffer
          PBS, pH 7.3, with 0.5% BSA, 50% glycerol
          Contains
          0.05% ProClin 300
          Storage conditions
          -20°C, store in dark
          Shipping conditions
          Wet ice
          Product Specific Information
          Sequence of this protein is as follows: MGDVEKGKKI FIMKCSQCHT VEKGGKHKTG PNLHGLFGRK TGQAPGYSYT AANKNKGIIW GEDTLMEYLE NPKKYIPGTK MIFVGIKKKE ERADLIAYLK KATNE
          Target Information
          Cytochrome C is an electron transporting protein that resides within the intermembrane space of the mitochondria, where it plays a critical role in oxidative phosphorylation and the production of cellular ATP. Release of cytochrome C from the mitochondria to the cytosol is triggered by apoptotic stimuli. In the cytoplasm, the cytochrome C binds apoptotic protease activating factor which activates the apoptotic initiator procaspase 9. Cytosolic cytochrome C functions in the activation of caspase 3, an ICE family molecule that is a key effector of apoptosis.
          Bioinformatics
          Protein Aliases: CRF; CYCS; Cytochrome c; Cytochrome c, somatic; hc1; unnamed protein product
          View more View less
          Gene Aliases: CYC; CYCS; HCS; THC4
          View more View less
          UniProt ID: (Human) P99999, (Mouse) P62897
          View more View less
          Entrez Gene ID: (Human) 54205, (Mouse) 13063
          View more View less
          Function(s)
          protein binding electron carrier activity heme binding metal ion binding electron transporter, transferring electrons from CoQH2-cytochrome c reductase complex and cytochrome c oxidase complex activity
          Process(es)
          mitochondrial electron transport, ubiquinol to cytochrome c mitochondrial electron transport, cytochrome c to oxygen apoptotic process cellular respiration apoptotic signaling pathway intrinsic apoptotic signaling pathway execution phase of apoptosis apoptotic DNA fragmentation activation of cysteine-type endopeptidase activity involved in apoptotic process by cytochrome c hydrogen peroxide metabolic process ATP synthesis coupled electron transport
          Ordering Plus Icon Minus Icon
          • Online Orderable Products
          • Privacy Policy for Online Ordering
          • Act on Specified Commercial Transactions
          • About Returns
          • About Displayed Prices
          • About Shipping Fees
          • Documents and Certificates
          • Study information disclosure
          • Promotions
          • Events and Seminars
          • Blog
          • Corporate Information
          • Social Responsibility (CSR)
          • Enhancing Customer Experience
          • Careers Careers
          • News
          Japan flag icon
          Japan
          Your items have has been added!
          Host server : magellan-search-green-7b6576f7bc-t4kr2:80/100.66.77.141:80.
          git-commit: 6d309ebe7a31b2ceadeb87f43b473eebab110cf3
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-2026.03.11-1.0