    • See all product categories
  • Applications
    • Bioprocessing
  • Services
    • 360° CDMO and CRO Solutions
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • ESRRB Antibodies
          Proteintech
          ESRRB Monoclonal Antibody (3E10C1), CoraLite® 594
          View all (27) ESRRB antibodies
          Questions & Answers
          Cite ESRRB Monoclonal Antibody (3E10C1), CoraLite® 594
          Additional Information:
          {{banner.description}}
          • Antibody Testing Data (2)
          ESRRB Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          ESRRB Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          FIGURE: 1 / 2
          ESRRB Antibody (CL594-66818) in ICC/IF
          Immunofluorescent analysis of (4% PFA) fixed Caco-2 cells using CoraLite594 ESRRB antibody (CL594-66818, Clone: 3E10C1 ) at dilution of 1:600. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product
          {{$ctrl.videoDesc}}
          {{$ctrl.videoLongDesc}}
          ESRRB Antibody in Immunocytochemistry (ICC/IF)
          ESRRB Antibody in Flow Cytometry (Flow)
          ESRRB Monoclonal Antibody (3E10C1), CoraLite® 594
          Product Details
          CL594-66818
          Applications
          Tested Dilution
          Publications
          Immunocytochemistry (ICC/IF)
          1:300-1:1,200
          -
          Flow Cytometry (Flow)
          1x10^6 cells
          -
          Product Specifications
          Species Reactivity
          Human, Mouse, Rat
          Host/Isotype
          Mouse / IgG1
          Class
          Monoclonal
          Type
          Antibody
          Clone
          3E10C1
          Immunogen
          ESRRB fusion protein Ag18626
          3D Epitope / Immunogen
          Conjugate
          CoraLite® 594 CoraLite® 594 CoraLite® 594
          • Unconjugated
          • CoraLite Plus 488
          • Request custom conjugation
          Excitation/Emission Max
          593/614 nm
          Form
          Liquid
          Concentration
          1000 µg/mL
          Amount
          100 µg
          Purification
          Affinity chromatography
          Storage buffer
          PBS, pH 7.3, with 50% glycerol, 0.5% BSA
          Contains
          0.05% ProClin 300
          Storage conditions
          -20°C, store in dark
          Shipping conditions
          Wet ice
          Product Specific Information
          Sequence of this protein is as follows: MSSDDRHLGS SCGSFIKTEP SSPSSGIDAL SHHSPSGSSD ASGGFGLALG THANGLDSPP MFAGAGLGGT PCRKSYEDCA SGIMEDSAIK C
          Target Information
          ESRRB (steroid hormone receptor ERR2) plays a role in maintenance of self-renewal and pluripotency of embryonic and trophoblast stem cells through different signaling pathways including FGF signaling pathway and Wnt signaling pathways. Upon FGF signaling pathway activation ESRRB interacts with KDM1A by directly binding to enhancer sites of ELF5 and EOMES and activates their transcription leading to self-renewal of trophoblast stem cells. ESSRB also regulates expression of multiple rod-specific genes and is required for survival of this cell type. Defects in the gene can cause deafness, autosomal recessive, 35 (DFNB35).
          Bioinformatics
          Protein Aliases: ERR beta-2; ERR-beta; ERRbeta; estrogen receptor related 2; Estrogen receptor-like 2; estrogen-related nuclear receptor beta; Estrogen-related receptor beta; estrogen-related receptor, beta; nuclear receptor ERRB2; Nuclear receptor subfamily 3 group B member 2; orphan nuclear receptor; Steroid hormone receptor ERR2; unnamed protein product
          View more View less
          Gene Aliases: DFNB35; ERR beta-2; Err-2; ERR2; ERRb; ERRB2; ERRbeta2; ESRL2; ESRRB; Estrrb; NR3B2
          View more View less
          UniProt ID: (Human) O95718, (Mouse) Q61539, (Rat) P11475
          View more View less
          Entrez Gene ID: (Human) 2103, (Mouse) 26380, (Rat) 299210
          View more View less
          Function(s)
          RNA polymerase II core promoter proximal region sequence-specific DNA binding RNA polymerase II transcription factor activity, sequence-specific DNA binding core promoter proximal region sequence-specific DNA binding RNA polymerase II core binding transcriptional activator activity, RNA polymerase II transcription regulatory region sequence-specific binding DNA binding transcription factor activity, sequence-specific DNA binding steroid hormone receptor activity RNA polymerase II transcription factor activity, ligand-activated sequence-specific DNA binding steroid binding protein binding zinc ion binding estrogen response element binding sequence-specific DNA binding metal ion binding RNA polymerase II sequence-specific DNA binding transcription factor binding sequence-specific double-stranded DNA binding RNA polymerase II distal enhancer sequence-specific DNA binding transcription coactivator activity transcription factor binding
          Process(es)
          embryonic placenta development regulation of transcription, DNA-templated regulation of transcription from RNA polymerase II promoter cell proliferation stem cell division stem cell population maintenance intracellular steroid hormone receptor signaling pathway intracellular receptor signaling pathway somatic stem cell population maintenance cell dedifferentiation photoreceptor cell maintenance positive regulation of transcription, DNA-templated positive regulation of transcription from RNA polymerase II promoter inner ear development morula formation positive regulation of stem cell population maintenance regulation of stem cell division negative regulation of stem cell differentiation in utero embryonic development trophectodermal cellular morphogenesis trophectodermal cell proliferation transcription, DNA-templated steroid hormone mediated signaling pathway
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          Your items have has been added!
          Host server : magellan-search-green-b49b87d85-8dsbr:80/100.66.76.150:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline