Search
Search
Proteintech
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.
Immunogen sequence: RKRQPDGPL GPLYASSNPE YLSASDVFPC SVYVPDEWEV SREKITLLRE LGQGSFGMVY EGNARDIIKG EAETRVAVKT VNESASLRER IEFLNEASVM KGFTCHHVVR LLGVVSKGQP TLVVMELMAH GDLKSYLRSL RPEAENNPGR PPPTLQEMIQ MAAEIADGMA YLNAKKFVHR DLAARNCMVA HDFTVKIGDF GMTRDIYETD YYRKGGKGLL PVRWMAPESL KDGVFTTSSD MWSFGVVLWE ITSLAEQPYQ GLSNEQVLKF VMDGGYLDQP DNCPERVTDL MRMCWQFNPK MRPTFLEIVN LLKDDLHPSF PEVSFFHSEE NKAPESEELE MEFEDMENVP LDRSSHCQRE EAGGRDGGSS LGFKRSYEEH IPYTHMNGGK KNGRILTLPR SNPS (980-1382 aa encoded by BC117172)
INSR is a receptor tyrosine kinase. Binding of insulin to the extracellular domain of INSR stimulates glucose uptake. INSR and IGF-1 receptors share major structural and functional similarity. The earliest cellular response to insulin stimulation is autophosphorylation of tyrosine in INSR. In humans, the INSR gene is located on chromosome 19. Defects in INSR are the cause of various insulin resistance syndromes and IGF-1R defects may also cause some forms of growth retardation.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: CD 220; CD220 antigen; CD220 beta; CD221; HHF 5; IGF-I receptor; insulin receptor precursor; IR; IR 1; IR beta; IR-1; IR1; unnamed protein product
Gene Aliases: CD220; HHF5
UniProt ID: (Human) P06213
Entrez Gene ID: (Human) 3643
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support