Search
The serine–threonine kinase Akt (also known as protein kinase B (PKB) plays a critical role in a broadly influential signaling network. Akt activation serves as a master switch for cellular signaling pathways by generating a multitude of intracellular responses through downstream targets and interacting partners. Akt has been implicated in diseases ranging from cancer and diabetes to neurodegeneration. Akt is one of the most actively studied kinases in both basic research and drug development. We offer a wide range of products to help with Akt research.
Akt/PKB is expressed as three isoforms [1]
Common targets in the Akt/PKB Pathway:
| 4E-BP1 | GβL | PDK1 |
| AR | GSK-3β | PI3K |
| ASK-1 | IκB | PIP2 |
| BAD | IKK | PIP3 |
| Β-CTN | MDM2 | PP2A |
| CASP9 | mTOR | PRAS40 |
| CCND1 | NFAT | PTEN |
| eIF2B | NFκB | Raptor |
| eIF4A | p110 | Rheb |
| eIF4B | p21 | Rictor |
| eIF4E | p27 | RTK |
| FKBP | p53 | SK6 |
| FoxO1 | p85 | TSC1 |
| GATA4 | PDCD4 | TSC2 |
An amino terminal pleckstrin homology (PH) domain, a central serine–threonine catalytic domain, and a small carboxy terminal regulatory domain characterize all the three isoforms. The PH domain binds to phosphatidylinositol-3,4-bisphosphate (PIP2) and phosphatidylinositol-3,4,5-trisphosphate (PIP3), products of PI3K. This binding causes Akt to locate to the plasma membrane, where it becomes phosphorylated by phosphoinositide-dependent kinase 1 (PDK1) on Thr308 in the activation loop of the catalytic domain. This phosphorylation leads to activation. Full activation requires phosphorylation at a second site (Ser473).
The mTOR–rictor complex (mTORC2) is the primary kinase for the second phosphorylation event, although other kinases like Ilk (integrin linked kinase) [2, 3], PDK1 [4, 5], DNA-dependent protein kinase (DNA-PK) [6, 7] and ATM (ataxia telangiectasia mutated) have also been identified [8].
Abnormal activation or blockage of the Akt/PKB pathway is implicated in tumor cell survival and proliferation. Thus, the Akt pathway is an attractive target for anticancer drug discovery [9]. Key targets for tumor suppression include PH domain Leucine-rich repeat Protein Phosphatase (PHLPP), phosphatase and tensin homolog (PTEN) and tuberous sclerosis complex (TSC) [10]. These targets provide suppression at multiple points throughout the pathway. PTEN dephosphorylates PIP3 and stops activation of Akt. PHLPP1/PHLPP2 negatively regulates PI3K and, in turn, regulates Akt [10]. TSC1 and TSC2 are activated by Akt and then hinder mTOR downstream [11].
We offer antibodies, ELISAs, Luminex multiplex assays and growth factors for key targets in the Akt signaling pathway.
Featured below is western blot, ELISA, and Luminex data using Thermo Scientific products.
![Western blot analysis of AKT [pS473]](/content/dam/LifeTech/global/life-sciences/cellanalysis/Images/PATHWAY%20IMAGES%207-15/700256_AKT-Ser473_WB.jpg)
Western blot analysis of AKT [pS473] was performed by loading 20 µg of NIH/3T3 (lane1), NIH/3T3 treated for 10 minutes with 25 ng/ml of PDGF (lane2) and U-87 MG (lane3) cell lysates. Proteins were transferred to a nitrocellulose membrane and blocked with 5 % skim milk for 1 hour at room temperature. AKT [pS473] was detected at ~55 kDa using ABfinity AKT [pS473] Recombinant Rabbit Monoclonal Antibody (Cat. No. 700256) at 0.5 µg-1 µg/ml in 2.5 % skim milk at 4°C overnight on a rocking platform. To confirm specificity, competition was performed with phosphopeptide (10 µg/mL) as shown in the corresponding blot on right. Goat Anti-Rabbit IgG-HRP Secondary Antibody (Cat. No. G21234) at 1:5000 dilution was used and chemiluminescent detection was performed using Novex ECL Chemiluminescent Substrate Reagent Kit (Cat. No. WP20005).
Immunofluorescent analysis of AKT (pT308) was performed on 70% confluent log phase HeLa cells. The cells were fixed with 4% paraformaldehyde for 15 minutes, permeabilized with 0. 25% Triton X-100 for 10 minutes, and blocked with 5% BSA for 1 hour at room temperature. The cells were labeled with ABfinity AKT (pT308) recombinant rabbit monoclonal antibody (Cat. No. 701052) at a dilution of 1:500 in 1% BSA and incubated for 3 hours at room temperature and then labeled with Alexa Fluor 488 goat anti-rabbit IgG secondary antibody (Cat. No. A11008) at a dilution of 1:400 for 30 minutes at room temperature (Panel a: green). Nuclei (Panel b: blue) were stained with SlowFade Gold Antifade Mountant DAPI (Cat. No. S36938). F-actin (Panel c: red) was stained with Alexa Fluor 594 phalloidin (Cat. No. A12381). Panel d is a merged image showing cytoplasmic and perinuclear localization of phosphorylated AKT (pT308). Panel e shows competition with the phospho-AKT (pT308) peptide.
| Product Name | Catalog Number |
| MDM2 Antibody | 337100 |
| GSK3 alpha + beta Antibody | 44610 |
| p53 Antibody | MA512557 |
| Phospho-PRAS40 pThr246 Antibody | 441100G |
| PI3 Kinase p110-alpha Antibody | MA514870 |
| FoxO1 Antibody | MA514846 |
| p21 / Cip1 Antibody | 337000 |
| Tuberin / TSC2 Antibody | AHO1422 |
| Raptor Antibody | 424000 |
| PTEN Antibody | 325800 |
| AKT1 + AKT2 + AKT3 Antibody | MA514999 |
| PDK1 Antibody | MA515797 |
| RICTOR Antibody | MA515681 |
| PP2A C Subunit Antibody | PA517510 |
| Bad Antibody | A16707 |
| P27 Kip1 Antibody | PA527188 |
| IKK alpha Antibody | MA516157 |
| PDCD4 Antibody | PA528150 |
| TSC1 Antibody | PA520131 |
| Phospho-EIF4EBP1pThr36 Antibody | PA512847 |
* This promotion is open to customers in the US and Canada. Discount will apply to orders received by Life Technologies no later than July 31, 2013, or until promotional supplies are depleted, whichever comes first. Customer can use the discount only once. Discount only applies to a maximum of 1 kit per customer. Discount applies to list price in effect at the time order is received by Life Technologies. Cannot be combined with other discounts or promotions. Offer void where prohibited, licensed, or restricted by federal, state, provincial, or local laws or regulation or agency/institutional policy. Other restrictions may apply.