TBS DataManager is a data management software designed to support your laboratory’s specialist protein testing. The software is used in conjunction with the Optilite® Analyser to enhance your laboratory’s efficiency and workflow. This will assist you in obtaining consistent and reliable results for your patients.
TBS DataManager has been optimized to run in conjunction with Binding Site instruments and integrates seamlessly with your current set up, allowing you to run our entire specialist test catalogue on one dedicated platform, with confidence.
Quality control management
| View a snapshot of all daily QCs in the profile screen |
| Color-coded alerts of abnormal QC results and flags where user intervention is required |
| Levey Jennings charts for each QC level |
| Full Westgard rules capability |
| Moving averages |
| All QC result details provided (user who validated QC, regent lot number etc.) |
Operational efficiency
| Reduces analyzer hands on time |
| Simplifies workload and increases laboratory productivity through automated workflows |
| Enables cross-functional collaboration and provides a holistic view due to centralized data processes |
| Ensures consistent performance by providing a common framework to share, compare, search, query, and use data |
| Uniform view of all patient data and test results. Allowing for quick follow-up and continuous monitoring aiding in timely patient care |
Validation and regulatory compliance
| Offers complete traceability of user actions, results and QC |
| Assay Validation Module dedicated to supporting validation procedures |
| User access is controlled by role-based security log in and automated log out |
| Maintenance diary |
| Supports laboratory compliance to ISO 15189 |