A breakthrough in monoclonal​ gammopathy assessment
The EXENT® System is an automated solution that enables M-protein identification and measurement in serum with enhanced sensitivity beyond conventional methods
The EXENT System is the latest innovative solution from the Binding Site, part of Thermo Fisher Scientific​
​The system combines the EXENT Analyser with the EXENT GAM Assay* to provide a fully automated solution to isotype, quantify and track monoclonal proteins throughout the patient journey.​
​The EXENT System is a regulatory compliant and easy to implement solution for routine laboratory use​.
EXENT® System shown before starting the virtual tour
Experience the EXENT® System
Also available on iPhone and iPad
Download on the App Store
Learn more about the EXENT System in the video below:
Watch this short video (01:58) to see the EXENT System in action!
What is the EXENT System?​
The EXENT System integrates immunoassay, MALDI-ToF mass spectrometry and intuitive data review software to provide enhanced analytical sensitivity and specificity for the measurement of monoclonal proteins. The analyzer has been designed to specifically address workflow and interpretation challenges faced by the clinical laboratory.
The EXENT Analyser consists of three integrated modules:​
Prepare
Automatically isolate immunoglobulins using isotype-specific paramagnetic reagents
Analyze
Rapidly analyze samples by MALDI-ToF mass spectrometry with load & click technology
Data Review
EXENT System automated analyzer with blue illuminated workspace, computer monitor, keyboard, and control tower unit
Automatically isolate immunoglobulins using isotype-specific paramagnetic reagents
Rapidly analyze samples by MALDI-ToF mass spectrometry with load & click technology
Easily review & release quantitative results using a simple intuitive interface
Integration for quantification
Optilite® Analyser*
Optilite analyser optimized for special protein testing in laboratory setting
*Optilite® Analyser is not part of the EXENT® Analyser
Primary serum sample to mass spectrometry ready
Immunoassay automatically isolates immunoglobulins using isotype-specific paramagnetic reagents
How does the EXENT System prepare the samples?
Exent System automated immunoassay analyzer for clinical laboratory testing
Rapidly analyze samples using MALDI-ToF mass spectrometry with load & click technology​
Integrated calibration and control routines are combined with automated sample measurement to remove the need for mass spectrometry expertise
How does the EXENT System analyze samples?
EXENT automated protein analysis system
Easily review & release results using an intuitive data analysis software
Automated peak picking and the ability to specifically track monoclonal proteins using their molecular mass reduces complexity and subjectivity when reviewing results​
EXENT System connectivity
The EXENT System is provided with ASTM connectivity
EXENT System automated analyzer with blue illuminated workspace, computer monitor, keyboard, and control tower unit
  • Ordering
  • Released Optilite® result
  • Ordering
LIS/Middleware
  • Released result
  • Released result
  • Released result
  • Ordering
  • Released Optilite® result
Middleware
  • Released result
  • Ordering
Optilite analyser optimized for special protein testing in laboratory setting
Unified patient test ordering
Unified patient test ordering
Automatic data transfer of Optilite assay results to the EXENT Analyser
Automatic data transfer of Optilite assay results to the EXENT Analyser
Automatic upload of released EXENT assay results
Automatic upload of released EXENT assay results
Related content
EXENT GAM Assay
Immunoassays
EXENT GAM Assay
The EXENT Analyser with the EXENT GAM Assay provides a sensitive and specific method for the assessment of M-proteins in patients with monoclonal gammopathies.
EXENT Clinical Case Studies
Expert Insights
EXENT Clinical Case Studies
Learn more about how the EXENT GAM Assay enhances monoclonal gammopathy assessment through our clinical case study series.
CE 2797
Product availability is subject to country specific regulatory requirements. Contact your local representative for availability in your country.
This product is not commercially available in the USA or China. Future commercial availability cannot be guaranteed.
EXENT and Optilite are registered trademarks of The Binding Site Group Limited (Birmingham, UK) in certain countries.
The EXENT® System combines EXENT® Analyser with Immunoglobulin Isotypes (GAM) for the EXENT® Analyser.
*EXENT GAM Assay refers to Immunoglobulin Isotypes (GAM) for the EXENT® Analyser.
Contact Us
Questions about our products or services? Our team is ready to hear them.
Complete this form and we will get back to you