Quality control with LabLink xL™ software
The secure, cloud-based solution for peer-to-peer QC comparison
LabLink xL software is the dedicated peer QC solution for Binding Site controls run on the Optilite™ analyser. Providing a more real time review for controls, allowing the laboratory to monitor their mean values for precision and accuracy within a peer-group using the same system and the same control lot
Timely and real-time QC insights
Identify trends and shifts faster to stay ahead of potential issues
Reporting and analysis tools
Option to receive automated email reports or notifications when reports are ready for download
Complement your QA program
Complementing external quality assurance (EQA) schemes and supports efforts related to ISO15189 accreditation
Facilitates alignment
Enables affiliated groups to stay aligned with each other, ensuring consistent and accurate QC practices across your organization
Seamless and secure access
Secure and cloud-based access at any time, using secure login credentials and based on anonymous peer information
As an integral part of laboratory testing quality assurance programs, peer comparison plays a crucial role. It entails comparing test results between laboratories to detect and rectify any systematic errors or biases that may exist.
LabLink xL software provides comparison of the accuracy and precision of analytical processes between laboratories and peer groups.
How does the LabLink xL system work?
Find out more about the LabLink xL software
Explore the benefits of LabLink xL software
  • Quality control: Peer comparison helps to ensure that laboratory test results are accurate and reliable
  • Accreditation: Supports peer comparison as part of laboratory accreditation
  • Economical: Peer comparison is an economical tool for laboratories, allowing early detection of potential shift in results
Kappa Free monthly trend report showing TSP and lab statistics from Sep 2022 to Aug 2023 with bar charts and QC data
  • Applicable to all special protein parameters that are part of Binding Site assay portfolio
  • No patient data is displayed or transmitted, all peer information is anonymized for end users
  • Allows affiliated groups to create their “own networked group” based on country or organization
  • Secured, long-term QC data storage with AWS™
  • Permissions hierarchy with multiple levels of user access
Optilite analyzer with three reagent compartments and monitor displaying LabLink xL peer review results
Perform all your daily QC activities inside LabLink xL software with interactive charts and graphs. Generate these reports on-demand or set the system to automatically generate reports e.g. at the beginning of each month for every active configured QC control lot. The generated reports can be automatically emailed to your inbox. Alternatively, choose to receive a notification when the reports are ready for download.
The standard reports available in LabLink xL software include the following:
  • Global review
  • Monthly summary report
  • Interactive peer statistics
  • Levey Jennings report
  • Youden Plot report
  • Affiliated lab report
  • Exception Notes report
  • Measurement of uncertainty report
LabLink xL global peer review chart showing Kappa Free test results with monthly trend data and statistical analysis
  • Percentage Bias – Percentage difference between your observed test results and your lab’s target mean
  • Coefficient of Variation (CVi) – Compares your lab’s CV to your target CV to evaluate precision with respect to your chosen target
  • Standard Deviation Index (SDi) – Compares your lab’s mean to your target mean to evaluate accuracy with respect to your chosen target
  • Sigma – Indicates how much your data varies from the total allowable error limit by calculating the number of standard deviations in a given test system that fit within your chosen quality goals
  • Z Score – Measures how many standard deviations away an individual QC result is from the target mean
  • No installation requirements: LabLink xL software is a web-based solution so any PC with web access can be used
  • Available in 15+ languages
  • Manual data entry or csv file data transfer. Download data direct from the Optilite system to a secure USB device and upload to LabLink xL software
LabLink xL software displaying a bell curve graph with peer comparison data for test system performance review
Thumbnail of Optilite analyser from a side angle with a textured background.
Product
Discover the Optilite™ analyser
The Optilite Analyser utilizes proven turbidimetric technology, which is known to be fast, robust and versatile, becoming the main technology for protein diagnostics worldwide
Image of a monitor with TBS data manager dashboard
Product
Intelligent data management with TBS DataManager
TBS DataManager has been optimised to run in conjunction with Binding Site systems and integrates seamlessly with your current set up, allowing you to run our entire specialist test catalogue on one dedicated platform, with confidence
Optilite is a registered trademark of The Binding Site Group Ltd (Birmingham, UK) in certain countries. © 2024, The Binding Site Group Ltd, Birmingham, UK.
© 2024 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific or its subsidiaries.
Product availability is subject to specific regulatory requirements. Contact your local representative for availability in your country.
Other brand or product names may be trademarks of their respective holders.
Contact Us?
Questions about our products or services? Our team is ready to hear them.
Complete this form and we will get back to you