Thermo Fisher Scientific
Your educational resource for biopharma, pharma, environmental, food and agriculture, industrial, and clinical labs
  • Categories
    • Advancing Materials
    • Advancing Mining
    • AnalyteGuru
    • Analyzing Metals
    • Ask a Scientist
    • Behind the Bench
    • Biotech at Scale
    • Clinical Conversations
    • Examining Food
    • Identifying Threats
    • Illuminating Semiconductors
    • Life in Atomic Resolution
    • Life in the Lab
    • OEMpowered
    • The Connected Lab
  • About Us
  • Contact
Accelerating ScienceAnalyteGuru / Ion Chromatography / Ion Chromatography for Vaccine Testing
Ion Chromatography for Vaccine Testing
By AnalyteGuru Staff, 03.06.2020
Viruses still represent a significant threat to human and animal health worldwide. Whether transmitted through water, air, blood, or animal vectors (insects, rodents), viral infections remain a significant threat to life worldwide. In addition to the common viral illnesses of childhood — such as measles, rubella, varicella and mumps — influenza, severe acute respiratory syndrome (SARS), Ebola and dengue, hemorrhagic fevers — such as yellow fever — acquired immune deficiency syndrome (AIDS) and chronic hepatitis (B, C) are some examples of severe viral diseases. Understandably, the fight against viral contamination is a great concern.
Vaccines have been used for centuries to immunize individuals against pathogenic organisms with the goal of averting the associated disease. Many important vaccines use bacterial capsular polysaccharides, or shorter polysaccharides or oligosaccharides, derived from the capsular polysaccharides, conjugated to a protein. Due to the rapidly increasing introduction of Haemophilus influenzaetype b, pneumococcal, meningococcal, group B streptococcal, and Salmonella polysaccharide and corresponding conjugate vaccines worldwide, a reliable and robust analytical method are needed for the quantitative monitoring of intermediate samples generated during fermentation and purification of polysaccharide vaccine components.
Why high-performance anion-exchange chromatography with pulsed amperometric detection (HPAE-PAD)?
Carbohydrates are difficult to analyze using common chromatography and detection methods. They are very polar compounds, exhibit similar structural characteristics, and lack a suitable chromophore. Methods for the liquid chromatographic analysis of carbohydrates often use refractive index (RI) detection. The RI detection methods are sensitive to eluent and sample matrix components. This usually precludes the use of gradients and often requires stringent sample cleanup prior to injection. Pulsed amperometry detects analytes containing functional groups which are oxidized at the applied detection voltage. HPAE-PAD is a major technique used to understand the carbohydrate composition of vaccines and ensure product quality.
Eluents for Polysaccharide Analysis in Vaccines
Carbohydrates are separated as oxyanions at high pH (>12) using hydroxide-based eluents for complex and hybrid oligosaccharides; separations can be facilitated and further improved by using sodium acetate gradients in sodium hydroxide. Preparing and maintaining a hydroxide eluent is very difficult, as the eluent concentration changes due to carbonate absorption. System contamination due to sodium acetate quality is another routine eluent preparation issue.
Thermo Fisher Scientific now offers a new operating mode for RFIC systems, called Dual Eluent Generation Cartridge (Dual EGC) mode. This novel option can replace the manual preparation of the sodium hydroxide/sodium acetate (NaOH/NaOAc) eluent gradients required for analyzing complex carbohydrates.
Dual EGC mode is available on supported instruments, such as the Thermo Scientific™ Dionex™ ICS-6000 HPIC™ System, to support the analysis of complex carbohydrates.
This novel option uses a methanesulfonic acid (MSA) EGC cartridge and a potassium hydroxide (KOH) EGC cartridge in series with each other to generate the extremely reproducible and accurate KOH/KMSA eluent gradient needed for analyzing complex carbohydrates such as oligosaccharides.
Which column is suitable for polysaccharide analysis in vaccines?
The Thermo Scientific™ Dionex™ CarboPac™ series of columns are made specifically for carbohydrate anion-exchange chromatography. These columns permit the separations based on size, charge, degree of branching, and linkage isomerism of mono-, oligo-, and polysaccharides, charged and neutral oligosaccharides. Carbopac columns provide fast, pH-stable and high-resolution mapping and analysis of charged and neutral oligosaccharides with no need for derivatization
Want to know more about carbohydrate analysis by HPAE-PAD?
    • Learn more about CarboPac PA300
    • Download Technical Note on Carbohydrates analysis by HPAE-PAD
    • Download our HPAE-PAD Notebook
    • thermofisher.com/Carbohydrates
    • https://www.thermofisher.com/us/en/home/industrial/chromatography/ion-chromatography-ic/ion-chromato…
References:
    • Vaccine Quality Ensured by High-Performance Anion-Exchange Chromatography with Pulsed Amperometric D…
    • USP General Chapter <1235> Vaccines for Human Use – General Considerations
1975 text image
Innovations of 1975 and the Birth of the First Commercialized Suppressed Ion Chromatograph
The year 1975 was a remarkable period in the history of tech...
Read More
Food packaging
Enhancing PFAS Analysis in Food Contact Materials Using Combustion Ion Chromatography
Per- and polyfluoroalkyl substances (PFAS) have been used in...
Read More
US EPA Method 1621_1200x600
Understanding PFAS and Combustion-Ion Chromatography
Per- and polyfluoroalkyl substances (PFAS) comprise more tha...
Read More
IC Golden Jubilee blog header image
What is the Ion Chromatography Golden Jubilee? Celebrating 50 Years of IC Innovation
The year 2025 marks the golden jubilee of ion chromatography...
Read More
AnalyteGuru Staff
Accelerate Analysis of Trace Elements for Routine Laboratory
Challenging Pharmaceutical Impurity Analyses Part 1: Derivatization