Thermo Fisher Scientific
Your educational resource for biopharma, pharma, environmental, food and agriculture, industrial, and clinical labs
  • Categories
    • Advancing Materials
    • Advancing Mining
    • AnalyteGuru
    • Analyzing Metals
    • Ask a Scientist
    • Behind the Bench
    • Biotech at Scale
    • Clinical Conversations
    • Examining Food
    • Identifying Threats
    • Illuminating Semiconductors
    • Life in Atomic Resolution
    • Life in the Lab
    • OEMpowered
    • The Connected Lab
  • About Us
  • Contact
Accelerating ScienceAnalyteGuru / Proteomics / Top 3 Tools Single-Cell Proteomics Researchers Need for a Big Breakthrough
Top 3 Tools Single-Cell Proteomics Researchers Need for a Big Breakthrough
By Alec Valenta, Product Marketing Specialist, HPLC, Thermo Fisher Scientific 05.24.2023
The emergence of single-cell proteomics (SCP) is set to revolutionize the field of precision medicine – a diagnostic approach that develops treatments to fit specific genetic and molecular characteristics.
Understanding the etiology of complex diseases is rooted in cellular biochemistry. But before you can develop effective and targeted therapeutics, you need to identify how the metabolic pathways, expression and regulation of proteins change in diseased versus normal cells.
Cells in single-cell proteomics
Also, when you analyze large groups of heterogenous cells together (e.g., tissues, plasma, etc.) in traditional proteomics approaches, a significant amount of information about the content of individual cell types is effectively lost or averaged out.
A deep understanding of disease states hinges on insight into the behavior of these individual cell types. The problem is the average “simple” cell has millions of proteins, so you need high-performance LC-MS technology to elucidate changes in protein expression, most importantly, without bias.
That’s where successful proteomics researchers like Karl Mechtler (Institute of Molecular Pathology, Vienna) shine. Our new manuscript in Biorxiv in collaboration with Dr. Mechtler’s group demonstrates a high throughput, proteome coverage, and quantitative precision in single-cell proteomics using our low-flow tools.
The major takeaway points from our groundbreaking work are:
  • Higher LC-MS sensitivity was achieved by employing a low nano-flow rate (100 nL/min) and narrow I.D. columns (50 µm)
  • High sample throughput (100 samples/day) is possible by using a trap-and-elute workflow and optimized methods
  • A 10-min elution window provides high MS utilization (~70 percent) without sacrificing data quality
  • Reproducible inter-lab performance was observed in single-cell and standard diluted samples
  • State-of-the-art separation technology enabled the identification of  >2,200 and >1,700 protein groups from 250 pg diluted standard and single-cell samples, respectively, using library-free DIA searches
Successful proteomics researchers
To our knowledge, this study represents the most comprehensive proteome coverage in the SCP workflow space to date.
Teamwork truly makes the dreamwork!
P.S. Don’t forget to explore our top three proteomics tools today:
  1. Thermo Scientific Vanquish Neo UHPLC System
  2. Thermo Scientific Orbitrap Exploris 480 Mass Spectrometer
  3. Thermo Scientific low-flow columns
Abstract flowing mesh of purple and blue textures with glowing particles.
From Archived FFPE Tissues to Biomarker Insights in a Day
Formalin-fixed, paraffin-embedded (FFPE) tissue is one of th...
Read More
mass spectrometry solutions for clinical research and toxicology
Accelerating Biomarker Discovery and Validation: From Discovery to Clinical Utility
At MSACL 2025, a Thermo Fisher Scientific–sponsored worksh...
Read More
Proteomics software solution
Accelerate Proteomics Research with Smart Automation
Modern proteomics research produces vast quantities of compl...
Read More
Dr. Stacy Malaker
Mucinomics: The Next Frontier of Mass Spectrometry
Mucinomics, the study of mucin domain glycoproteins, is emer...
Read More
Alec Valenta
Alec Valenta is a Product Marketing Specialist for Chromatography and Mass Spectrometry, Thermo Fisher Scientific
Quality Risk Management: Don’t Discard Standard Lab Practices
Complex Carbohydrate Analysis by HPAE-PAD Made Easier: ‘Just Add Water’