FTIR Basics: Videos
FTIR stands for Fourier transform infrared, the preferred method of infrared spectroscopy. When IR radiation is passed through a sample, some radiation is absorbed by the sample and some passes through (is transmitted). The resulting signal at the detector is a spectrum representing a molecular ‘fingerprint’ of the sample. The usefulness of infrared spectroscopy arises because different chemical structures (molecules) produce different spectral fingerprints. The Fourier Transform converts the detector output to an interpretable spectrum. The FTIR generates spectra with patterns that provide structural insights.
Dive deeper into these subjects by watching these three tutorials.
Three-Part Video Series
This three-part video series teaches the basics of the most common FTIR sampling technique: Attenuated Total Reflection (ATR).
7-Minute Video: The Basics of The Attenuated Total Reflection Technique
This is the first video in a series that teaches the basics of the most common FTIR sampling techniques: Attenuated Total Reflection (ATR). How do people use ATR and why? Find out in this classroom-style instruction video. In less than seven minutes, I’ll show you how infrared light moves through diamond and germanium crystals, what affects the depth of penetration can have on the sample, and the most critical things to consider when choosing between crystals.
Which crystal should you use for Carbon Black Rubber? View the ATR-Basics video and find out.
3-Minute Video: The ATR Sample Preparation
This is the second video in a series that teaches the basics of the most common FTIR sampling techniques: Attenuated Total Reflection (ATR). This video addresses ATR accessories and sample preparation. Get a close-up view of the diamond crystal, pressure anvil, and gasket in an FTIR instrument, and learn how they are used to analyze samples.
In less than four minutes, I will show you how the various components of an ATR accessory and explain how they aid in analyzing samples.
What would happen if you tried to run a black rubber ring sample on the diamond crystal? View the ATR-Sample Preparation video and find out.
6-Minute Video: FTIR Sampling Techniques: Attenuated Total Reflectance Data Collection
This is the third video in a series that teaches the basics of the most common FTIR sampling techniques: Attenuated Total Reflection (ATR). This video addresses ATR data collections. See various samples, including solids and liquids — being analyzed in real time with an FTIR instrument, and see the analysis results in seconds.
What spectra would you see if you scanned your own plastic store loyalty card? How about some granular powder? View the 6-minute ATR-Sample Preparation video and find out.
Further Reading
Leave a Reply