Where Did Those Gemstones Come From?
Energy Dispersive X-Ray Fluorescence (EDXRF) is a nondestructive tool for determining the authenticity of colored gemstones and their geographical origin
Energy Dispersive X-Ray Fluorescence (EDXRF) is a nondestructive tool for determining the authenticity of colored gemstones and their geographical origin
Determining the Origin and Authenticity of Gemstones using EDXRF
Precious gemstones include diamonds and colored stones such as rubies, emeralds, and sapphires. While diamonds remain the most expensive gemstone, the colored gems are rapidly gaining in popularity, as evidenced by their increasing use in engagement rings. As the value of colored gemstones increases, so does the need to verify their authenticity and source.
How Do Gemstones Form?
Most gemstones form in the Earth’s crust, approximately 3 to 25 miles beneath the Earth’s surface. Two gemstones, diamonds and peridot, are found much deeper in the Earth. Diamond forms in “kimberlite pipes” that originate in the Earth’s mantle (>125 miles) and end at the surface. Molten magma running through these pipes usually reaches the surface as lava and cools off, but if it doesn’t reach the surface, it cools more slowly, crystallizes, and forms coarse-grained minerals.
The first type of rock, igneous rock, is also formed by the cooling and crystallization of magma beneath the earth’s surface (intrusive or plutonic), or it can be formed when lava flows at the earth surface (extrusive or volcanic). Interlocking crystals grow within intrusive igneous rock and may form gemstones depending on the elements present, the cooling time, and the environment. The slower cooling (in the deep environment), the larger gemstones.
Gemstones found in igneous rock include the quartzes (including amethyst, citrine and ametrine), the garnets, moonstone, apatite, diamond, spinel, tanzanite, tourmaline, topaz and zircon. Some of these gemstones form in pegmatites and hydrothermal veins that are genetically related to igneous rocks.
Sedimentary rock is the second type and gives rise to the transported minerals such as jasper, malachite, opal and zircon. Sedimentary rocks are formed when rock is worn down and the fragments are carried by water or the wind, and they are finally compressed together over time.
The third variety, metamorphic rock, is formed when intense underground heat or pressure changes existing rocks. The beryls (emerald, morganite and aquamarine), jade, lapis lazuli, turquoise, spinel, ruby, sapphire and zircon are formed in metamorphic rock.
How Are Gemstones Found?
Diamond mining is an industrialized process. Diamonds are typically recovered through pit mining, which involves the use of heavy machinery, hydraulic shovels, and trucks to mine diamonds from kimberlite pipes. Colored gemstone (other than diamond) mining, however, is a very different process. Mining for precious colored gemstones is rigorous and time-consuming because the deposits are few and when found, tend to be characterized by small quantities of gems scattered throughout a large amount of rock. Modern mining techniques are of little value in these circumstances, and the deposits are often too small to be profitable for major mine outfits, who leave them to small, independent miners who rely on the same manual techniques they have been using for decades. Nevertheless, in recent years, several major mining companies have entered the gemstone market with new strategies for employing modern mining practice.
How Can You Be Sure a Colored Gemstone Is Real?
As with gold, gemstone counterfeits, or synthetic gemstones, can be mistaken for the real thing. Another important factor in establishing the value of a gemstone is to know where it came from, both geologically and geographically, which can’t be determined visually.
Energy Dispersive X-Ray Fluorescence (EDXRF) is an important tool for the determination of the authenticity of colored gemstones and their geographical origin. Depending on the geological setting, precious gemstones like rubies, emeralds or sapphires from different origins often exhibit a characteristic combination of trace elements at different concentrations. As an example, identification and quantification of such elements may allow tracking an emerald down to its location of origin such as Colombia, Brazil, Afghanistan, Zambia or Zimbabwe. Similarly, the presence of certain trace elements also helps to distinguish between a valuable naturally formed gemstone (e.g. ruby) and a quasi-worthless synthetic crystal (e.g. synthetic ruby.)
Ali Somarin
Written by:
Ali Somarin, PhD, P.Geo
Chief Geologist, Thermo Fisher Scientific
Ali Somarin performed research, drafted application notes, and provided technical knowledge on the application of portable XRF in various fields (mining, oil/gas, environmental, gems) for Thermo Fisher Scientific during 2010-2015.
Read more Somarin, Ali
Comments
    • nitonxrf says
      Hi Hawk — Thanks for the question. Our Chief Geologist says: “XRF
      is a non-destructive method meaning that after assay, you won’t see anything at
      the surface of your sample/gem. And you can analyze the same gem or any point
      on it as many times as you like with no destruction.“
  1. Greg says
    I have not checked in here for a while as I thought it was getting boring, but
    the last several posts are good quality so I guess I’ll add you
    back to my daily bloglist. You deserve it friend :
    )
    • Marlene Gasdia-Cochrane says
      Thanks for your comment, Greg. If you would like to offer any topics that would be of more interest to you, please let us know. We are always looking for new topics to educate our audience. You may even want to author a guest blog. Let us know.
  2. Bulk Gemstones says
    Nice i didn’t know about EDXRF Method, Your other articles are good. They are very informative.
    Thank you,
  3. Jahnavi Jaanu says
    Awesome! We read a lot of posts regarding the origin of the gemstones but our curiosity is never satisfied. Thank you for all the details shared to satisfy this knowledge hunger.
  4. Cassondra says
    Hi I found a rock when I was little playing at the gravel pit of my parents farm. I’ve always wondered what it was. Could you help?