Flexible lab water systems designed for your application
There are many models and configurations to choose from.  We’ve made it easy to find the right system for your needs by selecting from the five categories below which will launch an interactive selection guide.
Which water purification system is right for you?
Meet your most critical and everyday application needs, from electrodeionization to reverse osmosis and distillation with Thermo Scientific Barnstead Water Purification Systems. Depending on your requirements, we have systems that will produce Type 2 water and Reverse Osmosis water for general lab purpose, Type 1 water for analytical and life science applications, or a combination of water type capabilities.   Browse the systems below by water type that best meets your requirements and view our model selector tool for more detailed model information.   
Depending on configuration, your system may require certain accessories and optional components.  Be sure to note these accessories when requesting your quote (required and optional accessories are listed with each system in the selector guide).
Barnstead Water Purification Systems overview
Type 1 – Ultrapure
water systems 
Type 2 – Pure w
ater systems
Reverse osmosis
water systems
Water distillation systems
Cartridge systems
Typical applications and interest areas
Analytical
HPLC
Gas chromatography
Ion chromatography
ICP spectroscopy
Mass spectroscopy
Atomic absorption
Total organic carbon
Life sciences
Genomics
Proteomics
Immunology
Pharmacology
Cell and tissue culture
Drug discovery
Molecular biology
Microbiology
Semiconductor
General  lab purpose
Autoclave
Humidification
Glassware wash/rinse
General lab equipment
Feed water to type 1 water systems
Media prep
Buffer prep
Chemical and biochemical reagent prep
General lab purpose
Autoclave
Humidification
Glassware wash/rinse
General lab equipment (water baths, incubators, etc)
Life sciences
Microbiology
Cell and tissue culture 
General lab purpose
Autoclave
Humidification
Glassware wash/rinse
General lab equipment
 Feed Water to Type 1 Water Systems
General lab purpose
Autoclave
Humidification
Glassware wash/rinse
General lab equipment (water baths, incubators, etc)
Description
Type 1 ultrapure water is the highest water purity and can be made pyrogen-, nuclease-, bacteria- and particulate-free.
Type 2 water is produced through reverse osmosis, deionization and distillation processes. It provides a superb source of feedwater for Type 1 Water systems.
Reverse osmosis is commonly used to supply purified water for water baths, dishwashers, autoclaves and feedwater to obtain pure and ultrapure water.
A lab water distillation system is one of the broadest removal capabilities of any single form of water purification. No consumables needed.
Cartridge systems are crucial components to any pure water system and are easy to operate and maintain.
Flow rates
Up to 1L/min to 2L/min
Up to 0.6L/min to 40L/hr
Model dependent
Up to 7L/hr to 40L/hr
Model dependent
Up to 1L/hr to 6L/hr
Model dependent
Up to 0.63L/min to 4L/min
Capacity
Storage capacity from 15L/day to 200L/day
Model dependent
Storage capacity from 30L to 100L
Model dependent
Storage capacity from 30L to 100L
Model dependent
Storage capacity from 6L to 45L 
Model dependent
Storage capacity from 1 to 30 L/day depending on feed water
Feedwater
Pretreated or tap
Tap
Tap
Tap
Pretreated options for Megapure 
Pretreated or tap
Monitoring
Resistivity/co https nductivity
Total Organic Carbon
RO permeate and system resistivity/conductivity
Feedwater and RO conductivity
Resistivity/conductivity
Models
GenPure
GenPure Pro
GenPure xCAD Plus
Smart2Pure
Smart2Pure Pro
Aquanex
MicroPure
Labtower EDI
E-Pure
Pacific TII
LabTower TII
Mega-Pure glass stills
Pacific RO
LabTower RO
Mega-Pure stills
B-Pure
B-Pure ½ Size filters
Bantam Deionizer
Hose nipple cartridges
Aquanex Ultrapure Water Purification System
Effortless purity redefined.