Click below to check your order status.
Note, order lookup is only supported in the following countries: United States, Canada.
If you are outside one of those countries please reach out to One Lambda Customer Support to submit an order status inquiry.
The new One Lambda™ AllType™ FASTPlex™ NGS HLA assay for Illumina™ and Ion GeneStudio™ systems has several advantages over its previous version , including an optimized primer mix for 11 HLA loci, more robust PCR amplification, greater on-target accuracy, an improved DRB primer design, and better locus balance. Exon 1 can be included in the multiplex assay to improve HLA allele resolution. Only one tube is needed to pool 96 samples for NGS library preparation since each sample is given a unique barcode, and only one universal adapter is required. The assay takes less than seven hours to complete and less than 90 minutes of hands-on time.
Speaker:
Peter Brescia
Senior Technical Manager, HLA and MMDx | Thermo Fisher Scientific
West Hills, CA USA
Recorded:
Nov 3, 2021
Support
Please sign in to access your account. Customers must log in to access some content, recorded webinars, training materials, documents, and software.
Please provide the following to setup your user account. If you do not know your center ID, please leave the center ID field blank.