Pierce™ Sulfhydryl Addition Kit
Thermo Scientific™

Pierce™ Sulfhydryl Addition Kit

The Thermo Scientific Pierce Sulfhydryl Addition Kit contains all necessary components to introduce free sulfhydryl groups into any primary amine-containing자세히 알아보기
Have Questions?
카탈로그 번호수량
234601 Reaction
카탈로그 번호 23460
제품 가격(KRW)
466,000
온라인 행사
Ends: 30-Sep-2026
548,000
할인액 82,000 (15%)
Each
수량:
1 Reaction
대량 주문 또는 맞춤형 요청
제품 가격(KRW)
466,000
온라인 행사
Ends: 30-Sep-2026
548,000
할인액 82,000 (15%)
Each
The Thermo Scientific Pierce Sulfhydryl Addition Kit contains all necessary components to introduce free sulfhydryl groups into any primary amine-containing molecule, purify the modified molecule and quantify the added sulfhydryl groups. SATA is a short-chain (2.8 angstrom spacer arm) reagent for covalent modification of primary amines and addition of a protected yet exposable sulfhydryl group, enabling heterobifunctional crosslinking strategies.

Features of SATA:

• Adds a protected sulfhydryl that can be deprotected by hydroxylamine
• Allows long-term storage of the sulfhydryl-modified molecule
• Forms cleavable disulfide bonds with other sulfhydryl-containing molecules
• Reacts with primary amines (e.g., lysine residues proteins) to form stable amide bonds
• Preserves protein activity with its mild, non-denaturing reaction conditions

SATA (N-succinimidyl S-acetylthioacetate) adds sulfhydryl groups to proteins and other amine-containing molecules in a protected form. The modified molecule can be stored indefinitely and treated with hydroxylamine to expose the labile sulfhydryl group when needed for conjugation reactions. SATA contains an N-hydroxysuccinimide (NHS) ester, which forms a stable, covalent amide bond with primary amines (i.e., lysine residues and the amino termini of proteins) and releases NHS as a by-product. De-protection (deacylation) to generate a free sulfhydryl is accomplished using hydroxylamine-HCl.

Sulfhydryl groups present on proteins, peptides and other compounds are important in protein chemistry/modification reactions. Frequently, thiols are unavailable or absent within the molecules of interest. Several reagents and techniques are available for introducing sulfhydryl groups or disulfides into proteins and peptides, including Traut's Reagent, variants of SPDP, and variants of SATA.

Related Products
Pierce™ SATA (N-succinimidyl S-acetylthioacetate)
Pierce™ SATP
Pierce™ SAT(PEG)4
For Research Use Only. Not for use in diagnostic procedures.
사양
형태Powder
라벨 유형Conjugating Groups
라벨 또는 염료Sulfhydryl Group
제품라인Pierce
수량1 Reaction
반응성 부분Active Ester, Succinimidyl Ester, NHS Ester
시약 유형SATA
용해도DMF, DMSO
화학물질 반응성Amine
교차결합제 유형Heterobifunctional
제품 유형Crosslinker
스페이서Short (<10 Å)
Unit SizeEach
구성 및 보관
Sufficient For: One modification experiment with 0.5 to 5 mg of purified protein
• SATA, 2 mg
• Hydroxylamine·HCl, 5 mg
• Conjugation Buffer Stock (10X), 20 mL
• PBS pack (makes 500 mL), 1 pack
• Dimethylformamide (DMF), 1 mL
• Dextran Desalting Column, 1 x 5 mL
• Column Extender, 1
• Ellman's Reagent (DTNB), 2 mg
• Cysteine·HCl, 20 mg

Store at 4°C.