Pierce™ Hydroxylamine-HCl
Thermo Scientific™
Pierce™ Hydroxylamine-HCl
Thermo Scientific Pierce Hydroxylamine-HCl has many uses, which include cleaving Asn-Gly peptide bonds and deprotecting (exposing) sulfhydryls on SATA-modified moleculesRead more
Catalog NumberQuantity
2610325 g
Catalog number 26103
Price (GBP)
71.50
Each
Quantity:
25 g
Request bulk or custom format
Price (GBP)
71.50
Each
Thermo Scientific Pierce Hydroxylamine-HCl has many uses, which include cleaving Asn-Gly peptide bonds and deprotecting (exposing) sulfhydryls on SATA-modified molecules for conjugation.
Features of Hydroxylamine-HCl:
• Quench amine-labeling or crosslinking reactions (e.g., with NHS esters)
• Expose protected sulfhydryl groups on SATA-modified molecules
• Cleave carbonyl-containing crosslinkers such as EGS and Sulfo-EGS
Hydroxylamine·HCl is a strong reducing agent that is useful in biochemical crosslinking applications, including the deacetylation of SATA and chemical cleavage of EGS and Sulfo-EGS. Hydroxylamine converts carbonyl compounds (aldehydes and ketones) to their oxime derivatives in the presence of a weak base. Therefore, crosslinkers and other compounds that contain a carbonyl within their structure are cleavable with hydroxylamine·HCl.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
CAS5470-11-1
FormSolid
Product LinePierce
Quantity25 g
Reagent TypeSATA
Product TypeHydroxylamine Hydrochloride
Unit SizeEach
Contents & Storage
Upon receipt store desiccated at room temperature. Product shipped at ambient temperature.