Search
Search

Entrez Gene ID: | 1404 |
Gene Name: | hyaluronan and proteoglycan link protein 1 |
Gene Aliases: |
CRT1, CRTL1 |
Location: |
Chr.5:83638198-83721210 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| HAPLN1 | NM_001884.3 | NP_001875.1 | ||
| XM_017009051.1 | XP_016864540.1 | |||
| XM_017009053.1 | XP_016864542.1 | |||
| AK313713.1 | ||||
| U43328.1 | AAA85216.1 | |||
| X17405.1 | CAA35462.1 | |||
| X78076.1 |
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv830367 | Chr.5:83531833 - 83742562 on Build GRCh38 | Gain+Loss |
|
| nsv1015852 | Chr.5:83404850 - 84499122 on Build GRCh38 | Gain |
|
Set Membership: |
|