Search
Search

Entrez Gene ID: | 114792 |
Gene Name: | kelch like family member 32 |
Gene Aliases: |
BKLHD5, KIAA1900, UG0030H05, dJ21F7.1 |
Location: |
Chr.6:96898083-97142949 on Build GRCh38 |
Assay Gene Location: | Within Intron 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| KLHL32 | NM_001286250.1 | NP_001273179.1 | ||
| NM_001286251.1 | NP_001273180.1 | |||
| NM_001286252.1 | NP_001273181.1 | |||
| NM_001323252.1 | NP_001310181.1 | |||
| NM_001323253.1 | NP_001310182.1 | |||
| NM_001323254.1 | NP_001310183.1 | |||
| NM_001323255.1 | NP_001310184.1 | |||
| NM_001323256.1 | NP_001310185.1 | |||
| NM_001323257.1 | NP_001310186.1 | |||
| NM_001323258.1 | NP_001310187.1 | |||
| NM_001323259.1 | NP_001310188.1 | |||
| NM_001323260.1 | NP_001310189.1 | |||
| NM_052904.3 | NP_443136.2 | |||
| NR_104421.1 | ||||
| NR_136580.1 | ||||
| AB067487.1 | BAB67793.1 | |||
| AF351610.1 | AAN76510.1 | |||
| AK055292.1 | BAB70899.1 | |||
| AK295479.1 | ||||
| AK297240.1 | ||||
| BC071627.1 | AAH71627.1 | |||
| DA332224.1 |
Set Membership: |
|