Search
Search

Entrez Gene ID: | 7247 |
Gene Name: | translin |
Gene Aliases: |
BCLF-1, C3PO, RCHF1, REHF-1, TBRBP, TRSLN |
Location: |
Chr.2:121755545-121767853 on Build GRCh38 |
Assay Gene Location: | Within Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TSN | NM_001261401.1 | 5 | 2725 | NP_001248330.1 |
| NM_004622.2 | 6 | 2805 | NP_004613.1 | |
| NR_048556.1 | 5 | 2711 | ||
| NR_048557.1 | 4 | 2614 | ||
| NR_048558.1 | 3 | 2498 | ||
| NR_048559.1 | 4 | 2722 | ||
| AL122099.1 | 1 | 120 | ||
| X78627.1 | CAA55341.1 |
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv834351 | Chr.2:121660348 - 121891697 on Build GRCh38 | Loss |
|
| nsv1011665 | Chr.2:121423564 - 121788408 on Build GRCh38 | Gain |
|
| nsv459163 | Chr.2:121573899 - 122358430 on Build GRCh38 | Gain |
|
| esv3592209 | Chr.2:121558675 - 122131149 on Build GRCh38 | Gain |
|
| esv3592217 | Chr.2:121720414 - 121800640 on Build GRCh38 | Gain |
|
Set Membership: |
|