Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ? Create Account
  • Applications
    • Real-Time PCR
    • Oligos, Primers, Probes and Genes
    • Cloning
    • Protein Biology
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Cell Analysis
    • Mass Spectrometry
    • Chromatography
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Online Order
    • Custom Primers & TaqMan Probes
    • miRNA Mimics & Inhibitors
    • Stealth RNAi / siRNA
    • Silencer Select siRNAs
    • Custom DNA Oligos
    • GeneArt Services
    • GeneArt Strings DNA Fragments
    • TrueGuide CRISPR gRNA
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all online orderable products
  • Services
    • Custom Services
    • Instrument Qualification Services
    • Technical Services
    • Pipette Services
    • Sample Request
    • Instrument Maintenance Services
    • Repairs and Relocation Services
    • OEM and Licensing Services
    • Events, Seminars, Training
    • Unity Lab Services
    • See all services
  • Support
    • Catalogs
    • Application Notes
    • Safety Data Sheets
    • Legal and Regulatory
    • Certificates of Analysis Search
    • Nunc/Nalgene Product Certificates
    • e-learning
    • FAQs
    • Learning Centers
    • Technical Support Centers
    • See all help and support topics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign In
            Sign In
            Don't have an account ? Create Account
            • Connect Your Lab
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › 462713_mat
          Assay Name: mmu-miR-16-2*
          miRBase Accession Number: MI0000566
          miRBase Version: v22.1
          Chromosome Location: Chr.3: 69009902 - 69009996 [+] on Build GRCm38
          Mature miRNA Sequence: ACCAAUAUUAUUGUGCUGCUUU
          Species: Mouse, Cricetulus griseus, Goat
          Product Type: TaqMan™ MicroRNA Assay
          Assay ID 462713_mat
          Size
          Availability Inventoried
          Catalog # 4427975
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Family ID:

          MIPF0000006, mir-15

          Mature miRNA Sequence:

          ACCAAUAUUAUUGUGCUGCUUU

          Mature miRNA Details:



          Species miRBase ID miRBase Accession Number
          Mouse mmu-miR-16-2-3p MIMAT0017018
          Cricetulus griseus cgr-miR-16 MIMAT0041151
          Goat chi-miR-16b-3p MIMAT0035997

          Stem-loop Details



          Mouse

          Stem-loop ID mmu-mir-16-2
          Stem-loop Accession # MI0000566
          Stem-loop Sequence
          CAUGCUUGUUCCACUCUAGCAGCACGUAAAUAUUGGCGUAGUGAAAUAAAUAUUAAACACCAAUAUUAUUGUGCUGCUUUAGUGUGACAGGGAUA
          Chromosome Location Chr. 3 - 69009902 - 69009996 [+] on Build GRCm38
          Mature MicroRNA miRBase Accession #: MIMAT0017018
          miRBase ID: mmu-miR-16-2-3p
          Mature miRNA Sequence: ACCAAUAUUAUUGUGCUGCUUU
          Chromosome Location: Chr. 3 - 69009902 - 69009996 [+] on Build GRCm38

          Cricetulus griseus

          Stem-loop ID cgr-mir-16-2
          Stem-loop Accession # MI0033080
          Stem-loop Sequence
          CAUACUUGUUCCGCUCUAGCAGCACGUAAAUAUUGGCGUAGUGAAAUAAAUAUUAAACACCAAUAUUAUUGUGCUGCUUUAGUGUGACAGGCAUA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0041151
          miRBase ID: cgr-miR-16
          Mature miRNA Sequence: ACCAAUAUUAUUGUGCUGCUUU
          Chromosome Location: NA

          Goat

          Stem-loop ID chi-mir-16b
          Stem-loop Accession # MI0030647
          Stem-loop Sequence
          CUGACAUACUUGUUCCGCUCUAGCAGCACGUAAAUAUUGGGGUAGUAAAAUAAAUAUUAAACACCAAUAUUAUUGUGCUGCUUUAGCGUGACAGGGA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0035997
          miRBase ID: chi-miR-16b-3p
          Mature miRNA Sequence: ACCAAUAUUAUUGUGCUGCUUU
          Chromosome Location: NA


          Back To Top

          More Information




          Related Products

          TaqMan™ Pri-miRNA Assay : Mm03306169_pri
          Ambion® Anti-miR™ miRNA Inhibitor : AM19244
          Ambion® Pre-miR™ miRNA Precursor : PM19244
          mirVana® miRNA inhibitor : MH19244
          mirVana® miRNA mimic : MC19244
          TaqMan™ Advanced miRNA Assay : mmu481555_mir


          miRBase Alias:

          Mouse: mmu-miR-16-2* (v17)

          Back To Top

          Related Products

          • TaqMan® Universal Master Mix II, with UNG
          • TaqMan® Universal Master Mix II, no UNG
          • TaqMan® MicroRNA Reverse Transcription Kit
          Ordering Plus Icon Minus Icon
          • Online Orderable Products
          • Privacy Policy for Online Ordering
          • Act on Specified Commercial Transactions
          • About Returns
          • About Displayed Prices
          • About Shipping Fees
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Study information disclosure
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Seminars
          • Blog
          About Thermo Fisher Plus Icon Minus Icon
          • Corporate Information
          • Social Responsibility (CSR)
          • Enhancing Customer Experience
          • Careers Careers
          • News
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Japan flag icon
          Japan

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-b2k9d:80/100.66.79.173:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline