Search
Search

| Assay Name: | mmu-miR-16-2-3p |
|---|---|
| miRBase Accession Number: | MI0000566 |
| miRBase Version: | v22.1 |
| Mature miRNA Sequence: | ACCAAUAUUAUUGUGCUGCUUU |
| Species: | Mouse, Cricetulus griseus, Goat |
| Product Type: | Ambion® Anti-miR™ miRNA Inhibitor |
Gene Family ID: | MIPF0000006, mir-15 |
Mature miRNA Sequence: | ACCAAUAUUAUUGUGCUGCUUU |
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Mouse | mmu-miR-16-2-3p | MIMAT0017018 |
| Cricetulus griseus | cgr-miR-16 | MIMAT0041151 |
| Goat | chi-miR-16b-3p | MIMAT0035997 |