Search
Search

| Assay Name: | hsa-miR-30a-3p |
|---|---|
| miRBase Accession Number: | MI0000088 |
| miRBase Version: | v22.1 |
| Mature miRNA Sequence: | CUUUCAGUCGGAUGUUUGCAGC |
| Species: | Human, Mouse, Rat, Alligator mississippiensis, Anolis carolinensis, Astatotilapia burtoni, Chicken, Chrysemys picta, Columba livia, Cricetulus griseus, Dasypus novemcinctus, Gorilla gorilla, Guinea pig, Ictalurus punctatus, Ophiophagus hannah, Ornithorhynchus anatinus, Pan paniscus, Pan troglodytes, Pig, Pongo pygmaeus, Python bivittatus, Rabbit, Rhesus monkey, Salmo salar, Zebrafish |
| Product Type: | TaqMan™ MicroRNA Assay |
Gene Family ID: | MIPF0000005, mir-30 |
Mature miRNA Sequence: | CUUUCAGUCGGAUGUUUGCAGC |
| Species | miRBase ID | miRBase Accession Number | |
|---|---|---|---|
| Human | hsa-miR-30a-3p | MIMAT0000088 | |
| Mouse | mmu-miR-30a-3p | MIMAT0000129 | |
| Rat | rno-miR-30a-3p | MIMAT0000809 | |
| Alligator mississippiensis | ami-miR-30a-3p | MIMAT0038078 | |
| Anolis carolinensis | aca-miR-30a-3p | MIMAT0021921 | |
| Astatotilapia burtoni | abu-miR-30a-3p | MIMAT0042058 | |
| View More Rows | |||
| Chicken | gga-miR-30a-3p | MIMAT0001136 | |
| Chrysemys picta | cpi-miR-30a-3p | MIMAT0037675 | |
| Columba livia | cli-miR-30a-3p | MIMAT0038452 | |
| Columba livia | cli-miR-30e-3p | MIMAT0038459 | |
| Cricetulus griseus | cgr-miR-30a-3p | MIMAT0023895 | |
| Dasypus novemcinctus | dno-miR-30a-3p | MIMAT0047557 | |
| Gorilla gorilla | ggo-miR-30a-3p | MIMAT0002374 | |
| Guinea pig | cpo-miR-30a-3p | MIMAT0046904 | |
| Ictalurus punctatus | ipu-miR-30e | MIMAT0029500 | |
| Ophiophagus hannah | oha-miR-30a-3p | MIMAT0036889 | |
| Ornithorhynchus anatinus | oan-miR-30a-3p | MIMAT0006914 | |
| Pan paniscus | ppa-miR-30a-3p | MIMAT0002378 | |
| Pan troglodytes | ptr-miR-30a-3p | MIMAT0002372 | |
| Pig | ssc-miR-30a-3p | MIMAT0015300 | |
| Pongo pygmaeus | ppy-miR-30a-3p | MIMAT0002376 | |
| Python bivittatus | pbv-miR-30a-3p | MIMAT0038871 | |
| Rabbit | ocu-miR-30a-3p | MIMAT0048151 | |
| Rhesus monkey | mml-miR-30a-3p | MIMAT0002370 | |
| Salmo salar | ssa-miR-30c-3p | MIMAT0032607 | |
| Salmo salar | ssa-miR-30d-3p | MIMAT0032609 | |
| Zebrafish | dre-miR-30e-3p | MIMAT0003402 | |