Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › 478273_mir

          Specificity testing was performed using human anti-targets.

          Assay Name: hsa-miR-30a-3p
          Stem-loop Accession Number: MI0000088
          miRBase Version: v22.1
          Chromosome Location: Chr.6: 71403551 - 71403621 [-] on Build GRCh38
          Mature miRNA Sequence: CUUUCAGUCGGAUGUUUGCAGC
          Species: Human, Mouse, Rat, Alligator mississippiensis, Anolis carolinensis, Astatotilapia burtoni, Chicken, Chrysemys picta, Columba livia, Cricetulus griseus, Dasypus novemcinctus, Gorilla gorilla, Guinea pig, Ictalurus punctatus, Ophiophagus hannah, Ornithorhynchus anatinus, Pan paniscus, Pan troglodytes, Pig, Pongo pygmaeus, Python bivittatus, Rabbit, Rhesus monkey, Salmo salar, Zebrafish
          Product Type: TaqMan™ Advanced miRNA Assay
          Assay ID 478273_mir
          Size S: 250 rxns
          Availability Inventoried
          Catalog # A25576
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Family ID:

          MIPF0000005, mir-30

          Mature miRNA Sequence:

          CUUUCAGUCGGAUGUUUGCAGC

          miRBase Details:



          Species miRBase ID miRBase Accession Number
          Human hsa-miR-30a-3p MIMAT0000088
          Mouse mmu-miR-30a-3p MIMAT0000129
          Rat rno-miR-30a-3p MIMAT0000809
          Alligator mississippiensis ami-miR-30a-3p MIMAT0038078
          Anolis carolinensis aca-miR-30a-3p MIMAT0021921
          Astatotilapia burtoni abu-miR-30a-3p MIMAT0042058
          View More Rows
          Chicken gga-miR-30a-3p MIMAT0001136
          Chrysemys picta cpi-miR-30a-3p MIMAT0037675
          Columba livia cli-miR-30a-3p MIMAT0038452
          Columba livia cli-miR-30e-3p MIMAT0038459
          Cricetulus griseus cgr-miR-30a-3p MIMAT0023895
          Dasypus novemcinctus dno-miR-30a-3p MIMAT0047557
          Gorilla gorilla ggo-miR-30a-3p MIMAT0002374
          Guinea pig cpo-miR-30a-3p MIMAT0046904
          Ictalurus punctatus ipu-miR-30e MIMAT0029500
          Ophiophagus hannah oha-miR-30a-3p MIMAT0036889
          Ornithorhynchus anatinus oan-miR-30a-3p MIMAT0006914
          Pan paniscus ppa-miR-30a-3p MIMAT0002378
          Pan troglodytes ptr-miR-30a-3p MIMAT0002372
          Pig ssc-miR-30a-3p MIMAT0015300
          Pongo pygmaeus ppy-miR-30a-3p MIMAT0002376
          Python bivittatus pbv-miR-30a-3p MIMAT0038871
          Rabbit ocu-miR-30a-3p MIMAT0048151
          Rhesus monkey mml-miR-30a-3p MIMAT0002370
          Salmo salar ssa-miR-30c-3p MIMAT0032607
          Salmo salar ssa-miR-30d-3p MIMAT0032609
          Zebrafish dre-miR-30e-3p MIMAT0003402

          Stem-loop Details



          Human

          Stem-loop ID hsa-mir-30a
          Stem-loop Accession # MI0000088
          Stem-loop Sequence
          GCGACUGUAAACAUCCUCGACUGGAAGCUGUGAAGCCACAGAUGGGCUUUCAGUCGGAUGUUUGCAGCUGC
          Chromosome Location Chr. 6 - 71403551 - 71403621 [-] on Build GRCh38
          Mature MicroRNA miRBase Accession #: MIMAT0000088
          miRBase ID: hsa-miR-30a-3p
          Mature miRNA Sequence: CUUUCAGUCGGAUGUUUGCAGC
          Chromosome Location: Chr. 6 - 71403551 - 71403621 [-] on Build GRCh38

          Mouse

          Stem-loop ID mmu-mir-30a
          Stem-loop Accession # MI0000144
          Stem-loop Sequence
          GCGACUGUAAACAUCCUCGACUGGAAGCUGUGAAGCCACAAAUGGGCUUUCAGUCGGAUGUUUGCAGCUGC
          Chromosome Location Chr. 1 - 23272269 - 23272339 [+] on Build GRCm38
          Mature MicroRNA miRBase Accession #: MIMAT0000129
          miRBase ID: mmu-miR-30a-3p
          Mature miRNA Sequence: CUUUCAGUCGGAUGUUUGCAGC
          Chromosome Location: Chr. 1 - 23272269 - 23272339 [+] on Build GRCm38

          Rat

          Stem-loop ID rno-mir-30a
          Stem-loop Accession # MI0000870
          Stem-loop Sequence
          GCAACUGUAAACAUCCUCGACUGGAAGCUGUGAAGCCACAAAUGGGCUUUCAGUCGGAUGUUUGCAGCUGC
          Chromosome Location Chr. 9 - 29542635 - 29542705 [+] on Build Rnor_6.0
          Mature MicroRNA miRBase Accession #: MIMAT0000809
          miRBase ID: rno-miR-30a-3p
          Mature miRNA Sequence: CUUUCAGUCGGAUGUUUGCAGC
          Chromosome Location: Chr. 9 - 29542635 - 29542705 [+] on Build Rnor_6.0

          Alligator mississippiensis

          Stem-loop ID ami-mir-30a
          Stem-loop Accession # MI0029690
          Stem-loop Sequence
          CUGUUGACAGUGAGCGACUGUAAACAUCCUCGACUGGAAGCUGUGAAGCAGCAGAUGGGGCUUUCAGUCGGAUGUUUGCAGCUGCCAACUGCCACAG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0038078
          miRBase ID: ami-miR-30a-3p
          Mature miRNA Sequence: CUUUCAGUCGGAUGUUUGCAGC
          Chromosome Location: NA

          Anolis carolinensis

          Stem-loop ID aca-mir-30a
          Stem-loop Accession # MI0018847
          Stem-loop Sequence
          UGUUGACGGUGAGCGACUGUAAACAUCCUCGACUGGAAGCUGUGAAGCAGUAGAUGAAGCUUUCAGUCGGAUGUUUGCAGCUGCCAACUGCCAC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0021921
          miRBase ID: aca-miR-30a-3p
          Mature miRNA Sequence: CUUUCAGUCGGAUGUUUGCAGC
          Chromosome Location: NA

          Astatotilapia burtoni

          Stem-loop ID abu-mir-30a-1
          Stem-loop Accession # MI0033751
          Stem-loop Sequence
          UGUAAACAUCCUUGACUGGAAGCUGGUGUUAGUUUCUGGGGCUUUCAGUCGGAUGUUUGCAGC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0042058
          miRBase ID: abu-miR-30a-3p
          Mature miRNA Sequence: CUUUCAGUCGGAUGUUUGCAGC
          Chromosome Location: NA

          Stem-loop ID abu-mir-30a-2
          Stem-loop Accession # MI0033752
          Stem-loop Sequence
          UGUAAACAUCCUUGACUGGAAGCUGAGGUUUUGACAGCGAGGGCUUUCAGUCGGAUGUUUGCAGC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0042058
          miRBase ID: abu-miR-30a-3p
          Mature miRNA Sequence: CUUUCAGUCGGAUGUUUGCAGC
          Chromosome Location: NA

          Chicken

          Stem-loop ID gga-mir-30a
          Stem-loop Accession # MI0001204
          Stem-loop Sequence
          GCGACUGUAAACAUCCUCGACUGGAAGCUGUGAAGCAGCAGAUGGGGCUUUCAGUCGGAUGUUUGCAGCUGC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0001136
          miRBase ID: gga-miR-30a-3p
          Mature miRNA Sequence: CUUUCAGUCGGAUGUUUGCAGC
          Chromosome Location: NA

          Chrysemys picta

          Stem-loop ID cpi-mir-30a
          Stem-loop Accession # MI0029424
          Stem-loop Sequence
          GCUGUUGACAGUGAGCGACUGUAAACAUCCUCGACUGGAAGCUGUGAAGCAGCAGAUGGGGCUUUCAGUCGGAUGUUUGCAGCUGCCAACUGCCACAGACU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0037675
          miRBase ID: cpi-miR-30a-3p
          Mature miRNA Sequence: CUUUCAGUCGGAUGUUUGCAGC
          Chromosome Location: NA

          Columba livia

          Stem-loop ID cli-mir-30a
          Stem-loop Accession # MI0029930
          Stem-loop Sequence
          UGAGCGACUGUAAACAUCCUCGACUGGAAGCUGUGAAGCAGCAGAUGGGGCUUUCAGUCGGAUGUUUGCAGCUGCCAA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0038452
          miRBase ID: cli-miR-30a-3p
          Mature miRNA Sequence: CUUUCAGUCGGAUGUUUGCAGC
          Chromosome Location: NA

          Stem-loop ID cli-mir-30e
          Stem-loop Accession # MI0029934
          Stem-loop Sequence
          GUCCUUGCUGCUGUAAACAUCCUUGACUGGAAGCUGUGAGGUGCGAGCAGGAGCUUUCAGUCGGAUGUUUGCAGCAGCAGGCUG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0038459
          miRBase ID: cli-miR-30e-3p
          Mature miRNA Sequence: CUUUCAGUCGGAUGUUUGCAGC
          Chromosome Location: NA

          Cricetulus griseus

          Stem-loop ID cgr-mir-30a
          Stem-loop Accession # MI0020484
          Stem-loop Sequence
          AGUGAGCGACUGUAAACAUCCUCGACUGGAAGCUGUGAAGCCACAAAUGGGCUUUCAGUCGGAUGUUUGCAGCUGCCUACUG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0023895
          miRBase ID: cgr-miR-30a-3p
          Mature miRNA Sequence: CUUUCAGUCGGAUGUUUGCAGC
          Chromosome Location: NA

          Dasypus novemcinctus

          Stem-loop ID dno-mir-30a
          Stem-loop Accession # MI0038934
          Stem-loop Sequence
          UGUAAACAUCCUCGACUGGAAGCUGUGAAGCUGCAGAUGGGCUUUCAGUCGGAUGUUUGCAGC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0047557
          miRBase ID: dno-miR-30a-3p
          Mature miRNA Sequence: CUUUCAGUCGGAUGUUUGCAGC
          Chromosome Location: NA

          Gorilla gorilla

          Stem-loop ID ggo-mir-30a
          Stem-loop Accession # MI0002668
          Stem-loop Sequence
          GCGACUGUAAACAUCCUCGACUGGAAGCUGUGAAGCCACAGAUGGGCUUUCAGUCGGAUGUUUGCAGCUGC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0002374
          miRBase ID: ggo-miR-30a-3p
          Mature miRNA Sequence: CUUUCAGUCGGAUGUUUGCAGC
          Chromosome Location: NA

          Guinea pig

          Stem-loop ID cpo-mir-30a
          Stem-loop Accession # MI0038590
          Stem-loop Sequence
          UGUAAACAUCCUCGACUGGAAGCUGGGAGGCCAGAAUGGGCUUUCAGUCGGAUGUUUGCAGC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0046904
          miRBase ID: cpo-miR-30a-3p
          Mature miRNA Sequence: CUUUCAGUCGGAUGUUUGCAGC
          Chromosome Location: NA

          Ictalurus punctatus

          Stem-loop ID ipu-mir-30e
          Stem-loop Accession # MI0024617
          Stem-loop Sequence
          UGUAAACAUCCUUGACUGGAAGCUGGUGCACAUGAUGGAGCUUUCAGUCGGAUGUUUGCAGC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0029500
          miRBase ID: ipu-miR-30e
          Mature miRNA Sequence: CUUUCAGUCGGAUGUUUGCAGC
          Chromosome Location: NA

          Ophiophagus hannah

          Stem-loop ID oha-mir-30a
          Stem-loop Accession # MI0031450
          Stem-loop Sequence
          GGGAGCGAUUGUAAACAUCCUCGACUGGAAGCUGUGAAGAAGCAGACAGGGCUUUCAGUCGGAUGUUUGCAGCUGCCAACU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0036889
          miRBase ID: oha-miR-30a-3p
          Mature miRNA Sequence: CUUUCAGUCGGAUGUUUGCAGC
          Chromosome Location: NA

          Ornithorhynchus anatinus

          Stem-loop ID oan-mir-30a
          Stem-loop Accession # MI0006729
          Stem-loop Sequence
          GAGUGGAUAUUGGCACUGACAGUGAGCGACUGUAAACAUCCUCGACUGGAAGCUGUGAAGCAGCAGAUGGGGCUUUCAGUCGGAUGUUUGCAGCUGCCUACUGCCUUAGACUUCAAGAAAUGUUAAUAUAGUUGCUU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0006914
          miRBase ID: oan-miR-30a-3p
          Mature miRNA Sequence: CUUUCAGUCGGAUGUUUGCAGC
          Chromosome Location: NA

          Pan paniscus

          Stem-loop ID ppa-mir-30a
          Stem-loop Accession # MI0002670
          Stem-loop Sequence
          GCGACUGUAAACAUCCUCGACUGGAAGCUGUGAAGCCACAGAUGGGCUUUCAGUCGGAUGUUUGCAGCUGC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0002378
          miRBase ID: ppa-miR-30a-3p
          Mature miRNA Sequence: CUUUCAGUCGGAUGUUUGCAGC
          Chromosome Location: NA

          Pan troglodytes

          Stem-loop ID ptr-mir-30a
          Stem-loop Accession # MI0002667
          Stem-loop Sequence
          GCGACUGUAAACAUCCUCGACUGGAAGCUGUGAAGCCACAGAUGGGCUUUCAGUCGGAUGUUUGCAGCUGC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0002372
          miRBase ID: ptr-miR-30a-3p
          Mature miRNA Sequence: CUUUCAGUCGGAUGUUUGCAGC
          Chromosome Location: NA

          Pig

          Stem-loop ID ssc-mir-30a
          Stem-loop Accession # MI0010688
          Stem-loop Sequence
          UCCCUGCGACAGUGAGCGGCUGUAAACAUCCUCGACUGGAAGCUGUGAGGCUGAAGACGGGCUUUCAGUCGGAUGUUUGCAGCCGCCGACUGCCUCCCGCAUCAGGA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0015300
          miRBase ID: ssc-miR-30a-3p
          Mature miRNA Sequence: CUUUCAGUCGGAUGUUUGCAGC
          Chromosome Location: NA

          Pongo pygmaeus

          Stem-loop ID ppy-mir-30a
          Stem-loop Accession # MI0002669
          Stem-loop Sequence
          GCGACUGUAAACAUCCUCGACUGGAAGCUGUGAAGCCACAGAUGGGCUUUCAGUCGGAUGUUUGCAGCUGC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0002376
          miRBase ID: ppy-miR-30a-3p
          Mature miRNA Sequence: CUUUCAGUCGGAUGUUUGCAGC
          Chromosome Location: NA

          Python bivittatus

          Stem-loop ID pbv-mir-30a
          Stem-loop Accession # MI0030183
          Stem-loop Sequence
          CUGUUGACAGUGAGCGAUUGUAAACAUCCUCGACUGGAAGCUGUGAAGCAGCAGAUGGGGCUUUCAGUCGGAUGUUUGCAGCUGCCAACUGCCACAG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0038871
          miRBase ID: pbv-miR-30a-3p
          Mature miRNA Sequence: CUUUCAGUCGGAUGUUUGCAGC
          Chromosome Location: NA

          Rabbit

          Stem-loop ID ocu-mir-30a
          Stem-loop Accession # MI0039247
          Stem-loop Sequence
          UGUAAACAUCCUCGACUGGAAGCUGUGAGGCGCAGACGGGCUUUCAGUCGGAUGUUUGCAGC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0048151
          miRBase ID: ocu-miR-30a-3p
          Mature miRNA Sequence: CUUUCAGUCGGAUGUUUGCAGC
          Chromosome Location: NA

          Rhesus monkey

          Stem-loop ID mml-mir-30a
          Stem-loop Accession # MI0002666
          Stem-loop Sequence
          GCGACUGUAAACAUCCUCGACUGGAAGCUGUGAAGCCAUAGAUGGGCUUUCAGUCGGAUGUUUGCAGCUGC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0002370
          miRBase ID: mml-miR-30a-3p
          Mature miRNA Sequence: CUUUCAGUCGGAUGUUUGCAGC
          Chromosome Location: NA

          Salmo salar

          Stem-loop ID ssa-mir-30c-1
          Stem-loop Accession # MI0026679
          Stem-loop Sequence
          UGUAAACAUCCUUGACUGGAAGCUGGGGUUUGGACAGCAAGGGCUUUCAGUCGGAUGUUUGCAGC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0032607
          miRBase ID: ssa-miR-30c-3p
          Mature miRNA Sequence: CUUUCAGUCGGAUGUUUGCAGC
          Chromosome Location: NA

          Stem-loop ID ssa-mir-30c-2
          Stem-loop Accession # MI0026680
          Stem-loop Sequence
          UGUAAACAUCCUUGACUGGAAGCUGGUGUUCGGUCAGCGAGGGCUUUCAGUCGGAUGUUUGCAGC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0032607
          miRBase ID: ssa-miR-30c-3p
          Mature miRNA Sequence: CUUUCAGUCGGAUGUUUGCAGC
          Chromosome Location: NA

          Stem-loop ID ssa-mir-30d-1
          Stem-loop Accession # MI0026681
          Stem-loop Sequence
          UGUAAACAUCCUUGACUGAAAGCUGGUGUUUCAUACAGGGCUUUCAGUCGGAUGUUUGCAGC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0032609
          miRBase ID: ssa-miR-30d-3p
          Mature miRNA Sequence: CUUUCAGUCGGAUGUUUGCAGC
          Chromosome Location: NA

          Zebrafish

          Stem-loop ID dre-mir-30e-2
          Stem-loop Accession # MI0001950
          Stem-loop Sequence
          UACGGGCUACUGUAAACAUCCUUGACUGGAAGCUGGUGCACAUGAUGGAGCUUUCAGUCGGAUGUUUGCAGCAGCCAACU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0003402
          miRBase ID: dre-miR-30e-3p
          Mature miRNA Sequence: CUUUCAGUCGGAUGUUUGCAGC
          Chromosome Location: NA


          Back To Top

          More Information




          Related Products

          TaqMan™ Pri-miRNA Assay : Rn03465253_pri
          TaqMan™ Pri-miRNA Assay : Mm03306197_pri
          TaqMan™ Pri-miRNA Assay : Hs03302677_pri
          Ambion® Anti-miR™ miRNA Inhibitor : AM10611
          Ambion® Pre-miR™ miRNA Precursor : PM10611
          TaqMan™ MicroRNA Assay : 000416
          mirVana® miRNA inhibitor : MH10611
          mirVana® miRNA mimic : MC10611
          TaqMan™ Advanced miRNA Assay : rno478273_mir
          TaqMan™ Advanced miRNA Assay : mmu478273_mir
          FGMIR : TaqManTM Advanced miRNA Human B 96-well Plate 1
          FGMIR : TaqManTM Advanced miRNA Human B Card
          MIRNA : TaqMan® Advanced miRNA Human A and B Cards
          MIRNA : TaqMan® Advanced miRNA Human A and B 96-well Plates, Standard
          MIRNA : TaqMan® Advanced miRNA Human B Card
          FGMIR : QS TaqmanTM OpenArray Human Advanced miRNA Panel
          MIRNA : TaqMan® OpenArray® Human Advanced MicroRNA Panel
          MIRNA : TaqMan® Advanced miRNA Human B 96-well Plates, Standard
          FGMIR : TaqManTM Advanced miRNA Human B 96-well Plate 1

          Back To Top

          Related Products

          • TaqMan® Advanced miRNA cDNA Synthesis Kit
          • TaqMan® Fast Advanced Master Mix
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-7d94cb4b65-4lm45:80/100.66.75.98:80.
          git-commit: c9e08c96761173abe34e68f880379696776a4827
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.44.1-2026.02.97.1.0