Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › 002404
          Assay Name: hsa-let-7b*
          miRBase Accession Number: MI0000063
          miRBase Version: v22.1
          Chromosome Location: Chr.22: 46113686 - 46113768 [+] on Build GRCh38
          Mature miRNA Sequence: CUAUACAACCUACUGCCUUCCC
          Species: Human, Mouse, Rat, Alligator mississippiensis, Dasypus novemcinctus, Guinea pig, Rhesus monkey
          Product Type: TaqMan™ MicroRNA Assay
          Assay ID 002404
          Size
          Availability Inventoried
          Catalog # 4427975
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Family ID:

          MIPF0000002, let-7

          Mature miRNA Sequence:

          CUAUACAACCUACUGCCUUCCC

          Mature miRNA Details:



          Species miRBase ID miRBase Accession Number
          Human hsa-let-7b-3p MIMAT0004482
          Mouse mmu-let-7b-3p MIMAT0004621
          Rat rno-let-7b-3p MIMAT0004705
          Alligator mississippiensis ami-let-7b-3p MIMAT0038004
          Dasypus novemcinctus dno-let-7b-3p MIMAT0047478
          Guinea pig cpo-let-7b-3p MIMAT0046821
          View More Rows
          Rhesus monkey mml-let-7b-3p MIMAT0026790

          Stem-loop Details



          Human

          Stem-loop ID hsa-let-7b
          Stem-loop Accession # MI0000063
          Stem-loop Sequence
          CGGGGUGAGGUAGUAGGUUGUGUGGUUUCAGGGCAGUGAUGUUGCCCCUCGGAAGAUAACUAUACAACCUACUGCCUUCCCUG
          Chromosome Location Chr. 22 - 46113686 - 46113768 [+] on Build GRCh38
          Mature MicroRNA miRBase Accession #: MIMAT0004482
          miRBase ID: hsa-let-7b-3p
          Mature miRNA Sequence: CUAUACAACCUACUGCCUUCCC
          Chromosome Location: Chr. 22 - 46113686 - 46113768 [+] on Build GRCh38

          Mouse

          Stem-loop ID mmu-let-7b
          Stem-loop Accession # MI0000558
          Stem-loop Sequence
          GCAGGGUGAGGUAGUAGGUUGUGUGGUUUCAGGGCAGUGAUGUUGCCCCUCCGAAGAUAACUAUACAACCUACUGCCUUCCCUGA
          Chromosome Location Chr. 15 - 85707319 - 85707403 [+] on Build GRCm38
          Mature MicroRNA miRBase Accession #: MIMAT0004621
          miRBase ID: mmu-let-7b-3p
          Mature miRNA Sequence: CUAUACAACCUACUGCCUUCCC
          Chromosome Location: Chr. 15 - 85707319 - 85707403 [+] on Build GRCm38

          Rat

          Stem-loop ID rno-let-7b
          Stem-loop Accession # MI0000829
          Stem-loop Sequence
          GCGGGGUGAGGUAGUAGGUUGUGUGGUUUCAGGGCAGUGAUGUCGCCCCUCCGAAGAUAACUAUACAACCUACUGCCUUCCCUGA
          Chromosome Location Chr. 7 - 126590627 - 126590711 [+] on Build Rnor_6.0
          Mature MicroRNA miRBase Accession #: MIMAT0004705
          miRBase ID: rno-let-7b-3p
          Mature miRNA Sequence: CUAUACAACCUACUGCCUUCCC
          Chromosome Location: Chr. 7 - 126590627 - 126590711 [+] on Build Rnor_6.0

          Alligator mississippiensis

          Stem-loop ID ami-let-7b
          Stem-loop Accession # MI0029643
          Stem-loop Sequence
          GCUCUGGCAGGAUGAGGUAGUAGGUUGUGUGGUUUCAGGGUAGUGAUUUUGCCCCAAUCAGGAGAUAACUAUACAACCUACUGCCUUCCCUG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0038004
          miRBase ID: ami-let-7b-3p
          Mature miRNA Sequence: CUAUACAACCUACUGCCUUCCC
          Chromosome Location: NA

          Dasypus novemcinctus

          Stem-loop ID dno-let-7b
          Stem-loop Accession # MI0038888
          Stem-loop Sequence
          UGAGGUAGUAGGUUGUGUGGUUUCAGGGCAGUGAUGUUGCCCUCUCGGAAGAUAACUAUACAACCUACUGCCUUCCC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0047478
          miRBase ID: dno-let-7b-3p
          Mature miRNA Sequence: CUAUACAACCUACUGCCUUCCC
          Chromosome Location: NA

          Guinea pig

          Stem-loop ID cpo-let-7b
          Stem-loop Accession # MI0038543
          Stem-loop Sequence
          UGAGGUAGUAGGUUGUGUGGUUUCCGGGCAGUGAUGCCGCCCCUCGGAAGAUAACUAUACAACCUACUGCCUUCCC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0046821
          miRBase ID: cpo-let-7b-3p
          Mature miRNA Sequence: CUAUACAACCUACUGCCUUCCC
          Chromosome Location: NA

          Rhesus monkey

          Stem-loop ID mml-let-7b
          Stem-loop Accession # MI0007573
          Stem-loop Sequence
          CGGGGUGAGGUAGUAGGUUGUGUGGUUUCAGGGCAGUGAUGUUGCCCCUCAGAAGAUAACUAUACAACCUACUGCCUUCCCUG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0026790
          miRBase ID: mml-let-7b-3p
          Mature miRNA Sequence: CUAUACAACCUACUGCCUUCCC
          Chromosome Location: NA


          Back To Top

          More Information


          Associated Megaplex™ RT Primer Pools:

          Megaplex™ RT Primers, Human Pool B v3.0
          Megaplex™ RT Primers, Rodent Pool B v3.0


          Associated TaqMan® Array MicroRNA Cards:

          TaqMan® Rodent MicroRNA B Cards v3.0
          TaqMan® Human MicroRNA B Cards v3.0


          TaqMan® OpenArray® MicroRNA Panels:

          TaqMan® OpenArray® Human MicroRNA Panel
          TaqMan® OpenArray® Rodent MicroRNA Panel




          Related Products

          TaqMan™ Pri-miRNA Assay : Hs03302548_pri
          TaqMan™ Pri-miRNA Assay : Rn03465224_pri
          TaqMan™ Pri-miRNA Assay : Mm03306756_pri
          Ambion® Anti-miR™ miRNA Inhibitor : AM12489
          Ambion® Pre-miR™ miRNA Precursor : PM12489
          mirVana® miRNA inhibitor : MH12489
          mirVana® miRNA mimic : MC12489
          TaqMan™ Advanced miRNA Assay : 478221_mir
          miRNA Card : TaqMan® Rodent MicroRNA B Cards v2.0
          MIRNA : TaqMan® OpenArray Human MicroRNA Panel
          FGMIR : TaqManTM Human MicroRNA Array B v3
          miRNA Pool : Megaplex™ RT Primers, Human Pool B v3.0
          MIRNA : TaqMan® Array Human MicroRNA A+B Cards Set v3.0
          FGMIR : QS TaqMan OpenArray Rod miRNA Plate
          MIRNA : TaqMan® Array Human MicroRNA B Cards v3.0
          miRNA Card : TaqMan® Human MicroRNA B Cards v2.0
          miRNA Card : TaqMan® Rodent MicroRNA B Cards v3.0
          FGMIR : QS TaqMan OpenArray Hu miRNA Plate
          FGMIR : TaqManTM Array Rodent MicroRNA B Card v3
          MIRNA : TaqMan® OpenArray Rodent MicroRNA Panel
          MIRNA : TaqMan® Array Rodent MicroRNA A+B Cards Set v3.0
          miRNA Card : TaqMan® Human MicroRNA B Cards v3.0
          miRNA Pool : Megaplex™ RT Primers, Rodent Pool B v3.0
          MIRNA : TaqMan® Array Rodent MicroRNA B Cards v3.0


          miRBase Alias:

          Human: hsa-let-7b* (v17)
          Mouse: mmu-let-7b* (v17)
          Rat: rno-let-7b* (v18)

          Back To Top

          Related Products

          • TaqMan® Universal Master Mix II, with UNG
          • TaqMan® Universal Master Mix II, no UNG
          • TaqMan® MicroRNA Reverse Transcription Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Argentina flag icon
          Argentina

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-zsnhf:80/100.66.75.14:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline