Search
Search

| Assay Name: | hsa-let-7b-3p |
|---|---|
| miRBase Accession Number: | MI0000063 |
| miRBase Version: | v22.1 |
| Mature miRNA Sequence: | CUAUACAACCUACUGCCUUCCC |
| Species: | Human, Mouse, Rat, Alligator mississippiensis, Dasypus novemcinctus, Guinea pig, Rhesus monkey |
| Product Type: | mirVana® miRNA mimic |
Gene Family ID: | MIPF0000002, let-7 |
Mature miRNA Sequence: | CUAUACAACCUACUGCCUUCCC |
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Human | hsa-let-7b-3p | MIMAT0004482 |
| Mouse | mmu-let-7b-3p | MIMAT0004621 |
| Rat | rno-let-7b-3p | MIMAT0004705 |
| Alligator mississippiensis | ami-let-7b-3p | MIMAT0038004 |
| Dasypus novemcinctus | dno-let-7b-3p | MIMAT0047478 |
| Guinea pig | cpo-let-7b-3p | MIMAT0046821 |
| View More Rows | ||
| Rhesus monkey | mml-let-7b-3p | MIMAT0026790 |