Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ? Create Account
  • Applications
    • Real-Time PCR
    • Oligos, Primers, Probes and Genes
    • Cloning
    • Protein Biology
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Cell Analysis
    • Mass Spectrometry
    • Chromatography
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Online Order
    • Custom Primers & TaqMan Probes
    • miRNA Mimics & Inhibitors
    • Stealth RNAi / siRNA
    • Silencer Select siRNAs
    • Custom DNA Oligos
    • GeneArt Services
    • GeneArt Strings DNA Fragments
    • TrueGuide CRISPR gRNA
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all online orderable products
  • Services
    • Custom Services
    • Instrument Qualification Services
    • Technical Services
    • Pipette Services
    • Sample Request
    • Instrument Maintenance Services
    • Repairs and Relocation Services
    • OEM and Licensing Services
    • Events, Seminars, Training
    • Unity Lab Services
    • See all services
  • Support
    • Catalogs
    • Application Notes
    • Safety Data Sheets
    • Legal and Regulatory
    • Certificates of Analysis Search
    • Nunc/Nalgene Product Certificates
    • e-learning
    • FAQs
    • Learning Centers
    • Technical Support Centers
    • See all help and support topics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign In
            Sign In
            Don't have an account ? Create Account
            • Connect Your Lab
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › 006193_mat
          Assay Name: cap-miR-29a
          miRBase Accession Number: MI0010057
          miRBase Version: v22.1
          Chromosome Location: -
          Mature miRNA Sequence: UAGCACCAUUUGAAAUCAGUUU
          Species: Capitella teleta, Lottia gigantea, Melibe leonina
          Product Type: TaqMan™ MicroRNA Assay
          Assay ID 006193_mat
          Size
          Availability Made To Order
          Catalog # 4440886
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Family ID:

          MIPF0000009, mir-29

          Mature miRNA Sequence:

          UAGCACCAUUUGAAAUCAGUUU

          Mature miRNA Details:



          Species miRBase ID miRBase Accession Number
          Capitella teleta cte-miR-29a MIMAT0009513
          Lottia gigantea lgi-miR-29 MIMAT0009565
          Melibe leonina mle-miR-29a-3p MIMAT0048719

          Stem-loop Details



          Capitella teleta

          Stem-loop ID cte-mir-29a
          Stem-loop Accession # MI0010057
          Stem-loop Sequence
          AAUGGGGCUCUGGAAUCCUGGUUUUCUCUGGUGCAUAGAUUAACGCUUUGAAUUUGUGUCUAGCACCAUUUGAAAUCAGUUUUCCUGGGUGCUCAC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0009513
          miRBase ID: cte-miR-29a
          Mature miRNA Sequence: UAGCACCAUUUGAAAUCAGUUU
          Chromosome Location: NA

          Lottia gigantea

          Stem-loop ID lgi-mir-29
          Stem-loop Accession # MI0010108
          Stem-loop Sequence
          UUCAGAUAUGUUACCAUUACGGGGUCUGCUUUCAAAUGAUGUAUAGAUUCAUAAUUCCUCUAGCACCAUUUGAAAUCAGUUUCUCAAGAUGGCACACCAAC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0009565
          miRBase ID: lgi-miR-29
          Mature miRNA Sequence: UAGCACCAUUUGAAAUCAGUUU
          Chromosome Location: NA

          Melibe leonina

          Stem-loop ID mle-mir-29a
          Stem-loop Accession # MI0039547
          Stem-loop Sequence
          CCUGAUCUCUCCUGGUGCAUAGAUUUGAUCCCUGCGUCUAGCACCAUUUGAAAUCAGUUU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0048719
          miRBase ID: mle-miR-29a-3p
          Mature miRNA Sequence: UAGCACCAUUUGAAAUCAGUUU
          Chromosome Location: NA


          Back To Top

          More Information




          Related Products

          mirVana® miRNA inhibitor : MH15267
          mirVana® miRNA mimic : MC15267

          Back To Top

          Related Products

          • TaqMan® Universal Master Mix II, with UNG
          • TaqMan® Universal Master Mix II, no UNG
          • TaqMan® MicroRNA Reverse Transcription Kit
          Ordering Plus Icon Minus Icon
          • Online Orderable Products
          • Privacy Policy for Online Ordering
          • Act on Specified Commercial Transactions
          • About Returns
          • About Displayed Prices
          • About Shipping Fees
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Study information disclosure
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Seminars
          • Blog
          About Thermo Fisher Plus Icon Minus Icon
          • Corporate Information
          • Social Responsibility (CSR)
          • Enhancing Customer Experience
          • Careers Careers
          • News
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Japan flag icon
          Japan

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-n9r8p:80/100.66.76.150:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline