Search
Search

| Assay Name: | cte-miR-29a |
|---|---|
| miRBase Accession Number: | MI0010057 |
| miRBase Version: | v22.1 |
| Mature miRNA Sequence: | UAGCACCAUUUGAAAUCAGUUU |
| Species: | Capitella teleta, Lottia gigantea, Melibe leonina |
| Product Type: | mirVana® miRNA inhibitor |
Gene Family ID: | MIPF0000009, mir-29 |
Mature miRNA Sequence: | UAGCACCAUUUGAAAUCAGUUU |
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Capitella teleta | cte-miR-29a | MIMAT0009513 |
| Lottia gigantea | lgi-miR-29 | MIMAT0009565 |
| Melibe leonina | mle-miR-29a-3p | MIMAT0048719 |