Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ? Create Account
  • Applications
    • Real-Time PCR
    • Oligos, Primers, Probes and Genes
    • Cloning
    • Protein Biology
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Cell Analysis
    • Mass Spectrometry
    • Chromatography
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Online Order
    • Custom Primers & TaqMan Probes
    • miRNA Mimics & Inhibitors
    • Stealth RNAi / siRNA
    • Silencer Select siRNAs
    • Custom DNA Oligos
    • GeneArt Services
    • GeneArt Strings DNA Fragments
    • TrueGuide CRISPR gRNA
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all online orderable products
  • Services
    • Custom Services
    • Instrument Qualification Services
    • Technical Services
    • Pipette Services
    • Sample Request
    • Instrument Maintenance Services
    • Repairs and Relocation Services
    • OEM and Licensing Services
    • Events, Seminars, Training
    • Unity Lab Services
    • See all services
  • Support
    • Catalogs
    • Application Notes
    • Safety Data Sheets
    • Legal and Regulatory
    • Certificates of Analysis Search
    • Nunc/Nalgene Product Certificates
    • e-learning
    • FAQs
    • Learning Centers
    • Technical Support Centers
    • See all help and support topics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign In
            Sign In
            Don't have an account ? Create Account
            • Connect Your Lab
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › 245310_mat
          Assay Name: age-miR-29b
          miRBase Accession Number: MI0002730
          miRBase Version: v22.1
          Chromosome Location: -
          Mature miRNA Sequence: UAGCACCAUUUGAAAUCAGU
          Species: Apis mellifera, Ateles geoffroyi, Goat, Gorilla gorilla, Lagothrix lagotricha, Macaca nemestrina, Nasonia giraulti, Nasonia vitripennis, Pan paniscus, Pan troglodytes, Pongo pygmaeus, Saguinus labiatus, Salmo salar
          Product Type: TaqMan™ MicroRNA Assay
          Assay ID 245310_mat
          Size
          Availability Made To Order
          Catalog # 4440886
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Family ID:

          MIPF0000009, mir-29

          Mature miRNA Sequence:

          UAGCACCAUUUGAAAUCAGU

          Mature miRNA Details:



          Species miRBase ID miRBase Accession Number
          Apis mellifera ame-miR-29b-3p MIMAT0004427
          Ateles geoffroyi age-miR-29b MIMAT0002438
          Goat chi-miR-29b-3p MIMAT0036115
          Gorilla gorilla ggo-miR-29b MIMAT0002436
          Lagothrix lagotricha lla-miR-29b MIMAT0002437
          Macaca nemestrina mne-miR-29b MIMAT0002441
          View More Rows
          Nasonia giraulti ngi-miR-29b MIMAT0018401
          Nasonia vitripennis nvi-miR-29b MIMAT0015656
          Pan paniscus ppa-miR-29b MIMAT0002439
          Pan troglodytes ptr-miR-29b MIMAT0002435
          Pongo pygmaeus ppy-miR-29b MIMAT0002434
          Saguinus labiatus sla-miR-29b MIMAT0002440
          Salmo salar ssa-miR-29a-3p MIMAT0032584

          Stem-loop Details



          Apis mellifera

          Stem-loop ID ame-mir-29b
          Stem-loop Accession # MI0005736
          Stem-loop Sequence
          AUUUAAAGACAAUAAGAAGAUAGAGGUACUGACUUCUAUGCGUGCUGGGGUUUGUGCUAAAUCUCCUAGCACCAUUUGAAAUCAGUACUACUCUUCUUAG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0004427
          miRBase ID: ame-miR-29b-3p
          Mature miRNA Sequence: UAGCACCAUUUGAAAUCAGU
          Chromosome Location: NA

          Ateles geoffroyi

          Stem-loop ID age-mir-29b
          Stem-loop Accession # MI0002730
          Stem-loop Sequence
          CUUCAGGAAGCUGGUUUCAUAUGGUGGUUUAGAUUUAAAUAGUGAUUGUCUAGCACCAUUUGAAAUCAGUGUUCUUGGGGG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0002438
          miRBase ID: age-miR-29b
          Mature miRNA Sequence: UAGCACCAUUUGAAAUCAGU
          Chromosome Location: NA

          Goat

          Stem-loop ID chi-mir-29b
          Stem-loop Accession # MI0030727
          Stem-loop Sequence
          CUUCUGGAAGCUGGUUUCACAUGGUGGCUUAGAUUUUUCCAUCUUUGUAUCUAGCACCAUUUGAAAUCAGUGUUUUAGGAGUAA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0036115
          miRBase ID: chi-miR-29b-3p
          Mature miRNA Sequence: UAGCACCAUUUGAAAUCAGU
          Chromosome Location: NA

          Gorilla gorilla

          Stem-loop ID ggo-mir-29b-1
          Stem-loop Accession # MI0002728
          Stem-loop Sequence
          CUUCAGGAAGCUGGUUUCAUAUGGUGGUUUAGAUUUAAAUAGUGAUUGUCUAGCACCAUUUGAAAUCAGUGUUCUUGGGGG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0002436
          miRBase ID: ggo-miR-29b
          Mature miRNA Sequence: UAGCACCAUUUGAAAUCAGU
          Chromosome Location: NA

          Stem-loop ID ggo-mir-29b-2
          Stem-loop Accession # MI0002733
          Stem-loop Sequence
          CUUCUGGAAGCUGGUUUCACAUGGUGGCUUAGAUUUUUCCAUCUUUGUAUCUAGCACCAUUUGAAAUCAGUGUUUUAGGAG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0002436
          miRBase ID: ggo-miR-29b
          Mature miRNA Sequence: UAGCACCAUUUGAAAUCAGU
          Chromosome Location: NA

          Lagothrix lagotricha

          Stem-loop ID lla-mir-29b
          Stem-loop Accession # MI0002729
          Stem-loop Sequence
          CUUCAGGAAGCUGGUUUCAUAUGGUGGUUUAGAUUUAAAUAGUGAUUGUCUAGCACCAUUUGAAAUCAGUGUUCUUGGGGG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0002437
          miRBase ID: lla-miR-29b
          Mature miRNA Sequence: UAGCACCAUUUGAAAUCAGU
          Chromosome Location: NA

          Macaca nemestrina

          Stem-loop ID mne-mir-29b
          Stem-loop Accession # MI0002736
          Stem-loop Sequence
          CUUCUGGAAGCUGGUUUCACAUGGUGGCUUAGAUUUUUCCAUCUUUGUAUCUAGCACCAUUUGAAAUCAGUGUUUUAGGAG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0002441
          miRBase ID: mne-miR-29b
          Mature miRNA Sequence: UAGCACCAUUUGAAAUCAGU
          Chromosome Location: NA

          Nasonia giraulti

          Stem-loop ID ngi-mir-29b
          Stem-loop Accession # MI0016642
          Stem-loop Sequence
          AAGAAUUAAGAUAGCAGAGGUACUGACUUCGGUGCGUGCUGGGGUUACCGAUCAAGCGCCUAGCACCAUUUGAAAUCAGUACUACUCGUCUUAG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0018401
          miRBase ID: ngi-miR-29b
          Mature miRNA Sequence: UAGCACCAUUUGAAAUCAGU
          Chromosome Location: NA

          Nasonia vitripennis

          Stem-loop ID nvi-mir-29b
          Stem-loop Accession # MI0014717
          Stem-loop Sequence
          ACACGAAGAAUUAAGAUAGCAGAGGUACUGACUUCGGUGCGUGCUGGGGUUACCGAUCAAGCGCCUAGCACCAUUUGAAAUCAGUACUACUCGUCUUAG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0015656
          miRBase ID: nvi-miR-29b
          Mature miRNA Sequence: UAGCACCAUUUGAAAUCAGU
          Chromosome Location: NA

          Pan paniscus

          Stem-loop ID ppa-mir-29b-1
          Stem-loop Accession # MI0002731
          Stem-loop Sequence
          CUUCAGGAAGCUGGUUUCAUAUGGUGGUUUAGAUUUAAAUAGUGAUUGUCUAGCACCAUUUGAAAUCAGUGUUCUUGGGGG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0002439
          miRBase ID: ppa-miR-29b
          Mature miRNA Sequence: UAGCACCAUUUGAAAUCAGU
          Chromosome Location: NA

          Stem-loop ID ppa-mir-29b-2
          Stem-loop Accession # MI0002737
          Stem-loop Sequence
          CUUCUGGAAGCUGGUUUCACAUGGUGGCUUAGAUUUUUCCAUCUUUGUAUCUAGCACCAUUUGAAAUCAGUGUUUUAGGAG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0002439
          miRBase ID: ppa-miR-29b
          Mature miRNA Sequence: UAGCACCAUUUGAAAUCAGU
          Chromosome Location: NA

          Pan troglodytes

          Stem-loop ID ptr-mir-29b-1
          Stem-loop Accession # MI0002727
          Stem-loop Sequence
          CUUCAGGAAGCUGGUUUCAUAUGGUGGUUUAGAUUUAAAUAGUGAUUGUCUAGCACCAUUUGAAAUCAGUGUUCUUGGGGG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0002435
          miRBase ID: ptr-miR-29b
          Mature miRNA Sequence: UAGCACCAUUUGAAAUCAGU
          Chromosome Location: NA

          Stem-loop ID ptr-mir-29b-2
          Stem-loop Accession # MI0002732
          Stem-loop Sequence
          CUUCUGGAAGCUGGUUUCACAUGGUGGCUUAGAUUUUUCCAUCUUUGUAUCUAGCACCAUUUGAAAUCAGUGUUUUAGGAG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0002435
          miRBase ID: ptr-miR-29b
          Mature miRNA Sequence: UAGCACCAUUUGAAAUCAGU
          Chromosome Location: NA

          Pongo pygmaeus

          Stem-loop ID ppy-mir-29b-1
          Stem-loop Accession # MI0002726
          Stem-loop Sequence
          CUUCAGGAAGCUGGUUUCAUAUGGUGGUUUAGAUUUAAAUAGUGAUUGUCUAGCACCAUUUGAAAUCAGUGUUCUUGGGGG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0002434
          miRBase ID: ppy-miR-29b
          Mature miRNA Sequence: UAGCACCAUUUGAAAUCAGU
          Chromosome Location: NA

          Stem-loop ID ppy-mir-29b-2
          Stem-loop Accession # MI0002734
          Stem-loop Sequence
          CUUCUGGAAGCUGGUUUCACAUGGUGGCUUAGAUUUUUCCAUCUUUGUAUCUAGCACCAUUUGAAAUCAGUGUUUUAGGAG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0002434
          miRBase ID: ppy-miR-29b
          Mature miRNA Sequence: UAGCACCAUUUGAAAUCAGU
          Chromosome Location: NA

          Saguinus labiatus

          Stem-loop ID sla-mir-29b
          Stem-loop Accession # MI0002735
          Stem-loop Sequence
          CUUCUGGAAGCUGGUUUCACAUGGUGGCUUAGAUUUUUCCAUCUUUGUAUCUAGCACCAUUUGAAAUCAGUGUUUUAGGAG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0002440
          miRBase ID: sla-miR-29b
          Mature miRNA Sequence: UAGCACCAUUUGAAAUCAGU
          Chromosome Location: NA

          Salmo salar

          Stem-loop ID ssa-mir-29a
          Stem-loop Accession # MI0026664
          Stem-loop Sequence
          CUGGUUUCACAUGGUGGUUUAGAUUAAUGUUCCCAGUGUCUAGCACCAUUUGAAAUCAGU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0032584
          miRBase ID: ssa-miR-29a-3p
          Mature miRNA Sequence: UAGCACCAUUUGAAAUCAGU
          Chromosome Location: NA


          Back To Top

          More Information




          Related Products

          mirVana® miRNA inhibitor : MH10218
          mirVana® miRNA mimic : MC10218


          miRBase Alias:

          Anaerococcus mediterraneensis: ame-miR-29b (v21)
          Monodelphis domestica: mdo-miR-29b (v19)
          Staphylococcus schleiferi: ssc-miR-29b (v18)

          Back To Top

          Related Products

          • TaqMan® Universal Master Mix II, with UNG
          • TaqMan® Universal Master Mix II, no UNG
          • TaqMan® MicroRNA Reverse Transcription Kit
          Ordering Plus Icon Minus Icon
          • Online Orderable Products
          • Privacy Policy for Online Ordering
          • Act on Specified Commercial Transactions
          • About Returns
          • About Displayed Prices
          • About Shipping Fees
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Study information disclosure
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Seminars
          • Blog
          About Thermo Fisher Plus Icon Minus Icon
          • Corporate Information
          • Social Responsibility (CSR)
          • Enhancing Customer Experience
          • Careers Careers
          • News
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Japan flag icon
          Japan

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-9fm2r:80/100.66.79.29:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline