Search
Search

| Assay Name: | age-miR-29b |
|---|---|
| miRBase Accession Number: | MI0002730 |
| miRBase Version: | v22.1 |
| Mature miRNA Sequence: | UAGCACCAUUUGAAAUCAGU |
| Species: | Apis mellifera, Ateles geoffroyi, Goat, Gorilla gorilla, Lagothrix lagotricha, Macaca nemestrina, Nasonia giraulti, Nasonia vitripennis, Pan paniscus, Pan troglodytes, Pongo pygmaeus, Saguinus labiatus, Salmo salar |
| Product Type: | mirVana® miRNA inhibitor |
Gene Family ID: | MIPF0000009, mir-29 |
Mature miRNA Sequence: | UAGCACCAUUUGAAAUCAGU |
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Apis mellifera | ame-miR-29b-3p | MIMAT0004427 |
| Ateles geoffroyi | age-miR-29b | MIMAT0002438 |
| Goat | chi-miR-29b-3p | MIMAT0036115 |
| Gorilla gorilla | ggo-miR-29b | MIMAT0002436 |
| Lagothrix lagotricha | lla-miR-29b | MIMAT0002437 |
| Macaca nemestrina | mne-miR-29b | MIMAT0002441 |
| View More Rows | ||
| Nasonia giraulti | ngi-miR-29b | MIMAT0018401 |
| Nasonia vitripennis | nvi-miR-29b | MIMAT0015656 |
| Pan paniscus | ppa-miR-29b | MIMAT0002439 |
| Pan troglodytes | ptr-miR-29b | MIMAT0002435 |
| Pongo pygmaeus | ppy-miR-29b | MIMAT0002434 |
| Saguinus labiatus | sla-miR-29b | MIMAT0002440 |
| Salmo salar | ssa-miR-29a-3p | MIMAT0032584 |