Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ? Create Account
  • Applications
    • Real-Time PCR
    • Oligos, Primers, Probes and Genes
    • Cloning
    • Protein Biology
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Cell Analysis
    • Mass Spectrometry
    • Chromatography
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Online Order
    • Custom Primers & TaqMan Probes
    • miRNA Mimics & Inhibitors
    • Stealth RNAi / siRNA
    • Silencer Select siRNAs
    • Custom DNA Oligos
    • GeneArt Services
    • GeneArt Strings DNA Fragments
    • TrueGuide CRISPR gRNA
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all online orderable products
  • Services
    • Custom Services
    • Instrument Qualification Services
    • Technical Services
    • Pipette Services
    • Sample Request
    • Instrument Maintenance Services
    • Repairs and Relocation Services
    • OEM and Licensing Services
    • Events, Seminars, Training
    • Unity Lab Services
    • See all services
  • Support
    • Catalogs
    • Application Notes
    • Safety Data Sheets
    • Legal and Regulatory
    • Certificates of Analysis Search
    • Nunc/Nalgene Product Certificates
    • e-learning
    • FAQs
    • Learning Centers
    • Technical Support Centers
    • See all help and support topics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign In
            Sign In
            Don't have an account ? Create Account
            • Connect Your Lab
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs04226362_pri
          Assay Name:
          hsa-mir-3649
          Stem-loop ID:
          hsa-mir-3649
          Mapped to miRBase Version:
          v22.1
          Chromosome Location:
          Chr.12: 1660315 - 1660380 [-] on Build GRCh38
          Stem-loop Location:
          Chr.12: 1660315 - 1660380 [-] on Build GRCh38
          Mature miRNA ID:
          hsa-miR-3649
          Species:
          Human
          Product Type:
          TaqMan™ Pri-miRNA Assay
          Assay ID Hs04226362_pri
          Size
          Availability Made To Order
          Catalog # 4427012
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Amplicon Length:

          105

          Chromosome Location:

          Chr. 12 - 1660168 - 1660168 [+] on 1660168 Build GRCh38

          Species:

          Human

          miRBase Details:



          Species miRBase ID miRBase Accession Number
          Human hsa-miR-3649 MIMAT0018069

          Stem-loop Details



          Human

          Stem-loop ID hsa-mir-3649
          Stem-loop Accession # MI0016049
          Stem-loop Sequence
          GCUUGGAACAGGCACCUGUGUGUGCCCAAGUGUUUCUAGCAAACACAGGGACCUGAGUGUCUAAGC
          Chromosome Location Chr. 12 - 1660315 - 1660380 [-] on Build GRCh38
          Mature MicroRNA miRBase Accession #: MIMAT0018069
          miRBase ID: hsa-miR-3649
          Mature miRNA Sequence: AGGGACCUGAGUGUCUAAG
          Chromosome Location: Chr. 12 - 1660315 - 1660380 [-] on Build GRCh38

          Gene Family ID
          Distance to Amplicon 102 bases

          Back To Top

          More Information




          Related Products

          Ambion® Anti-miR™ miRNA Inhibitor : AM20431
          Ambion® Pre-miR™ miRNA Precursor : PM20431
          TaqMan™ MicroRNA Assay : 463578_mat
          mirVana® miRNA inhibitor : MH20431
          mirVana® miRNA mimic : MC20431
          TaqMan™ Advanced miRNA Assay : 480657_mir

          Back To Top

          Related Products

          • High Capacity RNA-to-cDNA Kit
          • SuperScript® IV VILO™ Master Mix
          • TaqMan® Universal PCR Master Mix
          • TaqMan® Fast Advanced Master Mix
          Ordering Plus Icon Minus Icon
          • Online Orderable Products
          • Privacy Policy for Online Ordering
          • Act on Specified Commercial Transactions
          • About Returns
          • About Displayed Prices
          • About Shipping Fees
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Study information disclosure
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Seminars
          • Blog
          About Thermo Fisher Plus Icon Minus Icon
          • Corporate Information
          • Social Responsibility (CSR)
          • Enhancing Customer Experience
          • Careers Careers
          • News
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Japan flag icon
          Japan

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-lnstr:80/100.66.74.145:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline