Search
Search

| Assay Name: | ppt-miR537a |
|---|---|
| miRBase Accession Number: | MI0003508 |
| miRBase Version: | v22.1 |
| Mature miRNA Sequence: | UUGAGGUGUUUCUACAGGCUA |
| Species: | Physcomitrella patens |
| Product Type: | mirVana® miRNA mimic |
Gene Family ID: | MIPF0000207, MIR537 |
Mature miRNA Sequence: | UUGAGGUGUUUCUACAGGCUA |
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Physcomitrella patens | ppt-miR537a | MIMAT0003145 |
| Physcomitrella patens | ppt-miR537b | MIMAT0003146 |
| Physcomitrella patens | ppt-miR537c | MIMAT0005067 |
| Physcomitrella patens | ppt-miR537d | MIMAT0005068 |