Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › MH10329
          Assay Name: ppt-miR537a
          miRBase Accession Number: MI0003508
          miRBase Version: v22.1
          Chromosome Location: -
          Mature miRNA Sequence: UUGAGGUGUUUCUACAGGCUA
          Species: Physcomitrella patens
          Product Type: mirVana® miRNA inhibitor
          Assay ID MH10329

          Purification
          Size
          Availability Made To Order
          Catalog # 4464084
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Family ID:

          MIPF0000207, MIR537

          Mature miRNA Sequence:

          UUGAGGUGUUUCUACAGGCUA

          miRBase Details:



          Species miRBase ID miRBase Accession Number
          Physcomitrella patens ppt-miR537a MIMAT0003145
          Physcomitrella patens ppt-miR537b MIMAT0003146
          Physcomitrella patens ppt-miR537c MIMAT0005067
          Physcomitrella patens ppt-miR537d MIMAT0005068

          Stem-loop Details



          Physcomitrella patens

          Stem-loop ID ppt-MIR537a
          Stem-loop Accession # MI0003508
          Stem-loop Sequence
          GUUGUCGUCAUAUAUGGACUGUAGAAACACCUGAAGCGAGUGAGGACCAUCCGAUUCUAGCCAUGGAAACAGAUAAGAGCUCAUGUCUGGGUUCCUGUUUCUGUGAUGCUUUUGAGGUUUGCGCAUGAGAUAGCAUCGGUUAGGCUUCAAACUCUUGAGGUGUUUCUACAGGCUACAUAUGAUUGAUAGC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0003145
          miRBase ID: ppt-miR537a
          Mature miRNA Sequence: UUGAGGUGUUUCUACAGGCUA
          Chromosome Location: NA

          Stem-loop ID ppt-MIR537b
          Stem-loop Accession # MI0003509
          Stem-loop Sequence
          UCAUAUCUGGACUGUAGAAACACCUGAAGUGACUGAAGUUAGUCCGGUUUAGGUAUGAGAAUGAUUGGAGCACCUCUCCAGAUGUUCCAGAUGUAAUGAAACCGGUGCAAGCUUCAAGCUCUUGAGGUGUUUCUACAGGCUACAUAUGA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0003146
          miRBase ID: ppt-miR537b
          Mature miRNA Sequence: UUGAGGUGUUUCUACAGGCUA
          Chromosome Location: NA

          Stem-loop ID ppt-MIR537c
          Stem-loop Accession # MI0005935
          Stem-loop Sequence
          UCAUAUCUGGACUGUAGAAACACCUGAAGUGACUGAAGUUAGUCCGGUUUAGGUAUGAGAAUGAUUGGAGCACCUCUCCAGAUGUUCCAGAUGUAAUGAAACCGGUGCAAGCUUCAAGCUCUUGAGGUGUUUCUACAGGCUACAUAUGA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0005067
          miRBase ID: ppt-miR537c
          Mature miRNA Sequence: UUGAGGUGUUUCUACAGGCUA
          Chromosome Location: NA

          Stem-loop ID ppt-MIR537d
          Stem-loop Accession # MI0005936
          Stem-loop Sequence
          UUGAACAAUGUCAUUAUCAUACAUAGACUGUAGAAACACCUGAAGCUAUUGAUCUCUACUUUGUUGUAACCGAUCUUUAUGGAAAGUGCACGAAAAUGUGUAUGCUGUGAUCGCACGAUUGGUGACAUCAGUAACUUGAGGUGUUUCUACAGGCUACUUAUGCUUGAUGAUUAUCUUA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0005068
          miRBase ID: ppt-miR537d
          Mature miRNA Sequence: UUGAGGUGUUUCUACAGGCUA
          Chromosome Location: NA


          Back To Top

          More Information




          Related Products

          TaqMan™ MicroRNA Assay : 008288_mat
          mirVana® miRNA mimic : MC10329

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-zpsr4:80/100.66.78.116:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0