Search
Search

| Assay Name: | hsa-miR-127-3p |
|---|---|
| miRBase Accession Number: | MI0000472 |
| miRBase Version: | v22.1 |
| Mature miRNA Sequence: | UCGGAUCCGUCUGAGCUUGGCU |
| Species: | Human, Mouse, Rat, Ateles geoffroyi, Bovine, Cricetulus griseus, Dasypus novemcinctus, Daubentonia madagascariensis, Dog, Guinea pig, Horse, Lagothrix lagotricha, Macaca nemestrina, Nomascus leucogenys, Otolemur garnettii, Pan paniscus, Pan troglodytes, Papio hamadryas, Pig, Pongo pygmaeus, Pteropus alecto, Rabbit, Rhesus monkey, Saguinus labiatus, Saimiri boliviensis, Tupaia chinensis |
| Product Type: | mirVana® miRNA mimic |
Gene Family ID: | MIPF0000080, mir-127 |
Mature miRNA Sequence: | UCGGAUCCGUCUGAGCUUGGCU |
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Human | hsa-miR-127-3p | MIMAT0000446 |
| Mouse | mmu-miR-127-3p | MIMAT0000139 |
| Rat | rno-miR-127-3p | MIMAT0000833 |
| Ateles geoffroyi | age-miR-127 | MIMAT0002287 |
| Bovine | bta-miR-127 | MIMAT0003787 |
| Cricetulus griseus | cgr-miR-127 | MIMAT0041149 |
| View More Rows | ||
| Dasypus novemcinctus | dno-miR-127-3p | MIMAT0047620 |
| Daubentonia madagascariensis | dma-miR-127 | MIMAT0049248 |
| Dog | cfa-miR-127 | MIMAT0006713 |
| Guinea pig | cpo-miR-127-3p | MIMAT0046973 |
| Horse | eca-miR-127 | MIMAT0013126 |
| Lagothrix lagotricha | lla-miR-127 | MIMAT0002285 |
| Macaca nemestrina | mne-miR-127 | MIMAT0002286 |
| Nomascus leucogenys | nle-miR-127 | MIMAT0049373 |
| Otolemur garnettii | oga-miR-127 | MIMAT0049678 |
| Pan paniscus | ppa-miR-127 | MIMAT0049050 |
| Pan troglodytes | ptr-miR-127 | MIMAT0002282 |
| Papio hamadryas | pha-miR-127 | MIMAT0049580 |
| Pig | ssc-miR-127 | MIMAT0013932 |
| Pongo pygmaeus | ppy-miR-127 | MIMAT0002283 |
| Pteropus alecto | pal-miR-127-3p | MIMAT0040147 |
| Rabbit | ocu-miR-127-3p | MIMAT0048209 |
| Rhesus monkey | mml-miR-127-3p | MIMAT0002281 |
| Saguinus labiatus | sla-miR-127 | MIMAT0002284 |
| Saimiri boliviensis | sbo-miR-127 | MIMAT0049479 |
| Tupaia chinensis | tch-miR-127-3p | MIMAT0036559 |