Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › mmu480910_mir

          Specificity testing was performed using mouse anti-targets.

          Assay Name: mmu-miR-127-3p
          Stem-loop Accession Number: MI0000472
          miRBase Version: v22.1
          Chromosome Location: Chr.14: 100882979 - 100883075 [+] on Build GRCh38
          Mature miRNA Sequence: UCGGAUCCGUCUGAGCUUGGCU
          Species: Human, Mouse, Rat, Ateles geoffroyi, Bovine, Cricetulus griseus, Dasypus novemcinctus, Daubentonia madagascariensis, Dog, Guinea pig, Horse, Lagothrix lagotricha, Macaca nemestrina, Nomascus leucogenys, Otolemur garnettii, Pan paniscus, Pan troglodytes, Papio hamadryas, Pig, Pongo pygmaeus, Pteropus alecto, Rabbit, Rhesus monkey, Saguinus labiatus, Saimiri boliviensis, Tupaia chinensis
          Product Type: TaqMan™ Advanced miRNA Assay
          Assay ID mmu480910_mir
          Size S: 250 rxns
          Availability Inventoried
          Catalog # A25576
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Family ID:

          MIPF0000080, mir-127

          Mature miRNA Sequence:

          UCGGAUCCGUCUGAGCUUGGCU

          miRBase Details:



          Species miRBase ID miRBase Accession Number
          Human hsa-miR-127-3p MIMAT0000446
          Mouse mmu-miR-127-3p MIMAT0000139
          Rat rno-miR-127-3p MIMAT0000833
          Ateles geoffroyi age-miR-127 MIMAT0002287
          Bovine bta-miR-127 MIMAT0003787
          Cricetulus griseus cgr-miR-127 MIMAT0041149
          View More Rows
          Dasypus novemcinctus dno-miR-127-3p MIMAT0047620
          Daubentonia madagascariensis dma-miR-127 MIMAT0049248
          Dog cfa-miR-127 MIMAT0006713
          Guinea pig cpo-miR-127-3p MIMAT0046973
          Horse eca-miR-127 MIMAT0013126
          Lagothrix lagotricha lla-miR-127 MIMAT0002285
          Macaca nemestrina mne-miR-127 MIMAT0002286
          Nomascus leucogenys nle-miR-127 MIMAT0049373
          Otolemur garnettii oga-miR-127 MIMAT0049678
          Pan paniscus ppa-miR-127 MIMAT0049050
          Pan troglodytes ptr-miR-127 MIMAT0002282
          Papio hamadryas pha-miR-127 MIMAT0049580
          Pig ssc-miR-127 MIMAT0013932
          Pongo pygmaeus ppy-miR-127 MIMAT0002283
          Pteropus alecto pal-miR-127-3p MIMAT0040147
          Rabbit ocu-miR-127-3p MIMAT0048209
          Rhesus monkey mml-miR-127-3p MIMAT0002281
          Saguinus labiatus sla-miR-127 MIMAT0002284
          Saimiri boliviensis sbo-miR-127 MIMAT0049479
          Tupaia chinensis tch-miR-127-3p MIMAT0036559

          Stem-loop Details



          Human

          Stem-loop ID hsa-mir-127
          Stem-loop Accession # MI0000472
          Stem-loop Sequence
          UGUGAUCACUGUCUCCAGCCUGCUGAAGCUCAGAGGGCUCUGAUUCAGAAAGAUCAUCGGAUCCGUCUGAGCUUGGCUGGUCGGAAGUCUCAUCAUC
          Chromosome Location Chr. 14 - 100882979 - 100883075 [+] on Build GRCh38
          Mature MicroRNA miRBase Accession #: MIMAT0000446
          miRBase ID: hsa-miR-127-3p
          Mature miRNA Sequence: UCGGAUCCGUCUGAGCUUGGCU
          Chromosome Location: Chr. 14 - 100882979 - 100883075 [+] on Build GRCh38

          Mouse

          Stem-loop ID mmu-mir-127
          Stem-loop Accession # MI0000154
          Stem-loop Sequence
          CCAGCCUGCUGAAGCUCAGAGGGCUCUGAUUCAGAAAGAUCAUCGGAUCCGUCUGAGCUUGGCUGGUCGG
          Chromosome Location Chr. 12 - 109592846 - 109592915 [+] on Build GRCm38
          Mature MicroRNA miRBase Accession #: MIMAT0000139
          miRBase ID: mmu-miR-127-3p
          Mature miRNA Sequence: UCGGAUCCGUCUGAGCUUGGCU
          Chromosome Location: Chr. 12 - 109592846 - 109592915 [+] on Build GRCm38

          Rat

          Stem-loop ID rno-mir-127
          Stem-loop Accession # MI0000899
          Stem-loop Sequence
          UUUGAUCACUGUCUCCAGCCUGCUGAAGCUCAGAGGGCUCUGAUUCAGAAAGAUCAUCGGAUCCGUCUGAGCUUGGCUGGUCGGAAGUCUCAUCAUC
          Chromosome Location Chr. 6 - 133713430 - 133713526 [+] on Build Rnor_6.0
          Mature MicroRNA miRBase Accession #: MIMAT0000833
          miRBase ID: rno-miR-127-3p
          Mature miRNA Sequence: UCGGAUCCGUCUGAGCUUGGCU
          Chromosome Location: Chr. 6 - 133713430 - 133713526 [+] on Build Rnor_6.0

          Ateles geoffroyi

          Stem-loop ID age-mir-127
          Stem-loop Accession # MI0002588
          Stem-loop Sequence
          UGUGAUCACUGUCUCCAGCCUGCUGAAGCUCAGAGGGCUCUGAUUCAGAAAGAUCAUCGGAUCCGUCUGAGCUUGGCUGGUCGGAAGUCUCAUCAUC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0002287
          miRBase ID: age-miR-127
          Mature miRNA Sequence: UCGGAUCCGUCUGAGCUUGGCU
          Chromosome Location: NA

          Bovine

          Stem-loop ID bta-mir-127
          Stem-loop Accession # MI0005008
          Stem-loop Sequence
          UGAUCACUGUCUCCAGCCUGCUGAAGCUCAGAGGGCUCUGAUUCAGAAAGAUCAUCGGAUCCGUCUGAGCUUGGCUGGUCGGAAGUCUCCUCAUC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0003787
          miRBase ID: bta-miR-127
          Mature miRNA Sequence: UCGGAUCCGUCUGAGCUUGGCU
          Chromosome Location: NA

          Cricetulus griseus

          Stem-loop ID cgr-mir-127
          Stem-loop Accession # MI0033078
          Stem-loop Sequence
          CCAGCCUGCUGAAGCUCAGAGGGCUCUGAUUCAGAAAGAUCAUCGGAUCCGUCUGAGCUUGGCUGGUCGG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0041149
          miRBase ID: cgr-miR-127
          Mature miRNA Sequence: UCGGAUCCGUCUGAGCUUGGCU
          Chromosome Location: NA

          Dasypus novemcinctus

          Stem-loop ID dno-mir-127
          Stem-loop Accession # MI0038969
          Stem-loop Sequence
          CUGAAGCUCAGAGGGCUCUGAUUCAGAAAGAUCAUCGGAUCCGUCUGAGCUUGGCU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0047620
          miRBase ID: dno-miR-127-3p
          Mature miRNA Sequence: UCGGAUCCGUCUGAGCUUGGCU
          Chromosome Location: NA

          Daubentonia madagascariensis

          Stem-loop ID dma-mir-127
          Stem-loop Accession # MI0039976
          Stem-loop Sequence
          CUGAAGCUCAGAGGGCUCUGAUUCAGAAAGAUCAUCGGAUCCGUCUGAGCUUGGCU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0049248
          miRBase ID: dma-miR-127
          Mature miRNA Sequence: UCGGAUCCGUCUGAGCUUGGCU
          Chromosome Location: NA

          Dog

          Stem-loop ID cfa-mir-127
          Stem-loop Accession # MI0008132
          Stem-loop Sequence
          CCUGCUGAAGCUCAGAGGGCUCUGAUUCAGAAAGAUCAUCGGAUCCGUCUGAGCUUGGCU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0006713
          miRBase ID: cfa-miR-127
          Mature miRNA Sequence: UCGGAUCCGUCUGAGCUUGGCU
          Chromosome Location: NA

          Guinea pig

          Stem-loop ID cpo-mir-127
          Stem-loop Accession # MI0038628
          Stem-loop Sequence
          CUGAAGCUCAGAGGGCUCUGAUUCAGAAAGAUCAUCGGAUCCGUCUGAGCUUGGCU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0046973
          miRBase ID: cpo-miR-127-3p
          Mature miRNA Sequence: UCGGAUCCGUCUGAGCUUGGCU
          Chromosome Location: NA

          Horse

          Stem-loop ID eca-mir-127
          Stem-loop Accession # MI0012876
          Stem-loop Sequence
          CCAGCCUGCUGAAGCUCAGAGGGCUCUGAUUCAGAAAGAUCAUCGGAUCCGUCUGAGCUUGGCUGGUCGG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0013126
          miRBase ID: eca-miR-127
          Mature miRNA Sequence: UCGGAUCCGUCUGAGCUUGGCU
          Chromosome Location: NA

          Lagothrix lagotricha

          Stem-loop ID lla-mir-127
          Stem-loop Accession # MI0002586
          Stem-loop Sequence
          UGUGAUCACUGUCUCCAGCCUGCUGAAGCUCAGAGGGCUCUGAUUCAGAAAGAUCAUCGGAUCCGUCUGAGCUUGGCUGGUCGGAAGUCUCAUCAUC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0002285
          miRBase ID: lla-miR-127
          Mature miRNA Sequence: UCGGAUCCGUCUGAGCUUGGCU
          Chromosome Location: NA

          Macaca nemestrina

          Stem-loop ID mne-mir-127
          Stem-loop Accession # MI0002587
          Stem-loop Sequence
          UGUGAUCACUGUCUCCAGCCUGCUGAAGCUCAGAGGGCUCUGAUUCAGAAAGAUCAUCGGAUCCGUCUGAGCUUGGCUGGUCGGAAGUCUCCUCAUC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0002286
          miRBase ID: mne-miR-127
          Mature miRNA Sequence: UCGGAUCCGUCUGAGCUUGGCU
          Chromosome Location: NA

          Nomascus leucogenys

          Stem-loop ID nle-mir-127
          Stem-loop Accession # MI0040110
          Stem-loop Sequence
          CCUGCUGAAGCUCAGAGGGCUCUGAUUCAGAAAGAUCAUCGGAUCCGUCUGAGCUUGGCU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0049373
          miRBase ID: nle-miR-127
          Mature miRNA Sequence: UCGGAUCCGUCUGAGCUUGGCU
          Chromosome Location: NA

          Otolemur garnettii

          Stem-loop ID oga-mir-127
          Stem-loop Accession # MI0040461
          Stem-loop Sequence
          CCUGCUGAAGCUCAGAGGGCUCUGAUUCAGAAAGAUCAUCGGAUCCGUCUGAGCUUGGCU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0049678
          miRBase ID: oga-miR-127
          Mature miRNA Sequence: UCGGAUCCGUCUGAGCUUGGCU
          Chromosome Location: NA

          Pan paniscus

          Stem-loop ID ppa-mir-127
          Stem-loop Accession # MI0039758
          Stem-loop Sequence
          CUGAAGCUCAGAGGGCUCUGAUUCAGAAAGAUCAUCGGAUCCGUCUGAGCUUGGCU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0049050
          miRBase ID: ppa-miR-127
          Mature miRNA Sequence: UCGGAUCCGUCUGAGCUUGGCU
          Chromosome Location: NA

          Pan troglodytes

          Stem-loop ID ptr-mir-127
          Stem-loop Accession # MI0002583
          Stem-loop Sequence
          UGUGAUCACUGUCUCCAGCCUGCUGAAGCUCAGAGGGCUCUGAUUCAGAAAGAUCAUCGGAUCCGUCUGAGCUUGGCUGGUCGGAAGUCUCAUCAUC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0002282
          miRBase ID: ptr-miR-127
          Mature miRNA Sequence: UCGGAUCCGUCUGAGCUUGGCU
          Chromosome Location: NA

          Papio hamadryas

          Stem-loop ID pha-mir-127
          Stem-loop Accession # MI0040355
          Stem-loop Sequence
          UGAAGCUCAGAGGGCUCUGAUUCAGAAAGAUCAUCGGAUCCGUCUGAGCUUGGCU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0049580
          miRBase ID: pha-miR-127
          Mature miRNA Sequence: UCGGAUCCGUCUGAGCUUGGCU
          Chromosome Location: NA

          Pig

          Stem-loop ID ssc-mir-127
          Stem-loop Accession # MI0013144
          Stem-loop Sequence
          AUCGCUGUCUCCAGCCUGCUGAAGCUCAGAGGGCUCUGAUUCAGAAAGAUCAUCGGAUCCGUCUGAGCUUGGCUGGUCGG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0013932
          miRBase ID: ssc-miR-127
          Mature miRNA Sequence: UCGGAUCCGUCUGAGCUUGGCU
          Chromosome Location: NA

          Pongo pygmaeus

          Stem-loop ID ppy-mir-127
          Stem-loop Accession # MI0002584
          Stem-loop Sequence
          UGUGAUCACUGUCUCCAGCCUGCUGAAGCUCAGAGGGCUCUGAUUCAGAAAGAUCAUCGGAUCCGUCUGAGCUUGGCUGGUCGGAAGUCUCAUCAUC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0002283
          miRBase ID: ppy-miR-127
          Mature miRNA Sequence: UCGGAUCCGUCUGAGCUUGGCU
          Chromosome Location: NA

          Pteropus alecto

          Stem-loop ID pal-mir-127
          Stem-loop Accession # MI0032517
          Stem-loop Sequence
          CUGAAGCUCAGAGGGCUCUGAUUCAGAAAGAUCAUCGGAUCCGUCUGAGCUUGGCU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0040147
          miRBase ID: pal-miR-127-3p
          Mature miRNA Sequence: UCGGAUCCGUCUGAGCUUGGCU
          Chromosome Location: NA

          Rabbit

          Stem-loop ID ocu-mir-127
          Stem-loop Accession # MI0039280
          Stem-loop Sequence
          CUGAAGCUCAGAGGGCUCUGAUUCAGAAAGAUCAUCGGAUCCGUCUGAGCUUGGCU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0048209
          miRBase ID: ocu-miR-127-3p
          Mature miRNA Sequence: UCGGAUCCGUCUGAGCUUGGCU
          Chromosome Location: NA

          Rhesus monkey

          Stem-loop ID mml-mir-127
          Stem-loop Accession # MI0002582
          Stem-loop Sequence
          UGUGAUCACUGUCUCCAGCCUGCUGAAGCUCAGAGGGCUCUGAUUCAGAAAGAUCAUCGGAUCCGUCUGAGCUUGGCUGGUCGGAAGUCUCCUCAUC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0002281
          miRBase ID: mml-miR-127-3p
          Mature miRNA Sequence: UCGGAUCCGUCUGAGCUUGGCU
          Chromosome Location: NA

          Saguinus labiatus

          Stem-loop ID sla-mir-127
          Stem-loop Accession # MI0002585
          Stem-loop Sequence
          UGUGAUCACUGUCUCCAGCCUGCUGAAGCUCAGAGGGCUCUGAUUCAGAAAGAUCAUCGGAUCCGUCUGAGCUUGGCUGGUCGGAAGUCUCAUCAUC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0002284
          miRBase ID: sla-miR-127
          Mature miRNA Sequence: UCGGAUCCGUCUGAGCUUGGCU
          Chromosome Location: NA

          Saimiri boliviensis

          Stem-loop ID sbo-mir-127
          Stem-loop Accession # MI0040236
          Stem-loop Sequence
          CCUGCUGAAGCUCAGAGGGCUCUGAUUCAGAAAGAUCAUCGGAUCCGUCUGAGCUUGGCU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0049479
          miRBase ID: sbo-miR-127
          Mature miRNA Sequence: UCGGAUCCGUCUGAGCUUGGCU
          Chromosome Location: NA

          Tupaia chinensis

          Stem-loop ID tch-mir-127
          Stem-loop Accession # MI0031219
          Stem-loop Sequence
          CUGAAGCUCAGAGGGCUCUGAUUCAGAAAGAUCAUCGGAUCCGUCUGAGCUUGGCU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0036559
          miRBase ID: tch-miR-127-3p
          Mature miRNA Sequence: UCGGAUCCGUCUGAGCUUGGCU
          Chromosome Location: NA


          Back To Top

          More Information




          Related Products

          TaqMan™ Pri-miRNA Assay : Mm03306252_pri
          TaqMan™ Pri-miRNA Assay : Rn03466450_pri
          TaqMan™ Pri-miRNA Assay : Hs03303235_pri
          Ambion® Anti-miR™ miRNA Inhibitor : AM10400
          Ambion® Pre-miR™ miRNA Precursor : PM10400
          TaqMan™ MicroRNA Assay : 000452
          mirVana® miRNA inhibitor : MH10400
          mirVana® miRNA mimic : MC10400
          TaqMan™ Advanced miRNA Assay : 477889_mir
          TaqMan™ Advanced miRNA Assay : rno480910_mir

          Back To Top

          Related Products

          • TaqMan® Advanced miRNA cDNA Synthesis Kit
          • TaqMan® Fast Advanced Master Mix
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-vzwvz:80/100.66.79.173:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline