Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › MC10678
          Assay Name: aca-miR-29a-3p
          miRBase Accession Number: MI0018840
          miRBase Version: v22.1
          Chromosome Location: -
          Mature miRNA Sequence: UAGCACCAUUUGAAAUCGGUU
          Species: Anolis carolinensis, Callithrix jacchus, Chicken, Cricetulus griseus, Monodelphis domestica, Ornithorhynchus anatinus, Taeniopygia guttata, Tupaia chinensis, Xenopus tropicalis
          Product Type: mirVana® miRNA mimic
          Assay ID MC10678

          Purification
          Size
          Availability Made To Order
          Catalog # 4464066
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Family ID:

          MIPF0000009, mir-29

          Mature miRNA Sequence:

          UAGCACCAUUUGAAAUCGGUU

          miRBase Details:



          Species miRBase ID miRBase Accession Number
          Anolis carolinensis aca-miR-29a-3p MIMAT0021912
          Callithrix jacchus cja-miR-29c MIMAT0039438
          Chicken gga-miR-29a-3p MIMAT0001096
          Cricetulus griseus cgr-miR-29c-3p MIMAT0023888
          Monodelphis domestica mdo-miR-29a-3p MIMAT0004164
          Ornithorhynchus anatinus oan-miR-29a-3p MIMAT0006807
          View More Rows
          Taeniopygia guttata tgu-miR-29a-3p MIMAT0014532
          Tupaia chinensis tch-miR-29c-3p MIMAT0036546
          Xenopus tropicalis xtr-miR-29a MIMAT0003656

          Stem-loop Details



          Anolis carolinensis

          Stem-loop ID aca-mir-29a-1
          Stem-loop Accession # MI0018840
          Stem-loop Sequence
          GUCACCCCUUUAAAGGAUGACUGAUUUCUUCUGGUGUUCGGAGUCUAGUUCUCUUUCUAGCACCAUUUGAAAUCGGUUAUAGUGAUUGGGGAAUAUC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0021912
          miRBase ID: aca-miR-29a-3p
          Mature miRNA Sequence: UAGCACCAUUUGAAAUCGGUU
          Chromosome Location: NA

          Stem-loop ID aca-mir-29a-2
          Stem-loop Accession # MI0018841
          Stem-loop Sequence
          CGUCUCUUACACAGGCUGACCGAUUUCUCUUGGUGUUCAGAGUCUUAGUUUUUGUCUAGCACCAUUUGAAAUCGGUUAUGAUGUAGGGGGAAAAGC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0021912
          miRBase ID: aca-miR-29a-3p
          Mature miRNA Sequence: UAGCACCAUUUGAAAUCGGUU
          Chromosome Location: NA

          Callithrix jacchus

          Stem-loop ID cja-mir-29c
          Stem-loop Accession # MI0031960
          Stem-loop Sequence
          UCUCUUACACAGGCUGACCGAUUUCUCCUGGUGUUCAGAGUCUGUUUUUGUCUAGCACCAUUUGAAAUCGGUUAUGAUGUAGGGGG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0039438
          miRBase ID: cja-miR-29c
          Mature miRNA Sequence: UAGCACCAUUUGAAAUCGGUU
          Chromosome Location: NA

          Chicken

          Stem-loop ID gga-mir-29a
          Stem-loop Accession # MI0001166
          Stem-loop Sequence
          ACCCCUUUAGAGGAUGACUGAUUUCUUUUGGUGUUCAGAGUCAAUAAUAUUUUCUAGCACCAUUUGAAAUCGGUUAUAGUGAUUGGGGA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0001096
          miRBase ID: gga-miR-29a-3p
          Mature miRNA Sequence: UAGCACCAUUUGAAAUCGGUU
          Chromosome Location: NA

          Cricetulus griseus

          Stem-loop ID cgr-mir-29c
          Stem-loop Accession # MI0020480
          Stem-loop Sequence
          CACAGGCUGACCGAUUUCUCCUGGUGUUCAGAGUCUGUUUUUGUCUAGCACCAUUUGAAAUCGGUUAUGAUGUAGG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0023888
          miRBase ID: cgr-miR-29c-3p
          Mature miRNA Sequence: UAGCACCAUUUGAAAUCGGUU
          Chromosome Location: NA

          Monodelphis domestica

          Stem-loop ID mdo-mir-29a-1
          Stem-loop Accession # MI0005353
          Stem-loop Sequence
          AUGACUGAUUUCUUUUGGUGUUCAGAGUCAAUAUCAUUUUCUAGCACCAUUUGAAAUCGGUUAU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0004164
          miRBase ID: mdo-miR-29a-3p
          Mature miRNA Sequence: UAGCACCAUUUGAAAUCGGUU
          Chromosome Location: NA

          Stem-loop ID mdo-mir-29a-2
          Stem-loop Accession # MI0023759
          Stem-loop Sequence
          CCGAUUUCUCUUGGUGUUCAGAGUCUGUUUUUGUCUAGCACCAUUUGAAAUCGGUU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0004164
          miRBase ID: mdo-miR-29a-3p
          Mature miRNA Sequence: UAGCACCAUUUGAAAUCGGUU
          Chromosome Location: NA

          Ornithorhynchus anatinus

          Stem-loop ID oan-mir-29a-1
          Stem-loop Accession # MI0006668
          Stem-loop Sequence
          UCAAAGGCUCCUGUGACCCCUCUAGAGGAUGACUGAUUUCUUUUGGUGUUCAGAGUCAAUAUCAUUUUCUAGCACCAUUUGAAAUCGGUUAUAAUGAUUGGGGAAUAGCGCCACGUGA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0006807
          miRBase ID: oan-miR-29a-3p
          Mature miRNA Sequence: UAGCACCAUUUGAAAUCGGUU
          Chromosome Location: NA

          Stem-loop ID oan-mir-29a-2
          Stem-loop Accession # MI0006702
          Stem-loop Sequence
          CAGGACCCAUCUCUUACACAGGCUGACCGAUUUCUCUUGGUGUUCAGAGUCUCAGUUCUUGUCUAGCACCAUUUGAAAUCGGUUAUGAUGUAGGGGGAAAAGCAGCAGUCCUG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0006807
          miRBase ID: oan-miR-29a-3p
          Mature miRNA Sequence: UAGCACCAUUUGAAAUCGGUU
          Chromosome Location: NA

          Taeniopygia guttata

          Stem-loop ID tgu-mir-29a-1
          Stem-loop Accession # MI0013745
          Stem-loop Sequence
          UGACUGAUUUCUUUUGGUGUUCAGAGUCAAUAAUAAUUUCUAGCACCAUUUGAAAUCGGUUAUAGUGAUUG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0014532
          miRBase ID: tgu-miR-29a-3p
          Mature miRNA Sequence: UAGCACCAUUUGAAAUCGGUU
          Chromosome Location: NA

          Stem-loop ID tgu-mir-29a-2
          Stem-loop Accession # MI0013746
          Stem-loop Sequence
          UGACCGAUUUCUCUUGGUGUUCAGAGUCUCAGUUUUUGUCUAGCACCAUUUGAAAUCGGUUAUGAUGUAGG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0014532
          miRBase ID: tgu-miR-29a-3p
          Mature miRNA Sequence: UAGCACCAUUUGAAAUCGGUU
          Chromosome Location: NA

          Tupaia chinensis

          Stem-loop ID tch-mir-29c
          Stem-loop Accession # MI0031206
          Stem-loop Sequence
          ACCGAUUUCUCCUGGUGUUCAGAGUCUGUUUUUGUCUAGCACCAUUUGAAAUCGGUU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0036546
          miRBase ID: tch-miR-29c-3p
          Mature miRNA Sequence: UAGCACCAUUUGAAAUCGGUU
          Chromosome Location: NA

          Xenopus tropicalis

          Stem-loop ID xtr-mir-29a
          Stem-loop Accession # MI0004897
          Stem-loop Sequence
          ACCCUUUUAAAGGAUGACUGAUUUCUUCUGGUGUUCAGAGUCUUUUGUUUUCUAGCACCAUUUGAAAUCGGUUAUAAUGAUGGGGUA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0003656
          miRBase ID: xtr-miR-29a
          Mature miRNA Sequence: UAGCACCAUUUGAAAUCGGUU
          Chromosome Location: NA


          Back To Top

          More Information




          Related Products

          TaqMan™ MicroRNA Assay : 004471_mat
          mirVana® miRNA inhibitor : MH10678


          miRBase Alias:

          Aeromonas caviae Ae398: aca-miR-29a (v19)
          Gallus gallus: gga-miR-29a (v19)
          Ochrobactrum anthropi: oan-miR-29a (v19)
          Trabulsiella guamensis ATCC 49490: tgu-miR-29a (v19)

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          France flag icon
          France

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-hznln:80/100.66.79.29:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline